Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130138125-130138688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM1
Entrez Gene ID
9131
Gene Name
apoptosis inducing factor, mitochondria associated 1
Gene Aliases
AIF, CMT2D, CMTX4, COWCK, COXPD6, DFNX5, NADMR, NAMSD, PDCD8
UniGene
Hs.424932
Gene Chromosome Location
Chr.X: 130129362-130165887 on Build GRCh38
Transcripts
NM_001130847.3 NM_004208.3 NM_145812.2 NR_132647.1 XM_017029963.1

Gene Symbol
RAB33A
Entrez Gene ID
9363
Gene Name
RAB33A, member RAS oncogene family
Gene Aliases
RabS10
UniGene
Hs.654356
Gene Chromosome Location
Chr.X: 130110633-130184872 on Build GRCh38
Transcripts
NM_001130847.3 NM_004208.3 NM_145812.2 NR_132647.1 XM_017029963.1

SNP Detail

SNP ID Chromosome Location
rs746271276 Chr.X: 130138247-130138247 on Build GRCh38
rs965690705 Chr.X: 130138526-130138526 on Build GRCh38
rs200114054 Chr.X: 130138649-130138649 on Build GRCh38
rs1009987927 Chr.X: 130138478-130138478 on Build GRCh38
rs758272982 Chr.X: 130138470-130138470 on Build GRCh38
rs148184613 Chr.X: 130138612-130138612 on Build GRCh38
rs965742909 Chr.X: 130138393-130138393 on Build GRCh38
rs1032739985 Chr.X: 130138344-130138344 on Build GRCh38
rs1005323571 Chr.X: 130138187-130138187 on Build GRCh38
rs948088391 Chr.X: 130138422-130138422 on Build GRCh38
rs752868065 Chr.X: 130138281-130138281 on Build GRCh38
rs74543749 Chr.X: 130138464-130138464 on Build GRCh38
rs960961537 Chr.X: 130138193-130138193 on Build GRCh38
rs767600125 Chr.X: 130138272-130138272 on Build GRCh38
rs1012453487 Chr.X: 130138322-130138322 on Build GRCh38
rs768118991 Chr.X: 130138621-130138621 on Build GRCh38
rs190558459 Chr.X: 130138460-130138460 on Build GRCh38
rs762537799 Chr.X: 130138419-130138419 on Build GRCh38
rs1045198869 Chr.X: 130138398-130138398 on Build GRCh38
rs921876086 Chr.X: 130138396-130138396 on Build GRCh38
rs907921544 Chr.X: 130138413-130138413 on Build GRCh38
rs929568092 Chr.X: 130138373-130138373 on Build GRCh38
rs955996950 Chr.X: 130138345-130138345 on Build GRCh38
rs965381749 Chr.X: 130138204-130138204 on Build GRCh38
rs369809173 Chr.X: 130138548-130138548 on Build GRCh38
rs976071788 Chr.X: 130138328-130138328 on Build GRCh38
rs775202035 Chr.X: 130138584-130138584 on Build GRCh38
rs759865484 Chr.X: 130138262-130138262 on Build GRCh38
rs907702631 Chr.X: 130138648-130138648 on Build GRCh38
rs1005712988 Chr.X: 130138244-130138244 on Build GRCh38
rs533350620 Chr.X: 130138241-130138241 on Build GRCh38
rs776589330 Chr.X: 130138560-130138560 on Build GRCh38
rs886435770 Chr.X: 130138169-130138169 on Build GRCh38
rs956500445 Chr.X: 130138166-130138166 on Build GRCh38
rs768793589 Chr.X: 130138267-130138267 on Build GRCh38
rs121965089 Chr.X: 130138622-130138622 on Build GRCh38
rs770815681 Chr.X: 130138550-130138550 on Build GRCh38
rs887939676 Chr.X: 130138389-130138389 on Build GRCh38
rs200686111 Chr.X: 130138474-130138474 on Build GRCh38
rs769856084 Chr.X: 130138248-130138248 on Build GRCh38
rs896487849 Chr.X: 130138421-130138421 on Build GRCh38
rs1023879712 Chr.X: 130138337-130138337 on Build GRCh38
rs190213794 Chr.X: 130138293-130138293 on Build GRCh38
rs372764527 Chr.X: 130138397-130138397 on Build GRCh38
rs1034694377 Chr.X: 130138471-130138474 on Build GRCh38
rs751119352 Chr.X: 130138452-130138452 on Build GRCh38
rs186284723 Chr.X: 130138420-130138420 on Build GRCh38
rs12014115 Chr.X: 130138642-130138642 on Build GRCh38
rs1045277210 Chr.X: 130138443-130138443 on Build GRCh38
rs756539240 Chr.X: 130138477-130138477 on Build GRCh38
rs1018626226 Chr.X: 130138382-130138382 on Build GRCh38
rs993597687 Chr.X: 130138205-130138205 on Build GRCh38
rs12014323 Chr.X: 130138545-130138545 on Build GRCh38
rs1012337816 Chr.X: 130138208-130138208 on Build GRCh38
rs776250139 Chr.X: 130138309-130138309 on Build GRCh38
rs1028888246 Chr.X: 130138549-130138549 on Build GRCh38
rs925727192 Chr.X: 130138240-130138240 on Build GRCh38
rs12010123 Chr.X: 130138308-130138308 on Build GRCh38
rs112432216 Chr.X: 130138282-130138282 on Build GRCh38
rs944680505 Chr.X: 130138323-130138323 on Build GRCh38
rs768366540 Chr.X: 130138596-130138596 on Build GRCh38
rs773680831 Chr.X: 130138608-130138608 on Build GRCh38
rs762443189 Chr.X: 130138594-130138594 on Build GRCh38
rs751927092 Chr.X: 130138251-130138251 on Build GRCh38
rs1035214940 Chr.X: 130138191-130138191 on Build GRCh38
rs915293063 Chr.X: 130138181-130138181 on Build GRCh38
rs773123094 Chr.X: 130138268-130138268 on Build GRCh38
rs1018740701 Chr.X: 130138490-130138490 on Build GRCh38
rs1049352137 Chr.X: 130138381-130138381 on Build GRCh38
rs1022541142 Chr.X: 130138289-130138289 on Build GRCh38
rs12010190 Chr.X: 130138466-130138466 on Build GRCh38
rs540630620 Chr.X: 130138236-130138236 on Build GRCh38
rs1023764906 Chr.X: 130138221-130138221 on Build GRCh38
rs1003237214 Chr.X: 130138259-130138259 on Build GRCh38
rs182805215 Chr.X: 130138474-130138474 on Build GRCh38
rs769632650 Chr.X: 130138578-130138578 on Build GRCh38
rs1038847509 Chr.X: 130138265-130138265 on Build GRCh38
rs974651871 Chr.X: 130138394-130138394 on Build GRCh38
rs752439789 Chr.X: 130138405-130138405 on Build GRCh38
rs111657175 Chr.X: 130138592-130138592 on Build GRCh38
rs1034914377 Chr.X: 130138261-130138261 on Build GRCh38
rs757796129 Chr.X: 130138232-130138232 on Build GRCh38
rs181905756 Chr.X: 130138341-130138341 on Build GRCh38
rs745640514 Chr.X: 130138562-130138562 on Build GRCh38
rs187385111 Chr.X: 130138276-130138276 on Build GRCh38
rs987983447 Chr.X: 130138180-130138180 on Build GRCh38
rs1040586653 Chr.X: 130138202-130138202 on Build GRCh38
rs1001469686 Chr.X: 130138342-130138342 on Build GRCh38
rs988442973 Chr.X: 130138361-130138361 on Build GRCh38

View Full Product Details