Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2904417-2904958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD1
Entrez Gene ID
118
Gene Name
adducin 1
Gene Aliases
ADDA
UniGene
Hs.183706
Gene Chromosome Location
Chr.4: 2843732-2930076 on Build GRCh38
Transcripts
NM_001119.4 NM_001286645.1 NM_014189.3 NM_014190.3 NM_176801.2 XM_005247933.2 XM_005247934.2 XM_005247935.2 XM_005247936.2 XM_005247937.2 XM_017007703.1 XM_017007704.1 XM_017007705.1 XM_017007706.1 XM_017007707.1 XM_017007708.1 XM_017007709.1 XM_017007710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660423 c.1285A>G p.T429A Chr.4: 2904887-2904887 on Build GRCh38
1694092 c.1255T>G p.Y419D Chr.4: 2904857-2904857 on Build GRCh38
4124292 c.1179C>T p.Y393Y Chr.4: 2904781-2904781 on Build GRCh38
1670864 c.1225G>A p.D409N Chr.4: 2904827-2904827 on Build GRCh38
5866279 c.1163G>T p.G388V Chr.4: 2904765-2904765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026362838 Chr.4: 2904759-2904759 on Build GRCh38
rs535777554 Chr.4: 2904501-2904501 on Build GRCh38
rs975086345 Chr.4: 2904826-2904826 on Build GRCh38
rs974465966 Chr.4: 2904556-2904556 on Build GRCh38
rs190509107 Chr.4: 2904938-2904938 on Build GRCh38
rs2285084 Chr.4: 2904558-2904558 on Build GRCh38
rs764711805 Chr.4: 2904832-2904832 on Build GRCh38
rs754258917 Chr.4: 2904747-2904747 on Build GRCh38
rs151011491 Chr.4: 2904624-2904624 on Build GRCh38
rs968262869 Chr.4: 2904731-2904731 on Build GRCh38
rs761612342 Chr.4: 2904795-2904795 on Build GRCh38
rs367978307 Chr.4: 2904883-2904883 on Build GRCh38
rs749096841 Chr.4: 2904936-2904936 on Build GRCh38
rs377019341 Chr.4: 2904757-2904757 on Build GRCh38
rs1055353422 Chr.4: 2904676-2904676 on Build GRCh38
rs762870470 Chr.4: 2904882-2904882 on Build GRCh38
rs775795547 Chr.4: 2904538-2904538 on Build GRCh38
rs751088506 Chr.4: 2904829-2904829 on Build GRCh38
rs543430304 Chr.4: 2904636-2904636 on Build GRCh38
rs762555520 Chr.4: 2904825-2904825 on Build GRCh38
rs375585792 Chr.4: 2904735-2904735 on Build GRCh38
rs376166483 Chr.4: 2904764-2904764 on Build GRCh38
rs34225447 Chr.4: 2904933-2904933 on Build GRCh38
rs140908749 Chr.4: 2904653-2904653 on Build GRCh38
rs748041609 Chr.4: 2904894-2904894 on Build GRCh38
rs866317695 Chr.4: 2904788-2904788 on Build GRCh38
rs747095109 Chr.4: 2904787-2904787 on Build GRCh38
rs780044563 Chr.4: 2904782-2904782 on Build GRCh38
rs765558070 Chr.4: 2904846-2904846 on Build GRCh38
rs776488630 Chr.4: 2904794-2904794 on Build GRCh38
rs984553548 Chr.4: 2904506-2904506 on Build GRCh38
rs780314941 Chr.4: 2904874-2904874 on Build GRCh38
rs1006047195 Chr.4: 2904773-2904773 on Build GRCh38
rs932980066 Chr.4: 2904607-2904607 on Build GRCh38
rs1038499573 Chr.4: 2904662-2904662 on Build GRCh38
rs778412779 Chr.4: 2904745-2904745 on Build GRCh38
rs960850684 Chr.4: 2904449-2904449 on Build GRCh38
rs145634840 Chr.4: 2904667-2904667 on Build GRCh38
rs750774758 Chr.4: 2904855-2904855 on Build GRCh38
rs904129400 Chr.4: 2904658-2904658 on Build GRCh38
rs36115602 Chr.4: 2904898-2904898 on Build GRCh38
rs913024832 Chr.4: 2904926-2904926 on Build GRCh38
rs896495825 Chr.4: 2904692-2904692 on Build GRCh38
rs142283650 Chr.4: 2904789-2904789 on Build GRCh38
rs923500764 Chr.4: 2904584-2904584 on Build GRCh38
rs572253145 Chr.4: 2904517-2904517 on Build GRCh38
rs555353861 Chr.4: 2904514-2904514 on Build GRCh38
rs374060610 Chr.4: 2904730-2904730 on Build GRCh38
rs749988256 Chr.4: 2904725-2904725 on Build GRCh38
rs1014098568 Chr.4: 2904484-2904484 on Build GRCh38
rs758602816 Chr.4: 2904868-2904868 on Build GRCh38
rs2239728 Chr.4: 2904726-2904726 on Build GRCh38
rs756734723 Chr.4: 2904739-2904739 on Build GRCh38
rs777519065 Chr.4: 2904737-2904740 on Build GRCh38
rs746035251 Chr.4: 2904769-2904769 on Build GRCh38
rs6833874 Chr.4: 2904655-2904655 on Build GRCh38
rs772304282 Chr.4: 2904775-2904775 on Build GRCh38
rs777173423 Chr.4: 2904572-2904572 on Build GRCh38
rs537655601 Chr.4: 2904915-2904915 on Build GRCh38
rs373130926 Chr.4: 2904714-2904714 on Build GRCh38
rs138254700 Chr.4: 2904796-2904796 on Build GRCh38
rs140440262 Chr.4: 2904909-2904909 on Build GRCh38
rs904970101 Chr.4: 2904663-2904663 on Build GRCh38
rs769689766 Chr.4: 2904899-2904899 on Build GRCh38
rs373803874 Chr.4: 2904835-2904835 on Build GRCh38
rs772950775 Chr.4: 2904810-2904810 on Build GRCh38
rs376233271 Chr.4: 2904724-2904724 on Build GRCh38
rs766033361 Chr.4: 2904827-2904827 on Build GRCh38
rs184957456 Chr.4: 2904545-2904545 on Build GRCh38
rs781132961 Chr.4: 2904888-2904888 on Build GRCh38
rs180688431 Chr.4: 2904448-2904448 on Build GRCh38
rs1026332705 Chr.4: 2904497-2904497 on Build GRCh38
rs768650483 Chr.4: 2904792-2904792 on Build GRCh38
rs747100756 Chr.4: 2904875-2904875 on Build GRCh38
rs774524748 Chr.4: 2904548-2904548 on Build GRCh38

View Full Product Details