Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 111075823-111076310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSLP
Entrez Gene ID
85480
Gene Name
thymic stromal lymphopoietin
Gene Aliases
-
UniGene
Hs.389874
Gene Chromosome Location
Chr.5: 111070080-111078024 on Build GRCh38
Transcripts
NM_033035.4 NM_138551.4 NR_045089.1 XM_011543698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
448421 c.412C>A p.L138M Chr.5: 111076006-111076006 on Build GRCh38
4736946 c.384delA p.R130fs*27 Chr.5: 111075978-111075978 on Build GRCh38
6038832 c.405T>A p.N135K Chr.5: 111075999-111075999 on Build GRCh38
4127061 c.459A>T p.R153R Chr.5: 111076053-111076053 on Build GRCh38
3719285 c.380G>A p.R127K Chr.5: 111075974-111075974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947571928 Chr.5: 111075863-111075863 on Build GRCh38
rs900082873 Chr.5: 111076257-111076257 on Build GRCh38
rs374842033 Chr.5: 111076043-111076043 on Build GRCh38
rs760066885 Chr.5: 111075945-111075945 on Build GRCh38
rs1050015808 Chr.5: 111076271-111076271 on Build GRCh38
rs901538347 Chr.5: 111075917-111075917 on Build GRCh38
rs777070141 Chr.5: 111075943-111075943 on Build GRCh38
rs866271073 Chr.5: 111075865-111075865 on Build GRCh38
rs187411102 Chr.5: 111076173-111076173 on Build GRCh38
rs550711691 Chr.5: 111076080-111076080 on Build GRCh38
rs1041322758 Chr.5: 111076251-111076251 on Build GRCh38
rs377076491 Chr.5: 111076208-111076208 on Build GRCh38
rs77769254 Chr.5: 111075891-111075891 on Build GRCh38
rs531054750 Chr.5: 111076177-111076177 on Build GRCh38
rs751225469 Chr.5: 111076114-111076114 on Build GRCh38
rs182364097 Chr.5: 111075946-111075946 on Build GRCh38
rs200237930 Chr.5: 111076107-111076107 on Build GRCh38
rs987651931 Chr.5: 111075985-111075985 on Build GRCh38
rs571481535 Chr.5: 111076261-111076261 on Build GRCh38
rs766974904 Chr.5: 111076152-111076152 on Build GRCh38
rs372152752 Chr.5: 111076012-111076012 on Build GRCh38
rs775918286 Chr.5: 111076147-111076148 on Build GRCh38
rs893206023 Chr.5: 111075855-111075855 on Build GRCh38
rs369561427 Chr.5: 111076093-111076093 on Build GRCh38
rs374398822 Chr.5: 111076049-111076049 on Build GRCh38
rs749927156 Chr.5: 111076188-111076188 on Build GRCh38
rs755633541 Chr.5: 111076234-111076234 on Build GRCh38
rs773370104 Chr.5: 111075928-111075928 on Build GRCh38
rs750095133 Chr.5: 111075909-111075909 on Build GRCh38
rs770269923 Chr.5: 111076071-111076071 on Build GRCh38
rs914668990 Chr.5: 111076164-111076164 on Build GRCh38
rs11466747 Chr.5: 111075996-111075996 on Build GRCh38
rs188293283 Chr.5: 111075906-111075906 on Build GRCh38
rs756093192 Chr.5: 111076023-111076023 on Build GRCh38
rs572018138 Chr.5: 111075850-111075850 on Build GRCh38
rs763312329 Chr.5: 111076078-111076078 on Build GRCh38
rs377757437 Chr.5: 111076092-111076092 on Build GRCh38
rs777991475 Chr.5: 111076042-111076042 on Build GRCh38
rs928527628 Chr.5: 111076106-111076106 on Build GRCh38
rs1031064440 Chr.5: 111075873-111075873 on Build GRCh38
rs150474294 Chr.5: 111076040-111076040 on Build GRCh38
rs369166421 Chr.5: 111076003-111076003 on Build GRCh38
rs947415384 Chr.5: 111076184-111076184 on Build GRCh38
rs765773773 Chr.5: 111075982-111075982 on Build GRCh38
rs763283488 Chr.5: 111075998-111075998 on Build GRCh38
rs796748481 Chr.5: 111076264-111076264 on Build GRCh38
rs372067633 Chr.5: 111075934-111075934 on Build GRCh38
rs552538362 Chr.5: 111076122-111076122 on Build GRCh38
rs989449618 Chr.5: 111075903-111075903 on Build GRCh38
rs11466746 Chr.5: 111075858-111075858 on Build GRCh38
rs1046716837 Chr.5: 111076254-111076254 on Build GRCh38
rs756896201 Chr.5: 111075897-111075897 on Build GRCh38
rs140730628 Chr.5: 111076004-111076004 on Build GRCh38
rs953638488 Chr.5: 111076133-111076133 on Build GRCh38
rs146834103 Chr.5: 111076002-111076002 on Build GRCh38
rs866770629 Chr.5: 111075870-111075870 on Build GRCh38
rs931089699 Chr.5: 111076193-111076193 on Build GRCh38
rs543101378 Chr.5: 111075899-111075899 on Build GRCh38
rs750348614 Chr.5: 111076016-111076016 on Build GRCh38
rs1005903508 Chr.5: 111076286-111076286 on Build GRCh38
rs1010862164 Chr.5: 111076099-111076099 on Build GRCh38
rs779907677 Chr.5: 111076032-111076032 on Build GRCh38
rs542212889 Chr.5: 111075875-111075875 on Build GRCh38
rs575772001 Chr.5: 111075898-111075898 on Build GRCh38
rs775926557 Chr.5: 111076077-111076077 on Build GRCh38
rs759662143 Chr.5: 111076069-111076069 on Build GRCh38
rs1029444493 Chr.5: 111076132-111076132 on Build GRCh38
rs570708928 Chr.5: 111076247-111076247 on Build GRCh38
rs988151250 Chr.5: 111076142-111076147 on Build GRCh38
rs760739045 Chr.5: 111075932-111075932 on Build GRCh38
rs749079513 Chr.5: 111075907-111075907 on Build GRCh38
rs887529607 Chr.5: 111076273-111076273 on Build GRCh38
rs374147223 Chr.5: 111076096-111076096 on Build GRCh38
rs552674759 Chr.5: 111076214-111076214 on Build GRCh38
rs139391725 Chr.5: 111076051-111076051 on Build GRCh38
rs981350286 Chr.5: 111076102-111076102 on Build GRCh38
rs749099078 Chr.5: 111076039-111076039 on Build GRCh38
rs769664192 Chr.5: 111075904-111075904 on Build GRCh38
rs772318913 Chr.5: 111075926-111075926 on Build GRCh38
rs867610850 Chr.5: 111076030-111076030 on Build GRCh38
rs924746683 Chr.5: 111076210-111076210 on Build GRCh38

View Full Product Details