Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73597041-73597609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLXIPL
Entrez Gene ID
51085
Gene Name
MLX interacting protein like
Gene Aliases
CHREBP, MIO, MONDOB, WBSCR14, WS-bHLH, bHLHd14
UniGene
Hs.647055
Gene Chromosome Location
Chr.7: 73593194-73624543 on Build GRCh38
Transcripts
NM_032951.2 NM_032952.2 NM_032953.2 NM_032954.2 NR_134541.1 XM_011516277.1 XM_011516278.1 XM_011516279.1 XM_011516281.2 XM_017012263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3087267 c.1472C>T p.P491L Chr.7: 73597313-73597313 on Build GRCh38
5545302 c.1286C>T p.P429L Chr.7: 73597499-73597499 on Build GRCh38
4465280 c.1374C>T p.T458T Chr.7: 73597411-73597411 on Build GRCh38
7137296 c.1511A>T p.K504I Chr.7: 73597274-73597274 on Build GRCh38
1257817 c.1445C>T p.P482L Chr.7: 73597340-73597340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782056579 Chr.7: 73597346-73597346 on Build GRCh38
rs781786111 Chr.7: 73597340-73597340 on Build GRCh38
rs782505684 Chr.7: 73597159-73597159 on Build GRCh38
rs186788005 Chr.7: 73597587-73597587 on Build GRCh38
rs1022267570 Chr.7: 73597443-73597443 on Build GRCh38
rs528177933 Chr.7: 73597321-73597321 on Build GRCh38
rs782532816 Chr.7: 73597267-73597267 on Build GRCh38
rs944107548 Chr.7: 73597233-73597233 on Build GRCh38
rs782180374 Chr.7: 73597297-73597297 on Build GRCh38
rs781897947 Chr.7: 73597354-73597354 on Build GRCh38
rs782345327 Chr.7: 73597285-73597285 on Build GRCh38
rs782699960 Chr.7: 73597521-73597521 on Build GRCh38
rs782276244 Chr.7: 73597408-73597408 on Build GRCh38
rs377515363 Chr.7: 73597506-73597506 on Build GRCh38
rs781815374 Chr.7: 73597133-73597133 on Build GRCh38
rs912561153 Chr.7: 73597468-73597468 on Build GRCh38
rs202093717 Chr.7: 73597220-73597220 on Build GRCh38
rs782082995 Chr.7: 73597201-73597201 on Build GRCh38
rs782435817 Chr.7: 73597393-73597393 on Build GRCh38
rs202062025 Chr.7: 73597167-73597167 on Build GRCh38
rs1034527360 Chr.7: 73597366-73597366 on Build GRCh38
rs782536435 Chr.7: 73597560-73597560 on Build GRCh38
rs782650586 Chr.7: 73597359-73597359 on Build GRCh38
rs781816046 Chr.7: 73597235-73597235 on Build GRCh38
rs781945971 Chr.7: 73597206-73597206 on Build GRCh38
rs200391124 Chr.7: 73597248-73597248 on Build GRCh38
rs781801078 Chr.7: 73597255-73597255 on Build GRCh38
rs372996225 Chr.7: 73597441-73597441 on Build GRCh38
rs782012438 Chr.7: 73597503-73597503 on Build GRCh38
rs782580492 Chr.7: 73597405-73597405 on Build GRCh38
rs781843857 Chr.7: 73597379-73597379 on Build GRCh38
rs782753841 Chr.7: 73597507-73597507 on Build GRCh38
rs782306170 Chr.7: 73597170-73597170 on Build GRCh38
rs990775615 Chr.7: 73597208-73597208 on Build GRCh38
rs375075524 Chr.7: 73597144-73597144 on Build GRCh38
rs975428992 Chr.7: 73597513-73597513 on Build GRCh38
rs782383095 Chr.7: 73597313-73597313 on Build GRCh38
rs782419145 Chr.7: 73597192-73597192 on Build GRCh38
rs199597960 Chr.7: 73597357-73597357 on Build GRCh38
rs371223699 Chr.7: 73597534-73597534 on Build GRCh38
rs781933211 Chr.7: 73597181-73597181 on Build GRCh38
rs372922241 Chr.7: 73597223-73597223 on Build GRCh38
rs369598943 Chr.7: 73597296-73597296 on Build GRCh38
rs115063951 Chr.7: 73597135-73597135 on Build GRCh38
rs782320509 Chr.7: 73597483-73597483 on Build GRCh38
rs542524167 Chr.7: 73597242-73597242 on Build GRCh38
rs782276575 Chr.7: 73597189-73597189 on Build GRCh38
rs782647344 Chr.7: 73597416-73597416 on Build GRCh38
rs782805105 Chr.7: 73597352-73597352 on Build GRCh38
rs201189325 Chr.7: 73597438-73597438 on Build GRCh38
rs782273030 Chr.7: 73597417-73597417 on Build GRCh38
rs781870697 Chr.7: 73597518-73597518 on Build GRCh38
rs782737967 Chr.7: 73597337-73597337 on Build GRCh38
rs368147395 Chr.7: 73597302-73597302 on Build GRCh38
rs782600073 Chr.7: 73597579-73597579 on Build GRCh38
rs563363573 Chr.7: 73597318-73597318 on Build GRCh38
rs782601360 Chr.7: 73597166-73597166 on Build GRCh38
rs1034517510 Chr.7: 73597147-73597147 on Build GRCh38
rs782102432 Chr.7: 73597519-73597519 on Build GRCh38
rs781980316 Chr.7: 73597195-73597195 on Build GRCh38
rs200798784 Chr.7: 73597550-73597550 on Build GRCh38
rs886734637 Chr.7: 73597063-73597063 on Build GRCh38
rs981628227 Chr.7: 73597145-73597145 on Build GRCh38
rs977848786 Chr.7: 73597467-73597467 on Build GRCh38
rs944133589 Chr.7: 73597472-73597472 on Build GRCh38
rs530397056 Chr.7: 73597271-73597271 on Build GRCh38
rs200441433 Chr.7: 73597231-73597231 on Build GRCh38
rs921113235 Chr.7: 73597533-73597533 on Build GRCh38
rs782020793 Chr.7: 73597306-73597306 on Build GRCh38
rs781857335 Chr.7: 73597554-73597554 on Build GRCh38
rs931248364 Chr.7: 73597298-73597298 on Build GRCh38
rs782420994 Chr.7: 73597413-73597413 on Build GRCh38
rs782807594 Chr.7: 73597549-73597550 on Build GRCh38
rs782047753 Chr.7: 73597442-73597442 on Build GRCh38
rs959088840 Chr.7: 73597372-73597372 on Build GRCh38
rs782538733 Chr.7: 73597544-73597544 on Build GRCh38
rs200967567 Chr.7: 73597222-73597222 on Build GRCh38
rs782339784 Chr.7: 73597437-73597437 on Build GRCh38
rs782165394 Chr.7: 73597312-73597312 on Build GRCh38
rs899641728 Chr.7: 73597263-73597263 on Build GRCh38
rs782245118 Chr.7: 73597281-73597281 on Build GRCh38
rs1003982657 Chr.7: 73597109-73597109 on Build GRCh38
rs967643101 Chr.7: 73597457-73597457 on Build GRCh38

View Full Product Details