Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 214327456-214327946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMYD2
Entrez Gene ID
56950
Gene Name
SET and MYND domain containing 2
Gene Aliases
HSKM-B, KMT3C, ZMYND14
UniGene
Hs.66170
Gene Chromosome Location
Chr.1: 214281101-214337134 on Build GRCh38
Transcripts
NM_020197.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
903692 c.612G>T p.L204F Chr.1: 214327631-214327631 on Build GRCh38
7204745 c.632C>A p.P211H Chr.1: 214327651-214327651 on Build GRCh38
4728608 c.688A>T p.I230F Chr.1: 214327707-214327707 on Build GRCh38
3483232 c.626G>A p.C209Y Chr.1: 214327645-214327645 on Build GRCh38
5016920 c.670G>T p.V224F Chr.1: 214327689-214327689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750954495 Chr.1: 214327638-214327638 on Build GRCh38
rs755437613 Chr.1: 214327654-214327654 on Build GRCh38
rs756141383 Chr.1: 214327640-214327640 on Build GRCh38
rs3754135 Chr.1: 214327866-214327866 on Build GRCh38
rs145848094 Chr.1: 214327715-214327715 on Build GRCh38
rs764794193 Chr.1: 214327723-214327723 on Build GRCh38
rs747886560 Chr.1: 214327683-214327683 on Build GRCh38
rs377331966 Chr.1: 214327631-214327631 on Build GRCh38
rs781716201 Chr.1: 214327665-214327665 on Build GRCh38
rs768411353 Chr.1: 214327576-214327579 on Build GRCh38
rs199700774 Chr.1: 214327646-214327646 on Build GRCh38
rs759292947 Chr.1: 214327599-214327599 on Build GRCh38
rs3754136 Chr.1: 214327847-214327847 on Build GRCh38
rs115367134 Chr.1: 214327509-214327509 on Build GRCh38
rs749396283 Chr.1: 214327851-214327851 on Build GRCh38
rs755113399 Chr.1: 214327770-214327770 on Build GRCh38
rs375997509 Chr.1: 214327713-214327713 on Build GRCh38
rs780140828 Chr.1: 214327644-214327644 on Build GRCh38
rs944215994 Chr.1: 214327849-214327849 on Build GRCh38
rs746774186 Chr.1: 214327891-214327891 on Build GRCh38
rs11577049 Chr.1: 214327792-214327792 on Build GRCh38
rs757886919 Chr.1: 214327838-214327838 on Build GRCh38
rs780591468 Chr.1: 214327752-214327752 on Build GRCh38
rs753490062 Chr.1: 214327598-214327598 on Build GRCh38
rs751857836 Chr.1: 214327609-214327609 on Build GRCh38
rs779249565 Chr.1: 214327671-214327671 on Build GRCh38
rs866032840 Chr.1: 214327701-214327701 on Build GRCh38
rs966307386 Chr.1: 214327572-214327572 on Build GRCh38
rs752399811 Chr.1: 214327727-214327727 on Build GRCh38
rs957729420 Chr.1: 214327625-214327625 on Build GRCh38
rs140909791 Chr.1: 214327714-214327714 on Build GRCh38
rs889185350 Chr.1: 214327832-214327832 on Build GRCh38
rs771903679 Chr.1: 214327689-214327689 on Build GRCh38
rs17022608 Chr.1: 214327564-214327564 on Build GRCh38
rs762028244 Chr.1: 214327737-214327737 on Build GRCh38
rs116184067 Chr.1: 214327895-214327895 on Build GRCh38
rs35007434 Chr.1: 214327909-214327909 on Build GRCh38
rs751934401 Chr.1: 214327573-214327573 on Build GRCh38
rs760033955 Chr.1: 214327592-214327592 on Build GRCh38
rs1032304447 Chr.1: 214327915-214327915 on Build GRCh38
rs966734350 Chr.1: 214327875-214327875 on Build GRCh38
rs1049823232 Chr.1: 214327766-214327766 on Build GRCh38
rs773264753 Chr.1: 214327582-214327582 on Build GRCh38
rs953373015 Chr.1: 214327508-214327508 on Build GRCh38
rs760710296 Chr.1: 214327710-214327710 on Build GRCh38
rs951747870 Chr.1: 214327590-214327590 on Build GRCh38
rs765998966 Chr.1: 214327593-214327593 on Build GRCh38
rs753905068 Chr.1: 214327760-214327760 on Build GRCh38
rs756585130 Chr.1: 214327744-214327744 on Build GRCh38
rs765020826 Chr.1: 214327608-214327608 on Build GRCh38
rs957572251 Chr.1: 214327918-214327918 on Build GRCh38
rs757998411 Chr.1: 214327532-214327532 on Build GRCh38
rs7547464 Chr.1: 214327739-214327739 on Build GRCh38
rs150512369 Chr.1: 214327611-214327611 on Build GRCh38
rs371305512 Chr.1: 214327678-214327678 on Build GRCh38
rs138323600 Chr.1: 214327858-214327858 on Build GRCh38
rs768846751 Chr.1: 214327660-214327660 on Build GRCh38
rs372383872 Chr.1: 214327742-214327742 on Build GRCh38

View Full Product Details