Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 90920845-90921392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD1
Entrez Gene ID
27063
Gene Name
ankyrin repeat domain 1
Gene Aliases
ALRP, C-193, CARP, CVARP, MCARP, bA320F15.2
UniGene
Hs.448589
Gene Chromosome Location
Chr.10: 90912100-90921275 on Build GRCh38
Transcripts
NM_014391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3415335 c.21G>T p.E7D Chr.10: 90921007-90921007 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs898417901 Chr.10: 90921130-90921130 on Build GRCh38
rs188706671 Chr.10: 90921025-90921025 on Build GRCh38
rs140454947 Chr.10: 90921316-90921316 on Build GRCh38
rs376540175 Chr.10: 90920997-90920997 on Build GRCh38
rs1002757724 Chr.10: 90921274-90921274 on Build GRCh38
rs1034282425 Chr.10: 90921283-90921283 on Build GRCh38
rs547506147 Chr.10: 90921097-90921097 on Build GRCh38
rs1054588778 Chr.10: 90921165-90921165 on Build GRCh38
rs934241121 Chr.10: 90921156-90921156 on Build GRCh38
rs759918625 Chr.10: 90921040-90921040 on Build GRCh38
rs776096785 Chr.10: 90921051-90921051 on Build GRCh38
rs79341122 Chr.10: 90921044-90921044 on Build GRCh38
rs527415499 Chr.10: 90921095-90921095 on Build GRCh38
rs560990868 Chr.10: 90920879-90920879 on Build GRCh38
rs1032767934 Chr.10: 90920927-90920927 on Build GRCh38
rs757472270 Chr.10: 90920972-90920972 on Build GRCh38
rs778760010 Chr.10: 90920974-90920974 on Build GRCh38
rs760196723 Chr.10: 90921257-90921257 on Build GRCh38
rs918783135 Chr.10: 90921153-90921153 on Build GRCh38
rs1010055657 Chr.10: 90920962-90920962 on Build GRCh38
rs889414603 Chr.10: 90921162-90921162 on Build GRCh38
rs959369267 Chr.10: 90921077-90921077 on Build GRCh38
rs370966027 Chr.10: 90921068-90921068 on Build GRCh38
rs764297291 Chr.10: 90920957-90920957 on Build GRCh38
rs568634889 Chr.10: 90921194-90921194 on Build GRCh38
rs112853885 Chr.10: 90921140-90921140 on Build GRCh38
rs892928579 Chr.10: 90920943-90920943 on Build GRCh38
rs752451600 Chr.10: 90921085-90921085 on Build GRCh38
rs779910001 Chr.10: 90921000-90921000 on Build GRCh38
rs907040410 Chr.10: 90921196-90921196 on Build GRCh38
rs866047359 Chr.10: 90921248-90921248 on Build GRCh38
rs529670906 Chr.10: 90921326-90921326 on Build GRCh38
rs538475043 Chr.10: 90921158-90921158 on Build GRCh38
rs536205329 Chr.10: 90921108-90921108 on Build GRCh38
rs764249980 Chr.10: 90921078-90921078 on Build GRCh38
rs1031930499 Chr.10: 90921279-90921279 on Build GRCh38
rs772628017 Chr.10: 90921223-90921223 on Build GRCh38
rs369861544 Chr.10: 90921106-90921106 on Build GRCh38
rs114861196 Chr.10: 90920899-90920899 on Build GRCh38
rs558535158 Chr.10: 90921067-90921067 on Build GRCh38
rs772918277 Chr.10: 90921027-90921027 on Build GRCh38
rs746973988 Chr.10: 90921013-90921013 on Build GRCh38
rs892875939 Chr.10: 90921171-90921171 on Build GRCh38
rs750255548 Chr.10: 90920990-90920990 on Build GRCh38
rs941465626 Chr.10: 90921107-90921107 on Build GRCh38
rs114144205 Chr.10: 90921061-90921061 on Build GRCh38
rs113762010 Chr.10: 90920998-90920998 on Build GRCh38
rs754063667 Chr.10: 90920964-90920964 on Build GRCh38
rs571902498 Chr.10: 90921267-90921267 on Build GRCh38
rs549813663 Chr.10: 90921129-90921129 on Build GRCh38
rs760785625 Chr.10: 90920955-90920955 on Build GRCh38
rs1034557657 Chr.10: 90920958-90920958 on Build GRCh38
rs569896171 Chr.10: 90921157-90921157 on Build GRCh38
rs10881855 Chr.10: 90921172-90921172 on Build GRCh38
rs867775337 Chr.10: 90921079-90921079 on Build GRCh38
rs768122383 Chr.10: 90921023-90921023 on Build GRCh38
rs747822205 Chr.10: 90921026-90921026 on Build GRCh38
rs959338441 Chr.10: 90921287-90921287 on Build GRCh38
rs34182231 Chr.10: 90921249-90921250 on Build GRCh38
rs563967213 Chr.10: 90920944-90920944 on Build GRCh38
rs904892768 Chr.10: 90921230-90921230 on Build GRCh38
rs1001834077 Chr.10: 90921262-90921262 on Build GRCh38
rs761048391 Chr.10: 90920955-90920955 on Build GRCh38
rs1049659603 Chr.10: 90921202-90921202 on Build GRCh38
rs184982275 Chr.10: 90920900-90920900 on Build GRCh38
rs1006553984 Chr.10: 90920925-90920925 on Build GRCh38
rs942380998 Chr.10: 90921163-90921163 on Build GRCh38

View Full Product Details