Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57623666-57624265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT1
Entrez Gene ID
2583
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 1
Gene Aliases
GALGT, GALNACT, GalNAc-T, SPG26
UniGene
Hs.159481
Gene Chromosome Location
Chr.12: 57623409-57633239 on Build GRCh38
Transcripts
XM_005268773.4 XM_011538147.2 XM_011538148.2 XM_017019140.1 XM_017019141.1 XM_017019142.1 NM_133489.2

Gene Symbol
SLC26A10
Entrez Gene ID
65012
Gene Name
solute carrier family 26 member 10
Gene Aliases
-
UniGene
Hs.727726
Gene Chromosome Location
Chr.12: 57619910-57626151 on Build GRCh38
Transcripts
XM_005268773.4 XM_011538147.2 XM_011538148.2 XM_017019140.1 XM_017019141.1 XM_017019142.1 NM_133489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5947668 c.1103T>A p.L368Q Chr.12: 57623885-57623885 on Build GRCh38
5947669 c.1103T>A p.L368Q Chr.12: 57623885-57623885 on Build GRCh38
694931 c.1180C>A p.Q394K Chr.12: 57624051-57624051 on Build GRCh38
1606447 c.1207C>T p.H403Y Chr.12: 57624078-57624078 on Build GRCh38
1677133 c.1072T>C p.C358R Chr.12: 57623854-57623854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769635151 Chr.12: 57624192-57624192 on Build GRCh38
rs781203734 Chr.12: 57624177-57624177 on Build GRCh38
rs561569782 Chr.12: 57623861-57623861 on Build GRCh38
rs760458004 Chr.12: 57623952-57623952 on Build GRCh38
rs997497820 Chr.12: 57623742-57623742 on Build GRCh38
rs150360114 Chr.12: 57623827-57623827 on Build GRCh38
rs568720714 Chr.12: 57624209-57624209 on Build GRCh38
rs373458144 Chr.12: 57624087-57624087 on Build GRCh38
rs747519022 Chr.12: 57623848-57623848 on Build GRCh38
rs959995538 Chr.12: 57623797-57623797 on Build GRCh38
rs781504426 Chr.12: 57624018-57624018 on Build GRCh38
rs761814292 Chr.12: 57623976-57623976 on Build GRCh38
rs774248092 Chr.12: 57624043-57624055 on Build GRCh38
rs1039261991 Chr.12: 57623689-57623689 on Build GRCh38
rs539071136 Chr.12: 57623812-57623812 on Build GRCh38
rs746834623 Chr.12: 57624069-57624069 on Build GRCh38
rs770271939 Chr.12: 57623785-57623785 on Build GRCh38
rs770467812 Chr.12: 57623914-57623914 on Build GRCh38
rs907318942 Chr.12: 57623792-57623792 on Build GRCh38
rs543934504 Chr.12: 57623901-57623901 on Build GRCh38
rs990798628 Chr.12: 57623970-57623970 on Build GRCh38
rs778350445 Chr.12: 57624036-57624036 on Build GRCh38
rs773371585 Chr.12: 57623988-57623988 on Build GRCh38
rs776672734 Chr.12: 57623972-57623972 on Build GRCh38
rs530957010 Chr.12: 57623691-57623691 on Build GRCh38
rs777223521 Chr.12: 57623705-57623705 on Build GRCh38
rs1006248320 Chr.12: 57623759-57623759 on Build GRCh38
rs998157588 Chr.12: 57623891-57623891 on Build GRCh38
rs113207856 Chr.12: 57624077-57624077 on Build GRCh38
rs200269774 Chr.12: 57623952-57623956 on Build GRCh38
rs1054716463 Chr.12: 57623776-57623776 on Build GRCh38
rs775642373 Chr.12: 57624138-57624140 on Build GRCh38
rs768411064 Chr.12: 57624176-57624176 on Build GRCh38
rs1016269314 Chr.12: 57623925-57623925 on Build GRCh38
rs141249305 Chr.12: 57623745-57623745 on Build GRCh38
rs764323752 Chr.12: 57623808-57623808 on Build GRCh38
rs780439666 Chr.12: 57623844-57623844 on Build GRCh38
rs7314152 Chr.12: 57623918-57623918 on Build GRCh38
rs753250941 Chr.12: 57624023-57624023 on Build GRCh38
rs368986810 Chr.12: 57623796-57623796 on Build GRCh38
rs759350823 Chr.12: 57624198-57624198 on Build GRCh38
rs185308379 Chr.12: 57624217-57624217 on Build GRCh38
rs770590600 Chr.12: 57623864-57623864 on Build GRCh38
rs942921127 Chr.12: 57623936-57623936 on Build GRCh38
rs748826429 Chr.12: 57623913-57623913 on Build GRCh38
rs749922541 Chr.12: 57624129-57624129 on Build GRCh38
rs557287174 Chr.12: 57623772-57623772 on Build GRCh38
rs544376585 Chr.12: 57623879-57623879 on Build GRCh38
rs760593764 Chr.12: 57623871-57623871 on Build GRCh38
rs1016223829 Chr.12: 57623845-57623845 on Build GRCh38
rs370384752 Chr.12: 57624132-57624132 on Build GRCh38
rs780340975 Chr.12: 57623900-57623900 on Build GRCh38
rs769250393 Chr.12: 57623805-57623806 on Build GRCh38
rs759729651 Chr.12: 57624119-57624119 on Build GRCh38
rs894298351 Chr.12: 57624093-57624093 on Build GRCh38
rs768952054 Chr.12: 57623854-57623854 on Build GRCh38
rs146080672 Chr.12: 57624071-57624071 on Build GRCh38
rs377348471 Chr.12: 57624130-57624130 on Build GRCh38
rs778283744 Chr.12: 57624128-57624128 on Build GRCh38
rs758654153 Chr.12: 57623834-57623834 on Build GRCh38
rs756599558 Chr.12: 57624025-57624025 on Build GRCh38
rs757555807 Chr.12: 57623813-57623813 on Build GRCh38
rs934137099 Chr.12: 57623719-57623720 on Build GRCh38
rs756617762 Chr.12: 57624127-57624127 on Build GRCh38
rs146286105 Chr.12: 57623870-57623870 on Build GRCh38
rs181004315 Chr.12: 57624206-57624206 on Build GRCh38
rs911361055 Chr.12: 57623922-57623922 on Build GRCh38
rs950852882 Chr.12: 57623756-57623756 on Build GRCh38
rs115563091 Chr.12: 57623882-57623882 on Build GRCh38
rs746709835 Chr.12: 57624172-57624172 on Build GRCh38
rs1006264991 Chr.12: 57623924-57623924 on Build GRCh38
rs988520674 Chr.12: 57623814-57623814 on Build GRCh38
rs771748322 Chr.12: 57623921-57623921 on Build GRCh38
rs750654660 Chr.12: 57623890-57623890 on Build GRCh38
rs928541256 Chr.12: 57623789-57623789 on Build GRCh38
rs776549675 Chr.12: 57624074-57624074 on Build GRCh38
rs779222335 Chr.12: 57623817-57623817 on Build GRCh38
rs551152685 Chr.12: 57624224-57624224 on Build GRCh38
rs766534361 Chr.12: 57624094-57624094 on Build GRCh38
rs77704265 Chr.12: 57623741-57623741 on Build GRCh38
rs750503191 Chr.12: 57623982-57623982 on Build GRCh38
rs564432437 Chr.12: 57624120-57624120 on Build GRCh38
rs758629015 Chr.12: 57623893-57623893 on Build GRCh38
rs781622323 Chr.12: 57623910-57623910 on Build GRCh38
rs755293464 Chr.12: 57624012-57624012 on Build GRCh38
rs947152551 Chr.12: 57623843-57623843 on Build GRCh38
rs752616306 Chr.12: 57623799-57623799 on Build GRCh38
rs763152859 Chr.12: 57623984-57623984 on Build GRCh38
rs369487777 Chr.12: 57623865-57623865 on Build GRCh38
rs753056447 Chr.12: 57624125-57624125 on Build GRCh38
rs756052175 Chr.12: 57623801-57623801 on Build GRCh38
rs546348968 Chr.12: 57624182-57624182 on Build GRCh38
rs764053855 Chr.12: 57623873-57623873 on Build GRCh38
rs768739554 Chr.12: 57623957-57623957 on Build GRCh38
rs942918735 Chr.12: 57623773-57623773 on Build GRCh38
rs753960059 Chr.12: 57623811-57623811 on Build GRCh38
rs771387746 Chr.12: 57624193-57624193 on Build GRCh38
rs757888053 Chr.12: 57624144-57624144 on Build GRCh38
rs755481490 Chr.12: 57623903-57623903 on Build GRCh38
rs187351673 Chr.12: 57623714-57623714 on Build GRCh38
rs759817543 Chr.12: 57624218-57624218 on Build GRCh38
rs773155266 Chr.12: 57624197-57624197 on Build GRCh38
rs759444224 Chr.12: 57624059-57624059 on Build GRCh38
rs759201708 Chr.12: 57623793-57623793 on Build GRCh38
rs751884673 Chr.12: 57624005-57624005 on Build GRCh38
rs376956247 Chr.12: 57623931-57623931 on Build GRCh38
rs746526883 Chr.12: 57623778-57623778 on Build GRCh38
rs765591316 Chr.12: 57623887-57623887 on Build GRCh38
rs765461569 Chr.12: 57623977-57623977 on Build GRCh38
rs767767286 Chr.12: 57624130-57624131 on Build GRCh38
rs772776805 Chr.12: 57623862-57623862 on Build GRCh38
rs369961371 Chr.12: 57624037-57624037 on Build GRCh38
rs142798108 Chr.12: 57624065-57624065 on Build GRCh38
rs763035220 Chr.12: 57624091-57624091 on Build GRCh38
rs769853441 Chr.12: 57624082-57624082 on Build GRCh38
rs1001302779 Chr.12: 57624035-57624035 on Build GRCh38
rs572812203 Chr.12: 57623782-57623782 on Build GRCh38
rs902070006 Chr.12: 57623889-57623889 on Build GRCh38
rs779401871 Chr.12: 57624150-57624150 on Build GRCh38
rs914857677 Chr.12: 57623997-57623997 on Build GRCh38
rs751844698 Chr.12: 57624116-57624116 on Build GRCh38
rs749177472 Chr.12: 57623885-57623886 on Build GRCh38
rs375009969 Chr.12: 57623863-57623863 on Build GRCh38
rs771013381 Chr.12: 57623895-57623896 on Build GRCh38

View Full Product Details