Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20501115-20501623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUL1
Entrez Gene ID
79594
Gene Name
mitochondrial E3 ubiquitin protein ligase 1
Gene Aliases
C1orf166, GIDE, MAPL, MULAN, RNF218
UniGene
Hs.10101
Gene Chromosome Location
Chr.1: 20499448-20508483 on Build GRCh38
Transcripts
NM_024544.2 XM_011542137.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
349949 c.633G>T p.Q211H Chr.1: 20501116-20501116 on Build GRCh38
903258 c.578C>T p.A193V Chr.1: 20501171-20501171 on Build GRCh38
903259 c.486G>A p.S162S Chr.1: 20501263-20501263 on Build GRCh38
5509669 c.625C>A p.R209S Chr.1: 20501124-20501124 on Build GRCh38
5545348 c.483C>T p.P161P Chr.1: 20501266-20501266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765538600 Chr.1: 20501251-20501251 on Build GRCh38
rs771758934 Chr.1: 20501420-20501420 on Build GRCh38
rs766306757 Chr.1: 20501369-20501369 on Build GRCh38
rs776763974 Chr.1: 20501340-20501340 on Build GRCh38
rs1024184961 Chr.1: 20501511-20501511 on Build GRCh38
rs11541821 Chr.1: 20501158-20501158 on Build GRCh38
rs749306191 Chr.1: 20501295-20501295 on Build GRCh38
rs547802214 Chr.1: 20501152-20501152 on Build GRCh38
rs553352028 Chr.1: 20501539-20501539 on Build GRCh38
rs903665772 Chr.1: 20501263-20501263 on Build GRCh38
rs764135854 Chr.1: 20501338-20501338 on Build GRCh38
rs956646615 Chr.1: 20501319-20501319 on Build GRCh38
rs1033074762 Chr.1: 20501446-20501446 on Build GRCh38
rs758966256 Chr.1: 20501217-20501217 on Build GRCh38
rs747096057 Chr.1: 20501374-20501385 on Build GRCh38
rs761693520 Chr.1: 20501343-20501343 on Build GRCh38
rs745955061 Chr.1: 20501156-20501156 on Build GRCh38
rs369085744 Chr.1: 20501248-20501248 on Build GRCh38
rs761186067 Chr.1: 20501301-20501301 on Build GRCh38
rs373150717 Chr.1: 20501402-20501402 on Build GRCh38
rs145815176 Chr.1: 20501291-20501291 on Build GRCh38
rs558350162 Chr.1: 20501596-20501596 on Build GRCh38
rs376968395 Chr.1: 20501384-20501384 on Build GRCh38
rs139245041 Chr.1: 20501247-20501247 on Build GRCh38
rs750565337 Chr.1: 20501346-20501346 on Build GRCh38
rs529262488 Chr.1: 20501137-20501137 on Build GRCh38
rs545543120 Chr.1: 20501580-20501580 on Build GRCh38
rs750721324 Chr.1: 20501252-20501252 on Build GRCh38
rs971220121 Chr.1: 20501525-20501525 on Build GRCh38
rs574179398 Chr.1: 20501574-20501574 on Build GRCh38
rs748499724 Chr.1: 20501293-20501293 on Build GRCh38
rs746499719 Chr.1: 20501437-20501437 on Build GRCh38
rs755109188 Chr.1: 20501375-20501375 on Build GRCh38
rs1000697457 Chr.1: 20501600-20501600 on Build GRCh38
rs372998635 Chr.1: 20501239-20501239 on Build GRCh38
rs900226227 Chr.1: 20501421-20501421 on Build GRCh38
rs768855958 Chr.1: 20501193-20501193 on Build GRCh38
rs927498311 Chr.1: 20501134-20501134 on Build GRCh38
rs201171244 Chr.1: 20501268-20501268 on Build GRCh38
rs112738588 Chr.1: 20501477-20501477 on Build GRCh38
rs774048986 Chr.1: 20501210-20501210 on Build GRCh38
rs770212403 Chr.1: 20501303-20501303 on Build GRCh38
rs758388916 Chr.1: 20501262-20501262 on Build GRCh38
rs142333872 Chr.1: 20501355-20501355 on Build GRCh38
rs371173943 Chr.1: 20501426-20501426 on Build GRCh38
rs376828034 Chr.1: 20501403-20501403 on Build GRCh38
rs964377314 Chr.1: 20501245-20501245 on Build GRCh38
rs780208591 Chr.1: 20501264-20501264 on Build GRCh38
rs1034425491 Chr.1: 20501597-20501597 on Build GRCh38
rs768345541 Chr.1: 20501441-20501441 on Build GRCh38
rs780418458 Chr.1: 20501167-20501167 on Build GRCh38
rs535123027 Chr.1: 20501478-20501478 on Build GRCh38
rs747603719 Chr.1: 20501172-20501172 on Build GRCh38
rs775522813 Chr.1: 20501217-20501217 on Build GRCh38
rs201135449 Chr.1: 20501452-20501452 on Build GRCh38
rs1039775777 Chr.1: 20501594-20501594 on Build GRCh38
rs1051015006 Chr.1: 20501566-20501566 on Build GRCh38
rs147902092 Chr.1: 20501383-20501383 on Build GRCh38
rs747437335 Chr.1: 20501266-20501266 on Build GRCh38
rs139475946 Chr.1: 20501194-20501194 on Build GRCh38
rs191490444 Chr.1: 20501216-20501216 on Build GRCh38
rs376986230 Chr.1: 20501168-20501168 on Build GRCh38
rs200114346 Chr.1: 20501356-20501356 on Build GRCh38
rs555995359 Chr.1: 20501401-20501401 on Build GRCh38
rs761887984 Chr.1: 20501459-20501459 on Build GRCh38
rs148047459 Chr.1: 20501166-20501166 on Build GRCh38
rs200015271 Chr.1: 20501200-20501200 on Build GRCh38
rs751677130 Chr.1: 20501374-20501374 on Build GRCh38
rs766747227 Chr.1: 20501231-20501231 on Build GRCh38
rs372420757 Chr.1: 20501307-20501307 on Build GRCh38
rs369750780 Chr.1: 20501223-20501223 on Build GRCh38
rs775095645 Chr.1: 20501323-20501323 on Build GRCh38
rs974247537 Chr.1: 20501468-20501468 on Build GRCh38
rs376478251 Chr.1: 20501317-20501317 on Build GRCh38
rs370886196 Chr.1: 20501344-20501344 on Build GRCh38
rs41265065 Chr.1: 20501333-20501333 on Build GRCh38
rs112773790 Chr.1: 20501224-20501224 on Build GRCh38
rs769674610 Chr.1: 20501467-20501467 on Build GRCh38
rs942717565 Chr.1: 20501586-20501586 on Build GRCh38
rs552383357 Chr.1: 20501230-20501230 on Build GRCh38
rs770584555 Chr.1: 20501198-20501198 on Build GRCh38
rs941378215 Chr.1: 20501331-20501331 on Build GRCh38
rs773963790 Chr.1: 20501306-20501306 on Build GRCh38

View Full Product Details