Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40462566-40463130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP69B
Entrez Gene ID
65243
Gene Name
ZFP69 zinc finger protein B
Gene Aliases
ZKSCAN23B, ZNF643, ZSCAN54B
UniGene
Hs.133034
Gene Chromosome Location
Chr.1: 40450068-40466765 on Build GRCh38
Transcripts
NM_023070.2 XM_005271136.1 XM_005271137.2 XM_005271138.3 XM_005271139.3 XM_017002147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3489572 c.709G>A p.E237K Chr.1: 40462999-40462999 on Build GRCh38
5817707 c.640T>A p.F214I Chr.1: 40462930-40462930 on Build GRCh38
4744538 c.609T>G p.S203R Chr.1: 40462800-40462800 on Build GRCh38
3489565 c.604C>T p.Q202* Chr.1: 40462588-40462588 on Build GRCh38
3489564 c.280G>A p.E94K Chr.1: 40462570-40462570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112085724 Chr.1: 40463108-40463108 on Build GRCh38
rs774812261 Chr.1: 40462901-40462901 on Build GRCh38
rs747719612 Chr.1: 40462976-40462976 on Build GRCh38
rs927910998 Chr.1: 40462920-40462920 on Build GRCh38
rs904715631 Chr.1: 40462619-40462619 on Build GRCh38
rs755104442 Chr.1: 40462653-40462653 on Build GRCh38
rs757011618 Chr.1: 40462827-40462827 on Build GRCh38
rs143573677 Chr.1: 40462646-40462646 on Build GRCh38
rs116091241 Chr.1: 40462736-40462736 on Build GRCh38
rs762732159 Chr.1: 40463033-40463033 on Build GRCh38
rs373622720 Chr.1: 40462955-40462955 on Build GRCh38
rs772628352 Chr.1: 40462986-40462986 on Build GRCh38
rs749391392 Chr.1: 40463024-40463024 on Build GRCh38
rs768286935 Chr.1: 40462754-40462754 on Build GRCh38
rs757034017 Chr.1: 40462659-40462659 on Build GRCh38
rs955594442 Chr.1: 40462735-40462735 on Build GRCh38
rs531715913 Chr.1: 40462686-40462686 on Build GRCh38
rs781223132 Chr.1: 40463054-40463054 on Build GRCh38
rs764361071 Chr.1: 40462818-40462818 on Build GRCh38
rs748984948 Chr.1: 40463061-40463061 on Build GRCh38
rs997014385 Chr.1: 40462706-40462706 on Build GRCh38
rs773126037 Chr.1: 40462922-40462922 on Build GRCh38
rs778862276 Chr.1: 40462719-40462720 on Build GRCh38
rs143880922 Chr.1: 40462880-40462880 on Build GRCh38
rs760012840 Chr.1: 40462909-40462909 on Build GRCh38
rs768165759 Chr.1: 40462790-40462790 on Build GRCh38
rs1024635645 Chr.1: 40462708-40462708 on Build GRCh38
rs750758286 Chr.1: 40462620-40462620 on Build GRCh38
rs777576148 Chr.1: 40462982-40462982 on Build GRCh38
rs375926428 Chr.1: 40462668-40462668 on Build GRCh38
rs537532571 Chr.1: 40462992-40462992 on Build GRCh38
rs747430571 Chr.1: 40463083-40463083 on Build GRCh38
rs774931726 Chr.1: 40462731-40462731 on Build GRCh38
rs759323159 Chr.1: 40463100-40463100 on Build GRCh38
rs151064151 Chr.1: 40462893-40462893 on Build GRCh38
rs753564338 Chr.1: 40462606-40462606 on Build GRCh38
rs371518930 Chr.1: 40462912-40462912 on Build GRCh38
rs548981075 Chr.1: 40462671-40462671 on Build GRCh38
rs562615056 Chr.1: 40462676-40462676 on Build GRCh38
rs763365431 Chr.1: 40462764-40462764 on Build GRCh38
rs12407929 Chr.1: 40463059-40463059 on Build GRCh38
rs868535806 Chr.1: 40462927-40462927 on Build GRCh38
rs61748795 Chr.1: 40462803-40462803 on Build GRCh38
rs761972282 Chr.1: 40462952-40462952 on Build GRCh38
rs751494351 Chr.1: 40463038-40463038 on Build GRCh38
rs946487160 Chr.1: 40462978-40462978 on Build GRCh38
rs758471306 Chr.1: 40462852-40462852 on Build GRCh38
rs769225195 Chr.1: 40463002-40463002 on Build GRCh38
rs148910082 Chr.1: 40462915-40462915 on Build GRCh38
rs772457918 Chr.1: 40463067-40463067 on Build GRCh38
rs752192494 Chr.1: 40463056-40463056 on Build GRCh38
rs140502577 Chr.1: 40462890-40462890 on Build GRCh38
rs754482617 Chr.1: 40462966-40462966 on Build GRCh38
rs144187238 Chr.1: 40462641-40462641 on Build GRCh38
rs780029539 Chr.1: 40462958-40462958 on Build GRCh38
rs776532371 Chr.1: 40463013-40463013 on Build GRCh38
rs776030132 Chr.1: 40462988-40462988 on Build GRCh38
rs147133462 Chr.1: 40463036-40463036 on Build GRCh38
rs377590446 Chr.1: 40462715-40462715 on Build GRCh38
rs997045463 Chr.1: 40463046-40463046 on Build GRCh38
rs1015598524 Chr.1: 40463079-40463079 on Build GRCh38
rs557878781 Chr.1: 40463018-40463018 on Build GRCh38
rs763232411 Chr.1: 40462635-40462635 on Build GRCh38
rs768111486 Chr.1: 40462891-40462891 on Build GRCh38
rs373664999 Chr.1: 40462592-40462592 on Build GRCh38
rs1052515651 Chr.1: 40462610-40462610 on Build GRCh38
rs769534623 Chr.1: 40462977-40462977 on Build GRCh38
rs771403576 Chr.1: 40462900-40462900 on Build GRCh38
rs778726084 Chr.1: 40462852-40462852 on Build GRCh38
rs146396152 Chr.1: 40463072-40463072 on Build GRCh38
rs899960990 Chr.1: 40462661-40462661 on Build GRCh38
rs750378184 Chr.1: 40462780-40462783 on Build GRCh38
rs746180211 Chr.1: 40462983-40462983 on Build GRCh38
rs763673910 Chr.1: 40462615-40462615 on Build GRCh38
rs766918354 Chr.1: 40462765-40462765 on Build GRCh38
rs771547294 Chr.1: 40462729-40462729 on Build GRCh38
rs771633065 Chr.1: 40462591-40462591 on Build GRCh38
rs767748640 Chr.1: 40462657-40462657 on Build GRCh38
rs777353372 Chr.1: 40462667-40462667 on Build GRCh38
rs749050942 Chr.1: 40462682-40462682 on Build GRCh38
rs754072556 Chr.1: 40462822-40462822 on Build GRCh38
rs367759748 Chr.1: 40463030-40463030 on Build GRCh38
rs761884118 Chr.1: 40463028-40463028 on Build GRCh38
rs754826548 Chr.1: 40463042-40463042 on Build GRCh38
rs141195830 Chr.1: 40463058-40463058 on Build GRCh38
rs371400359 Chr.1: 40463104-40463104 on Build GRCh38
rs892231289 Chr.1: 40462614-40462614 on Build GRCh38
rs778813963 Chr.1: 40462723-40462723 on Build GRCh38
rs747220835 Chr.1: 40463075-40463075 on Build GRCh38
rs139917327 Chr.1: 40462789-40462789 on Build GRCh38
rs143377630 Chr.1: 40462816-40462816 on Build GRCh38
rs150144613 Chr.1: 40462910-40462910 on Build GRCh38
rs370968880 Chr.1: 40462757-40462757 on Build GRCh38
rs527970050 Chr.1: 40462866-40462866 on Build GRCh38
rs758201525 Chr.1: 40462956-40462956 on Build GRCh38
rs754065879 Chr.1: 40462924-40462924 on Build GRCh38
rs760800372 Chr.1: 40462917-40462917 on Build GRCh38
rs372387090 Chr.1: 40462727-40462727 on Build GRCh38
rs780617088 Chr.1: 40462970-40462970 on Build GRCh38
rs745685335 Chr.1: 40462863-40462863 on Build GRCh38
rs148278676 Chr.1: 40462606-40462606 on Build GRCh38
rs947927350 Chr.1: 40462908-40462908 on Build GRCh38
rs776607691 Chr.1: 40463040-40463055 on Build GRCh38
rs560379802 Chr.1: 40463085-40463085 on Build GRCh38
rs137945908 Chr.1: 40462964-40462964 on Build GRCh38
rs770833768 Chr.1: 40463031-40463031 on Build GRCh38
rs774968060 Chr.1: 40462787-40462787 on Build GRCh38

View Full Product Details