Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93896250-93896823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCLM
Entrez Gene ID
2730
Gene Name
glutamate-cysteine ligase modifier subunit
Gene Aliases
GLCLR
UniGene
Hs.315562
Gene Chromosome Location
Chr.1: 93885199-93909598 on Build GRCh38
Transcripts
NM_001308253.1 NM_002061.3 XM_011541261.2 XM_017001056.1 XM_017001057.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4488857 c.339C>T p.A113A Chr.1: 93896819-93896819 on Build GRCh38
1172181 c.389C>T p.P130L Chr.1: 93896769-93896769 on Build GRCh38
281338 c.383C>T p.A128V Chr.1: 93896775-93896775 on Build GRCh38
913438 c.523C>A p.L175M Chr.1: 93896635-93896635 on Build GRCh38
5014134 c.404G>A p.G135E Chr.1: 93896754-93896754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057292024 Chr.1: 93896308-93896308 on Build GRCh38
rs1052344083 Chr.1: 93896478-93896478 on Build GRCh38
rs768409891 Chr.1: 93896628-93896628 on Build GRCh38
rs749744134 Chr.1: 93896768-93896768 on Build GRCh38
rs780522121 Chr.1: 93896767-93896767 on Build GRCh38
rs746859746 Chr.1: 93896764-93896764 on Build GRCh38
rs914503045 Chr.1: 93896316-93896316 on Build GRCh38
rs781586151 Chr.1: 93896749-93896749 on Build GRCh38
rs770263513 Chr.1: 93896593-93896593 on Build GRCh38
rs752882203 Chr.1: 93896696-93896696 on Build GRCh38
rs777843165 Chr.1: 93896725-93896725 on Build GRCh38
rs527366545 Chr.1: 93896357-93896357 on Build GRCh38
rs760736712 Chr.1: 93896677-93896677 on Build GRCh38
rs911712763 Chr.1: 93896557-93896557 on Build GRCh38
rs193270904 Chr.1: 93896509-93896509 on Build GRCh38
rs750225700 Chr.1: 93896660-93896660 on Build GRCh38
rs773867359 Chr.1: 93896775-93896775 on Build GRCh38
rs758423656 Chr.1: 93896724-93896724 on Build GRCh38
rs551308131 Chr.1: 93896542-93896542 on Build GRCh38
rs1037883029 Chr.1: 93896564-93896564 on Build GRCh38
rs1803636 Chr.1: 93896799-93896799 on Build GRCh38
rs201660483 Chr.1: 93896279-93896279 on Build GRCh38
rs950614652 Chr.1: 93896496-93896496 on Build GRCh38
rs560036794 Chr.1: 93896297-93896297 on Build GRCh38
rs994878319 Chr.1: 93896395-93896395 on Build GRCh38
rs142756309 Chr.1: 93896737-93896737 on Build GRCh38
rs933112511 Chr.1: 93896477-93896477 on Build GRCh38
rs554987315 Chr.1: 93896370-93896370 on Build GRCh38
rs775019592 Chr.1: 93896601-93896601 on Build GRCh38
rs746396137 Chr.1: 93896591-93896591 on Build GRCh38
rs188669727 Chr.1: 93896419-93896419 on Build GRCh38
rs766417928 Chr.1: 93896683-93896683 on Build GRCh38
rs1025940625 Chr.1: 93896460-93896460 on Build GRCh38
rs964927403 Chr.1: 93896597-93896597 on Build GRCh38
rs988699880 Chr.1: 93896312-93896312 on Build GRCh38
rs921668275 Chr.1: 93896379-93896379 on Build GRCh38
rs535381519 Chr.1: 93896347-93896347 on Build GRCh38
rs776941374 Chr.1: 93896584-93896584 on Build GRCh38
rs895999830 Chr.1: 93896315-93896315 on Build GRCh38
rs751761445 Chr.1: 93896734-93896734 on Build GRCh38
rs955876902 Chr.1: 93896303-93896303 on Build GRCh38
rs377507485 Chr.1: 93896417-93896417 on Build GRCh38
rs899427361 Chr.1: 93896569-93896569 on Build GRCh38
rs768052355 Chr.1: 93896769-93896769 on Build GRCh38
rs761459298 Chr.1: 93896644-93896644 on Build GRCh38
rs776170924 Chr.1: 93896570-93896570 on Build GRCh38
rs767446073 Chr.1: 93896647-93896647 on Build GRCh38
rs762363248 Chr.1: 93896626-93896626 on Build GRCh38
rs560889470 Chr.1: 93896491-93896491 on Build GRCh38
rs200732106 Chr.1: 93896633-93896633 on Build GRCh38
rs940874438 Chr.1: 93896531-93896531 on Build GRCh38
rs377397123 Chr.1: 93896504-93896504 on Build GRCh38

View Full Product Details