Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41434313-41434866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOB2
Entrez Gene ID
10766
Gene Name
transducer of ERBB2, 2
Gene Aliases
APRO5, TOB4, TOBL, TROB2
UniGene
Hs.474978
Gene Chromosome Location
Chr.22: 41433488-41448230 on Build GRCh38
Transcripts
NM_016272.3 XM_005261315.2 XM_006724105.3 XM_017028539.1

SNP Detail

SNP ID Chromosome Location
rs143705293 Chr.22: 41434805-41434805 on Build GRCh38
rs540704799 Chr.22: 41434563-41434563 on Build GRCh38
rs1022802433 Chr.22: 41434765-41434765 on Build GRCh38
rs370341833 Chr.22: 41434614-41434614 on Build GRCh38
rs957456541 Chr.22: 41434500-41434500 on Build GRCh38
rs865872772 Chr.22: 41434541-41434541 on Build GRCh38
rs982895194 Chr.22: 41434570-41434570 on Build GRCh38
rs941524101 Chr.22: 41434346-41434346 on Build GRCh38
rs56176102 Chr.22: 41434607-41434607 on Build GRCh38
rs370712667 Chr.22: 41434774-41434774 on Build GRCh38
rs921396679 Chr.22: 41434618-41434618 on Build GRCh38
rs532758375 Chr.22: 41434640-41434640 on Build GRCh38
rs1039048028 Chr.22: 41434371-41434371 on Build GRCh38
rs200324753 Chr.22: 41434395-41434395 on Build GRCh38
rs1051262655 Chr.22: 41434683-41434683 on Build GRCh38
rs564491250 Chr.22: 41434601-41434601 on Build GRCh38
rs535986703 Chr.22: 41434798-41434798 on Build GRCh38
rs900077748 Chr.22: 41434376-41434376 on Build GRCh38
rs1050725983 Chr.22: 41434415-41434415 on Build GRCh38
rs890804098 Chr.22: 41434697-41434697 on Build GRCh38
rs1011708513 Chr.22: 41434755-41434755 on Build GRCh38
rs941562352 Chr.22: 41434609-41434609 on Build GRCh38
rs932907364 Chr.22: 41434673-41434673 on Build GRCh38
rs970275042 Chr.22: 41434770-41434770 on Build GRCh38
rs141317378 Chr.22: 41434668-41434668 on Build GRCh38
rs960719762 Chr.22: 41434825-41434825 on Build GRCh38
rs1053319170 Chr.22: 41434712-41434712 on Build GRCh38
rs927971131 Chr.22: 41434561-41434561 on Build GRCh38
rs1023338433 Chr.22: 41434548-41434548 on Build GRCh38
rs576581365 Chr.22: 41434443-41434443 on Build GRCh38
rs996948568 Chr.22: 41434404-41434404 on Build GRCh38
rs41276315 Chr.22: 41434704-41434704 on Build GRCh38
rs892879291 Chr.22: 41434715-41434715 on Build GRCh38
rs1047156991 Chr.22: 41434332-41434332 on Build GRCh38
rs964556252 Chr.22: 41434830-41434830 on Build GRCh38
rs113362539 Chr.22: 41434478-41434478 on Build GRCh38
rs184637794 Chr.22: 41434489-41434489 on Build GRCh38
rs567444875 Chr.22: 41434781-41434781 on Build GRCh38
rs41276313 Chr.22: 41434555-41434555 on Build GRCh38
rs866018003 Chr.22: 41434442-41434442 on Build GRCh38
rs887288107 Chr.22: 41434337-41434337 on Build GRCh38
rs569807170 Chr.22: 41434669-41434669 on Build GRCh38
rs868180767 Chr.22: 41434521-41434521 on Build GRCh38
rs1011407381 Chr.22: 41434528-41434528 on Build GRCh38

View Full Product Details