Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19504726-19505289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDE1
Entrez Gene ID
51573
Gene Name
glycerophosphodiester phosphodiesterase 1
Gene Aliases
363E6.2, MIR16
UniGene
Hs.512607
Gene Chromosome Location
Chr.16: 19501693-19522145 on Build GRCh38
Transcripts
NM_001324066.1 NM_001324067.1 NM_016641.3 NR_136689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5631794 c.754A>G p.M252V Chr.16: 19504975-19504975 on Build GRCh38
3988300 c.748T>G p.F250V Chr.16: 19504981-19504981 on Build GRCh38
702577 c.719G>A p.R240H Chr.16: 19505010-19505010 on Build GRCh38
3817398 c.652C>G p.R218G Chr.16: 19505077-19505077 on Build GRCh38
6484247 c.759G>A p.M253I Chr.16: 19504970-19504970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780579441 Chr.16: 19504876-19504877 on Build GRCh38
rs755539443 Chr.16: 19505025-19505025 on Build GRCh38
rs140559686 Chr.16: 19504799-19504799 on Build GRCh38
rs779413798 Chr.16: 19505094-19505094 on Build GRCh38
rs762760428 Chr.16: 19505004-19505004 on Build GRCh38
rs780076983 Chr.16: 19505119-19505119 on Build GRCh38
rs939491354 Chr.16: 19505092-19505092 on Build GRCh38
rs998334154 Chr.16: 19504806-19504806 on Build GRCh38
rs200268131 Chr.16: 19504899-19504899 on Build GRCh38
rs140966841 Chr.16: 19504984-19504984 on Build GRCh38
rs747148336 Chr.16: 19505134-19505134 on Build GRCh38
rs765003403 Chr.16: 19504839-19504839 on Build GRCh38
rs750924151 Chr.16: 19504861-19504862 on Build GRCh38
rs374623694 Chr.16: 19504968-19504968 on Build GRCh38
rs775629516 Chr.16: 19504958-19504958 on Build GRCh38
rs756658093 Chr.16: 19504890-19504890 on Build GRCh38
rs879409705 Chr.16: 19504766-19504766 on Build GRCh38
rs758784142 Chr.16: 19504877-19504877 on Build GRCh38
rs773096688 Chr.16: 19505002-19505002 on Build GRCh38
rs1003314202 Chr.16: 19504992-19504992 on Build GRCh38
rs777977948 Chr.16: 19504831-19504832 on Build GRCh38
rs758663648 Chr.16: 19505101-19505101 on Build GRCh38
rs773182614 Chr.16: 19505166-19505166 on Build GRCh38
rs527799526 Chr.16: 19505209-19505209 on Build GRCh38
rs968935377 Chr.16: 19505106-19505106 on Build GRCh38
rs571341439 Chr.16: 19505010-19505010 on Build GRCh38
rs536938668 Chr.16: 19505210-19505210 on Build GRCh38
rs754531926 Chr.16: 19504858-19504858 on Build GRCh38
rs1005421126 Chr.16: 19504884-19504884 on Build GRCh38
rs929958655 Chr.16: 19504828-19504828 on Build GRCh38
rs369924505 Chr.16: 19504928-19504928 on Build GRCh38
rs751189103 Chr.16: 19504976-19504978 on Build GRCh38
rs766430568 Chr.16: 19505130-19505130 on Build GRCh38
rs766140341 Chr.16: 19505005-19505005 on Build GRCh38
rs946858179 Chr.16: 19505242-19505242 on Build GRCh38
rs376550940 Chr.16: 19505007-19505007 on Build GRCh38
rs766052454 Chr.16: 19504847-19504847 on Build GRCh38
rs554574797 Chr.16: 19504816-19504816 on Build GRCh38
rs953827172 Chr.16: 19504836-19504836 on Build GRCh38
rs911408257 Chr.16: 19505261-19505261 on Build GRCh38
rs751165180 Chr.16: 19504850-19504850 on Build GRCh38
rs762672600 Chr.16: 19504845-19504845 on Build GRCh38
rs901181962 Chr.16: 19504812-19504812 on Build GRCh38
rs374592814 Chr.16: 19505143-19505143 on Build GRCh38
rs532212787 Chr.16: 19504964-19504964 on Build GRCh38
rs2072086 Chr.16: 19505076-19505076 on Build GRCh38
rs757643915 Chr.16: 19504929-19504929 on Build GRCh38
rs752254773 Chr.16: 19505020-19505020 on Build GRCh38
rs1047424277 Chr.16: 19504834-19504834 on Build GRCh38
rs756684532 Chr.16: 19504756-19504756 on Build GRCh38
rs374308081 Chr.16: 19505034-19505034 on Build GRCh38
rs780723543 Chr.16: 19504880-19504880 on Build GRCh38
rs1023458711 Chr.16: 19504885-19504885 on Build GRCh38
rs769779940 Chr.16: 19504966-19504966 on Build GRCh38
rs374018494 Chr.16: 19505060-19505060 on Build GRCh38
rs772107691 Chr.16: 19504957-19504957 on Build GRCh38
rs201968694 Chr.16: 19504936-19504936 on Build GRCh38
rs779324598 Chr.16: 19504934-19504934 on Build GRCh38
rs770780728 Chr.16: 19505178-19505178 on Build GRCh38
rs1041083874 Chr.16: 19504804-19504804 on Build GRCh38
rs750233613 Chr.16: 19504843-19504843 on Build GRCh38
rs201526728 Chr.16: 19504961-19504961 on Build GRCh38
rs757733912 Chr.16: 19505077-19505077 on Build GRCh38
rs200327001 Chr.16: 19504881-19504881 on Build GRCh38
rs767017831 Chr.16: 19505011-19505011 on Build GRCh38
rs765589950 Chr.16: 19504822-19504822 on Build GRCh38
rs184769837 Chr.16: 19505196-19505196 on Build GRCh38
rs749195346 Chr.16: 19505168-19505168 on Build GRCh38
rs370745528 Chr.16: 19505098-19505098 on Build GRCh38
rs1043973063 Chr.16: 19505260-19505260 on Build GRCh38
rs765815603 Chr.16: 19504844-19504847 on Build GRCh38
rs942776070 Chr.16: 19504791-19504791 on Build GRCh38
rs972788447 Chr.16: 19505040-19505040 on Build GRCh38

View Full Product Details