Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30707418-30708013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRCAP
Entrez Gene ID
10847
Gene Name
Snf2-related CREBBP activator protein
Gene Aliases
DOMO1, EAF1, FLHS, SWR1
UniGene
Hs.620916
Gene Chromosome Location
Chr.16: 30699141-30740129 on Build GRCh38
Transcripts
NM_006662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7301277 c.574C>T p.R192C Chr.16: 30707653-30707653 on Build GRCh38
335422 c.592A>G p.M198V Chr.16: 30707671-30707671 on Build GRCh38
6907647 c.605T>A p.V202D Chr.16: 30707684-30707684 on Build GRCh38
6144394 c.550C>T p.R184W Chr.16: 30707629-30707629 on Build GRCh38
6575624 c.575G>T p.R192L Chr.16: 30707654-30707654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756665720 Chr.16: 30707521-30707521 on Build GRCh38
rs191080503 Chr.16: 30707915-30707915 on Build GRCh38
rs933830927 Chr.16: 30707451-30707451 on Build GRCh38
rs1016780742 Chr.16: 30707507-30707507 on Build GRCh38
rs942928260 Chr.16: 30707965-30707965 on Build GRCh38
rs533814930 Chr.16: 30707782-30707782 on Build GRCh38
rs925564807 Chr.16: 30707981-30707981 on Build GRCh38
rs972589745 Chr.16: 30707846-30707846 on Build GRCh38
rs1023523307 Chr.16: 30707604-30707604 on Build GRCh38
rs961481363 Chr.16: 30707979-30707979 on Build GRCh38
rs570285253 Chr.16: 30707780-30707782 on Build GRCh38
rs771182909 Chr.16: 30707687-30707687 on Build GRCh38
rs769983508 Chr.16: 30707667-30707667 on Build GRCh38
rs970414220 Chr.16: 30707468-30707468 on Build GRCh38
rs748556220 Chr.16: 30707654-30707654 on Build GRCh38
rs1014889097 Chr.16: 30707892-30707892 on Build GRCh38
rs189183429 Chr.16: 30707773-30707773 on Build GRCh38
rs563925784 Chr.16: 30707768-30707768 on Build GRCh38
rs763088830 Chr.16: 30707542-30707542 on Build GRCh38
rs970267721 Chr.16: 30707637-30707637 on Build GRCh38
rs143206233 Chr.16: 30707835-30707835 on Build GRCh38
rs376999171 Chr.16: 30707745-30707745 on Build GRCh38
rs186571696 Chr.16: 30707847-30707847 on Build GRCh38
rs528022534 Chr.16: 30707843-30707843 on Build GRCh38
rs1014029647 Chr.16: 30707443-30707443 on Build GRCh38
rs1043481804 Chr.16: 30707827-30707827 on Build GRCh38
rs758019448 Chr.16: 30707643-30707643 on Build GRCh38
rs773529599 Chr.16: 30707541-30707541 on Build GRCh38
rs781518661 Chr.16: 30707534-30707534 on Build GRCh38
rs774540726 Chr.16: 30707702-30707702 on Build GRCh38
rs896158848 Chr.16: 30707838-30707838 on Build GRCh38
rs544740086 Chr.16: 30707823-30707823 on Build GRCh38
rs896096717 Chr.16: 30707577-30707577 on Build GRCh38
rs759713844 Chr.16: 30707449-30707449 on Build GRCh38
rs181335500 Chr.16: 30707520-30707520 on Build GRCh38
rs369504517 Chr.16: 30707537-30707537 on Build GRCh38
rs906009644 Chr.16: 30707917-30707917 on Build GRCh38
rs147177072 Chr.16: 30707986-30707986 on Build GRCh38
rs763757740 Chr.16: 30707748-30707748 on Build GRCh38
rs562285842 Chr.16: 30707961-30707961 on Build GRCh38
rs772410409 Chr.16: 30707729-30707729 on Build GRCh38
rs984149226 Chr.16: 30707698-30707698 on Build GRCh38
rs759808882 Chr.16: 30707558-30707558 on Build GRCh38
rs1016999544 Chr.16: 30707725-30707725 on Build GRCh38
rs764004100 Chr.16: 30707735-30707735 on Build GRCh38
rs142017487 Chr.16: 30707480-30707480 on Build GRCh38
rs376390874 Chr.16: 30707572-30707572 on Build GRCh38
rs529482989 Chr.16: 30707983-30707983 on Build GRCh38
rs1024212949 Chr.16: 30707466-30707466 on Build GRCh38
rs150267250 Chr.16: 30707721-30707721 on Build GRCh38
rs979674075 Chr.16: 30707973-30707973 on Build GRCh38
rs762595769 Chr.16: 30707685-30707685 on Build GRCh38
rs72793372 Chr.16: 30707552-30707552 on Build GRCh38
rs750152871 Chr.16: 30707777-30707777 on Build GRCh38
rs1001465380 Chr.16: 30707788-30707788 on Build GRCh38
rs781590472 Chr.16: 30707644-30707644 on Build GRCh38
rs184474420 Chr.16: 30707736-30707736 on Build GRCh38
rs1050063057 Chr.16: 30707458-30707458 on Build GRCh38
rs556337718 Chr.16: 30707831-30707831 on Build GRCh38
rs181922670 Chr.16: 30707844-30707844 on Build GRCh38
rs558749357 Chr.16: 30707790-30707790 on Build GRCh38
rs574895830 Chr.16: 30707832-30707832 on Build GRCh38
rs201747150 Chr.16: 30707561-30707561 on Build GRCh38
rs750129041 Chr.16: 30707639-30707639 on Build GRCh38
rs986903667 Chr.16: 30707615-30707615 on Build GRCh38
rs756793325 Chr.16: 30707625-30707625 on Build GRCh38
rs72793373 Chr.16: 30707837-30707837 on Build GRCh38
rs769034500 Chr.16: 30707532-30707532 on Build GRCh38
rs760587957 Chr.16: 30707598-30707598 on Build GRCh38
rs889630101 Chr.16: 30707485-30707485 on Build GRCh38
rs756436067 Chr.16: 30707787-30707787 on Build GRCh38
rs189739931 Chr.16: 30707452-30707452 on Build GRCh38
rs979930252 Chr.16: 30707483-30707483 on Build GRCh38
rs139272113 Chr.16: 30707819-30707819 on Build GRCh38
rs753653928 Chr.16: 30707609-30707609 on Build GRCh38
rs986441826 Chr.16: 30707929-30707929 on Build GRCh38
rs778348276 Chr.16: 30707629-30707629 on Build GRCh38
rs771092696 Chr.16: 30707543-30707543 on Build GRCh38
rs745484540 Chr.16: 30707525-30707525 on Build GRCh38
rs1005705389 Chr.16: 30707934-30707934 on Build GRCh38
rs1025381885 Chr.16: 30707889-30707889 on Build GRCh38
rs764108371 Chr.16: 30707606-30707606 on Build GRCh38
rs778544087 Chr.16: 30707522-30707522 on Build GRCh38

View Full Product Details