Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49532546-49533122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCBTB1
Entrez Gene ID
55213
Gene Name
RCC1 and BTB domain containing protein 1
Gene Aliases
CLLD7, CLLL7, GLP
UniGene
Hs.508021
Gene Chromosome Location
Chr.13: 49531944-49585606 on Build GRCh38
Transcripts
NM_018191.3 XM_005266441.2 XM_011535133.1 XM_011535134.1 XM_011535135.2

SNP Detail

SNP ID Chromosome Location
rs200694108 Chr.13: 49533083-49533083 on Build GRCh38
rs898996285 Chr.13: 49532617-49532617 on Build GRCh38
rs41284784 Chr.13: 49532646-49532646 on Build GRCh38
rs7995684 Chr.13: 49532819-49532819 on Build GRCh38
rs1049591152 Chr.13: 49532780-49532780 on Build GRCh38
rs41284782 Chr.13: 49532604-49532604 on Build GRCh38
rs71216548 Chr.13: 49533076-49533076 on Build GRCh38
rs142944522 Chr.13: 49532998-49532998 on Build GRCh38
rs1039178781 Chr.13: 49532811-49532811 on Build GRCh38
rs899048708 Chr.13: 49532894-49532894 on Build GRCh38
rs762032989 Chr.13: 49532661-49532661 on Build GRCh38
rs562115260 Chr.13: 49532674-49532674 on Build GRCh38
rs757579708 Chr.13: 49533070-49533071 on Build GRCh38
rs937257030 Chr.13: 49532915-49532915 on Build GRCh38
rs199788678 Chr.13: 49533082-49533082 on Build GRCh38
rs958258156 Chr.13: 49533089-49533089 on Build GRCh38
rs999446365 Chr.13: 49533066-49533066 on Build GRCh38
rs140027458 Chr.13: 49533071-49533085 on Build GRCh38
rs1046034 Chr.13: 49532675-49532675 on Build GRCh38
rs974852061 Chr.13: 49532812-49532812 on Build GRCh38
rs917842303 Chr.13: 49532692-49532692 on Build GRCh38
rs199527104 Chr.13: 49533085-49533085 on Build GRCh38
rs1000644636 Chr.13: 49532875-49532876 on Build GRCh38
rs950680474 Chr.13: 49532960-49532960 on Build GRCh38
rs959449416 Chr.13: 49532677-49532677 on Build GRCh38
rs1032317981 Chr.13: 49533070-49533070 on Build GRCh38
rs1055614566 Chr.13: 49532931-49532931 on Build GRCh38
rs929414020 Chr.13: 49532777-49532777 on Build GRCh38
rs1025095459 Chr.13: 49533008-49533008 on Build GRCh38
rs919281423 Chr.13: 49532952-49532952 on Build GRCh38
rs544584726 Chr.13: 49532830-49532830 on Build GRCh38
rs1042072068 Chr.13: 49532853-49532853 on Build GRCh38
rs1001329923 Chr.13: 49532632-49532632 on Build GRCh38
rs200696623 Chr.13: 49533080-49533080 on Build GRCh38
rs534636863 Chr.13: 49532570-49532570 on Build GRCh38
rs868585445 Chr.13: 49532773-49532773 on Build GRCh38
rs902595353 Chr.13: 49533060-49533060 on Build GRCh38
rs16659 Chr.13: 49533076-49533090 on Build GRCh38
rs193232071 Chr.13: 49532891-49532891 on Build GRCh38
rs978746003 Chr.13: 49532709-49532709 on Build GRCh38
rs886128528 Chr.13: 49532598-49532598 on Build GRCh38
rs199588684 Chr.13: 49533078-49533078 on Build GRCh38
rs1025379195 Chr.13: 49532697-49532697 on Build GRCh38
rs907526021 Chr.13: 49532868-49532868 on Build GRCh38
rs958288622 Chr.13: 49532755-49532755 on Build GRCh38
rs1005195916 Chr.13: 49532599-49532599 on Build GRCh38
rs767227992 Chr.13: 49533000-49533000 on Build GRCh38
rs1128767 Chr.13: 49533095-49533095 on Build GRCh38
rs533230938 Chr.13: 49532890-49532890 on Build GRCh38
rs573592323 Chr.13: 49532633-49532633 on Build GRCh38
rs1032217024 Chr.13: 49532673-49532673 on Build GRCh38
rs1032514565 Chr.13: 49532731-49532731 on Build GRCh38
rs1032494404 Chr.13: 49532876-49532876 on Build GRCh38
rs966767690 Chr.13: 49532699-49532699 on Build GRCh38
rs867929386 Chr.13: 49532873-49532873 on Build GRCh38
rs546456410 Chr.13: 49532595-49532595 on Build GRCh38
rs1013691634 Chr.13: 49532679-49532679 on Build GRCh38
rs902186781 Chr.13: 49532871-49532871 on Build GRCh38
rs943545231 Chr.13: 49532800-49532800 on Build GRCh38
rs970642870 Chr.13: 49532896-49532896 on Build GRCh38
rs933046724 Chr.13: 49532837-49532837 on Build GRCh38
rs909408751 Chr.13: 49532789-49532789 on Build GRCh38
rs909306380 Chr.13: 49533062-49533062 on Build GRCh38
rs972640607 Chr.13: 49532934-49532934 on Build GRCh38
rs562676216 Chr.13: 49532840-49532840 on Build GRCh38
rs1037657080 Chr.13: 49532608-49532608 on Build GRCh38
rs201058461 Chr.13: 49533084-49533084 on Build GRCh38
rs200403352 Chr.13: 49533076-49533076 on Build GRCh38
rs201154849 Chr.13: 49533077-49533077 on Build GRCh38

View Full Product Details