Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72272402-72272953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS4X
Entrez Gene ID
6191
Gene Name
ribosomal protein S4, X-linked
Gene Aliases
CCG2, DXS306, RPS4, S4, SCAR, SCR10
UniGene
Hs.118076
Gene Chromosome Location
Chr.X: 72272603-72277291 on Build GRCh38
Transcripts
NM_001007.4

SNP Detail

SNP ID Chromosome Location
rs772547575 Chr.X: 72272808-72272809 on Build GRCh38
rs373139828 Chr.X: 72272660-72272660 on Build GRCh38
rs774926516 Chr.X: 72272776-72272776 on Build GRCh38
rs773305995 Chr.X: 72272519-72272519 on Build GRCh38
rs755197506 Chr.X: 72272666-72272666 on Build GRCh38
rs113927316 Chr.X: 72272602-72272602 on Build GRCh38
rs372262584 Chr.X: 72272498-72272498 on Build GRCh38
rs910926687 Chr.X: 72272762-72272762 on Build GRCh38
rs773765328 Chr.X: 72272819-72272820 on Build GRCh38
rs10783 Chr.X: 72272730-72272730 on Build GRCh38
rs747526505 Chr.X: 72272763-72272763 on Build GRCh38
rs11545423 Chr.X: 72272754-72272754 on Build GRCh38
rs182095808 Chr.X: 72272692-72272692 on Build GRCh38
rs907138915 Chr.X: 72272893-72272893 on Build GRCh38
rs762491291 Chr.X: 72272777-72272777 on Build GRCh38
rs900311156 Chr.X: 72272583-72272585 on Build GRCh38
rs762312428 Chr.X: 72272538-72272538 on Build GRCh38
rs968685805 Chr.X: 72272828-72272828 on Build GRCh38
rs780058676 Chr.X: 72272658-72272658 on Build GRCh38
rs767368222 Chr.X: 72272625-72272625 on Build GRCh38
rs778045446 Chr.X: 72272761-72272761 on Build GRCh38
rs904190589 Chr.X: 72272612-72272612 on Build GRCh38
rs185166384 Chr.X: 72272891-72272891 on Build GRCh38
rs768238370 Chr.X: 72272796-72272796 on Build GRCh38
rs1009974335 Chr.X: 72272681-72272682 on Build GRCh38
rs761674944 Chr.X: 72272622-72272622 on Build GRCh38
rs761493456 Chr.X: 72272816-72272816 on Build GRCh38
rs376367408 Chr.X: 72272797-72272797 on Build GRCh38
rs376839099 Chr.X: 72272636-72272636 on Build GRCh38
rs1056475803 Chr.X: 72272907-72272907 on Build GRCh38
rs1051137585 Chr.X: 72272650-72272650 on Build GRCh38
rs942496953 Chr.X: 72272801-72272801 on Build GRCh38
rs183857827 Chr.X: 72272561-72272561 on Build GRCh38
rs928932275 Chr.X: 72272463-72272463 on Build GRCh38
rs767437734 Chr.X: 72272819-72272819 on Build GRCh38
rs997366738 Chr.X: 72272633-72272633 on Build GRCh38
rs748504019 Chr.X: 72272775-72272775 on Build GRCh38
rs779367864 Chr.X: 72272657-72272658 on Build GRCh38
rs747826302 Chr.X: 72272582-72272582 on Build GRCh38
rs772458199 Chr.X: 72272760-72272760 on Build GRCh38
rs191773447 Chr.X: 72272559-72272559 on Build GRCh38
rs767848530 Chr.X: 72272573-72272573 on Build GRCh38
rs757217550 Chr.X: 72272905-72272905 on Build GRCh38
rs111354545 Chr.X: 72272601-72272601 on Build GRCh38
rs12564 Chr.X: 72272656-72272656 on Build GRCh38
rs765922668 Chr.X: 72272558-72272558 on Build GRCh38
rs748321540 Chr.X: 72272693-72272693 on Build GRCh38
rs1804391 Chr.X: 72272707-72272707 on Build GRCh38
rs772338492 Chr.X: 72272890-72272890 on Build GRCh38
rs941983223 Chr.X: 72272574-72272576 on Build GRCh38
rs1001101562 Chr.X: 72272851-72272855 on Build GRCh38
rs1804390 Chr.X: 72272679-72272679 on Build GRCh38
rs990384732 Chr.X: 72272863-72272863 on Build GRCh38
rs113969908 Chr.X: 72272484-72272484 on Build GRCh38
rs914433610 Chr.X: 72272874-72272874 on Build GRCh38
rs777512684 Chr.X: 72272480-72272483 on Build GRCh38
rs189512872 Chr.X: 72272574-72272574 on Build GRCh38
rs750393551 Chr.X: 72272635-72272635 on Build GRCh38

View Full Product Details