Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98515685-98516279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDND1
Entrez Gene ID
56650
Gene Name
claudin domain containing 1
Gene Aliases
C3orf4, GENX-3745
UniGene
Hs.531371
Gene Chromosome Location
Chr.3: 98515473-98523066 on Build GRCh38
Transcripts
NM_001040181.1 NM_001040182.1 NM_001040183.1 NM_001040199.1 NM_001040200.1 NM_019895.2

SNP Detail

SNP ID Chromosome Location
rs887289040 Chr.3: 98515987-98515987 on Build GRCh38
rs189921456 Chr.3: 98516109-98516109 on Build GRCh38
rs576094399 Chr.3: 98515969-98515969 on Build GRCh38
rs950199966 Chr.3: 98515964-98515964 on Build GRCh38
rs886211382 Chr.3: 98515760-98515760 on Build GRCh38
rs978888913 Chr.3: 98515835-98515835 on Build GRCh38
rs750496714 Chr.3: 98515815-98515815 on Build GRCh38
rs566279438 Chr.3: 98515866-98515866 on Build GRCh38
rs890867040 Chr.3: 98515842-98515842 on Build GRCh38
rs1028896935 Chr.3: 98516027-98516027 on Build GRCh38
rs979223664 Chr.3: 98516111-98516111 on Build GRCh38
rs1050854285 Chr.3: 98515838-98515838 on Build GRCh38
rs967428210 Chr.3: 98515829-98515829 on Build GRCh38
rs939755557 Chr.3: 98516170-98516170 on Build GRCh38
rs975440081 Chr.3: 98516071-98516071 on Build GRCh38
rs1020107538 Chr.3: 98516055-98516055 on Build GRCh38
rs144826203 Chr.3: 98516126-98516126 on Build GRCh38
rs937368733 Chr.3: 98515850-98515850 on Build GRCh38
rs1028945342 Chr.3: 98515806-98515806 on Build GRCh38
rs745516983 Chr.3: 98515819-98515819 on Build GRCh38
rs940411153 Chr.3: 98515995-98515995 on Build GRCh38
rs11553644 Chr.3: 98515754-98515754 on Build GRCh38
rs765660913 Chr.3: 98515820-98515820 on Build GRCh38
rs1004491109 Chr.3: 98515766-98515766 on Build GRCh38
rs752425412 Chr.3: 98516160-98516161 on Build GRCh38
rs565265888 Chr.3: 98516029-98516029 on Build GRCh38
rs753166627 Chr.3: 98515822-98515822 on Build GRCh38
rs922189058 Chr.3: 98516102-98516102 on Build GRCh38
rs999524724 Chr.3: 98516019-98516019 on Build GRCh38
rs759118215 Chr.3: 98515837-98515840 on Build GRCh38
rs912209531 Chr.3: 98516130-98516130 on Build GRCh38
rs898792594 Chr.3: 98516006-98516006 on Build GRCh38
rs995839538 Chr.3: 98515966-98515966 on Build GRCh38
rs543574749 Chr.3: 98516234-98516234 on Build GRCh38
rs771688593 Chr.3: 98516001-98516001 on Build GRCh38
rs773641078 Chr.3: 98515785-98515785 on Build GRCh38
rs936920048 Chr.3: 98515708-98515708 on Build GRCh38
rs182491245 Chr.3: 98516247-98516247 on Build GRCh38
rs532937761 Chr.3: 98516218-98516218 on Build GRCh38
rs556215959 Chr.3: 98515847-98515847 on Build GRCh38
rs756793614 Chr.3: 98515711-98515711 on Build GRCh38
rs1014406823 Chr.3: 98515731-98515731 on Build GRCh38
rs898289727 Chr.3: 98515962-98515962 on Build GRCh38
rs553759249 Chr.3: 98515777-98515777 on Build GRCh38
rs780366293 Chr.3: 98516008-98516008 on Build GRCh38
rs539589594 Chr.3: 98515831-98515831 on Build GRCh38
rs958952870 Chr.3: 98516131-98516131 on Build GRCh38
rs564119365 Chr.3: 98516144-98516144 on Build GRCh38
rs777015234 Chr.3: 98515776-98515776 on Build GRCh38
rs917536677 Chr.3: 98515918-98515918 on Build GRCh38
rs878871359 Chr.3: 98516164-98516164 on Build GRCh38
rs991528078 Chr.3: 98516157-98516157 on Build GRCh38
rs917485824 Chr.3: 98516200-98516200 on Build GRCh38
rs186015823 Chr.3: 98515790-98515790 on Build GRCh38
rs1008833657 Chr.3: 98516042-98516042 on Build GRCh38
rs983012515 Chr.3: 98516249-98516249 on Build GRCh38
rs139437845 Chr.3: 98515975-98515975 on Build GRCh38
rs534177984 Chr.3: 98515769-98515769 on Build GRCh38
rs1037291778 Chr.3: 98515912-98515912 on Build GRCh38
rs895178679 Chr.3: 98515990-98515990 on Build GRCh38
rs932301646 Chr.3: 98515788-98515788 on Build GRCh38
rs895951692 Chr.3: 98515715-98515715 on Build GRCh38
rs1046127222 Chr.3: 98515735-98515735 on Build GRCh38
rs1056440047 Chr.3: 98515916-98515916 on Build GRCh38
rs561684687 Chr.3: 98516073-98516073 on Build GRCh38

View Full Product Details