Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 137939136-137939695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13B
Entrez Gene ID
51306
Gene Name
family with sequence similarity 13 member B
Gene Aliases
ARHGAP49, C5orf5, FAM13B1, KHCHP, N61
UniGene
Hs.567453
Gene Chromosome Location
Chr.5: 137937909-138051961 on Build GRCh38
Transcripts
NM_001101800.2 NM_001101801.2 NM_016603.3 XM_005272007.1 XM_006714647.1 XM_006714648.1 XM_006714649.1 XM_011543448.1 XM_011543449.1 XM_011543450.1 XM_017009549.1 XM_017009550.1 XM_017009551.1 NM_001258448.1 NM_001258449.1 NM_001300921.1 NM_014386.3 XM_017009343.1

Gene Symbol
PKD2L2
Entrez Gene ID
27039
Gene Name
polycystin 2 like 2, transient receptor potential cation channel
Gene Aliases
TRPP5
UniGene
Hs.716884
Gene Chromosome Location
Chr.5: 137889420-137942747 on Build GRCh38
Transcripts
NM_001101800.2 NM_001101801.2 NM_016603.3 XM_005272007.1 XM_006714647.1 XM_006714648.1 XM_006714649.1 XM_011543448.1 XM_011543449.1 XM_011543450.1 XM_017009549.1 XM_017009550.1 XM_017009551.1 NM_001258448.1 NM_001258449.1 NM_001300921.1 NM_014386.3 XM_017009343.1

SNP Detail

SNP ID Chromosome Location
rs755554402 Chr.5: 137939623-137939623 on Build GRCh38
rs568297439 Chr.5: 137939344-137939344 on Build GRCh38
rs570627432 Chr.5: 137939258-137939258 on Build GRCh38
rs947166550 Chr.5: 137939303-137939303 on Build GRCh38
rs971956756 Chr.5: 137939186-137939186 on Build GRCh38
rs891455501 Chr.5: 137939227-137939227 on Build GRCh38
rs896201307 Chr.5: 137939336-137939336 on Build GRCh38
rs1023681657 Chr.5: 137939461-137939461 on Build GRCh38
rs551828739 Chr.5: 137939595-137939595 on Build GRCh38
rs982209763 Chr.5: 137939177-137939177 on Build GRCh38
rs1042598175 Chr.5: 137939371-137939371 on Build GRCh38
rs778489092 Chr.5: 137939309-137939309 on Build GRCh38
rs3209743 Chr.5: 137939214-137939214 on Build GRCh38
rs139187964 Chr.5: 137939552-137939552 on Build GRCh38
rs1001131950 Chr.5: 137939414-137939414 on Build GRCh38
rs925461629 Chr.5: 137939663-137939663 on Build GRCh38
rs555642739 Chr.5: 137939540-137939540 on Build GRCh38
rs945177289 Chr.5: 137939604-137939604 on Build GRCh38
rs917569108 Chr.5: 137939598-137939598 on Build GRCh38
rs748836713 Chr.5: 137939178-137939178 on Build GRCh38
rs544347055 Chr.5: 137939559-137939559 on Build GRCh38
rs546815173 Chr.5: 137939338-137939338 on Build GRCh38
rs947081384 Chr.5: 137939567-137939567 on Build GRCh38
rs945110556 Chr.5: 137939355-137939355 on Build GRCh38
rs7711607 Chr.5: 137939569-137939569 on Build GRCh38
rs961524272 Chr.5: 137939501-137939501 on Build GRCh38
rs114637319 Chr.5: 137939179-137939179 on Build GRCh38
rs3777118 Chr.5: 137939426-137939426 on Build GRCh38
rs145927578 Chr.5: 137939527-137939527 on Build GRCh38
rs915442062 Chr.5: 137939561-137939561 on Build GRCh38
rs900271878 Chr.5: 137939605-137939605 on Build GRCh38
rs1053229188 Chr.5: 137939220-137939220 on Build GRCh38
rs914979592 Chr.5: 137939447-137939447 on Build GRCh38
rs569495800 Chr.5: 137939427-137939427 on Build GRCh38
rs890391790 Chr.5: 137939422-137939422 on Build GRCh38
rs904012292 Chr.5: 137939375-137939375 on Build GRCh38
rs1141538 Chr.5: 137939510-137939510 on Build GRCh38
rs973335904 Chr.5: 137939454-137939454 on Build GRCh38
rs143662048 Chr.5: 137939518-137939518 on Build GRCh38
rs545062568 Chr.5: 137939164-137939164 on Build GRCh38
rs182224747 Chr.5: 137939379-137939379 on Build GRCh38
rs1042504673 Chr.5: 137939647-137939647 on Build GRCh38
rs930585714 Chr.5: 137939227-137939227 on Build GRCh38
rs539080029 Chr.5: 137939428-137939428 on Build GRCh38
rs926501408 Chr.5: 137939542-137939542 on Build GRCh38
rs935571253 Chr.5: 137939187-137939187 on Build GRCh38
rs969414171 Chr.5: 137939462-137939462 on Build GRCh38
rs115109738 Chr.5: 137939333-137939333 on Build GRCh38
rs546994832 Chr.5: 137939378-137939378 on Build GRCh38
rs931701394 Chr.5: 137939622-137939622 on Build GRCh38
rs959304605 Chr.5: 137939566-137939566 on Build GRCh38

View Full Product Details