Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90384855-90385431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP10
Entrez Gene ID
85865
Gene Name
GTP binding protein 10
Gene Aliases
ObgH2, UG0751c10
UniGene
Hs.593547
Gene Chromosome Location
Chr.7: 90346665-90391455 on Build GRCh38
Transcripts
NM_001042717.2 NM_033107.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6333050 c.997G>T p.E333* Chr.7: 90384987-90384987 on Build GRCh38
4807457 c.921A>T p.E307D Chr.7: 90384911-90384911 on Build GRCh38
4689221 c.1058C>T p.A353V Chr.7: 90385048-90385048 on Build GRCh38
2863350 c.919_920insA p.N309fs*37 Chr.7: 90384909-90384910 on Build GRCh38
4689220 c.920delA p.N309fs*2 Chr.7: 90384910-90384910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781172282 Chr.7: 90384941-90384941 on Build GRCh38
rs756608010 Chr.7: 90384901-90384901 on Build GRCh38
rs758529195 Chr.7: 90385071-90385071 on Build GRCh38
rs551715366 Chr.7: 90385063-90385063 on Build GRCh38
rs757620202 Chr.7: 90385054-90385054 on Build GRCh38
rs984959143 Chr.7: 90385346-90385346 on Build GRCh38
rs534740402 Chr.7: 90385258-90385258 on Build GRCh38
rs1012236470 Chr.7: 90385329-90385329 on Build GRCh38
rs999573164 Chr.7: 90384948-90384948 on Build GRCh38
rs533190751 Chr.7: 90385037-90385037 on Build GRCh38
rs1028471710 Chr.7: 90385228-90385228 on Build GRCh38
rs991284170 Chr.7: 90385007-90385007 on Build GRCh38
rs778015258 Chr.7: 90384910-90384910 on Build GRCh38
rs567455204 Chr.7: 90385179-90385179 on Build GRCh38
rs997070167 Chr.7: 90385358-90385358 on Build GRCh38
rs374949799 Chr.7: 90385118-90385118 on Build GRCh38
rs774027772 Chr.7: 90385009-90385009 on Build GRCh38
rs555447082 Chr.7: 90385399-90385399 on Build GRCh38
rs34277629 Chr.7: 90385350-90385351 on Build GRCh38
rs556870701 Chr.7: 90385310-90385310 on Build GRCh38
rs767125180 Chr.7: 90385035-90385035 on Build GRCh38
rs772885312 Chr.7: 90384959-90384959 on Build GRCh38
rs754166184 Chr.7: 90384924-90384924 on Build GRCh38
rs1011400127 Chr.7: 90385237-90385237 on Build GRCh38
rs373738415 Chr.7: 90385177-90385177 on Build GRCh38
rs17867238 Chr.7: 90385279-90385279 on Build GRCh38
rs369162751 Chr.7: 90385169-90385169 on Build GRCh38
rs745885335 Chr.7: 90384910-90384910 on Build GRCh38
rs745531795 Chr.7: 90385123-90385123 on Build GRCh38
rs1028501368 Chr.7: 90385375-90385375 on Build GRCh38
rs267601622 Chr.7: 90384957-90384957 on Build GRCh38
rs748062317 Chr.7: 90384942-90384942 on Build GRCh38
rs1013827347 Chr.7: 90384998-90384998 on Build GRCh38
rs775174019 Chr.7: 90385133-90385133 on Build GRCh38
rs949309564 Chr.7: 90385284-90385284 on Build GRCh38
rs1020279104 Chr.7: 90385203-90385203 on Build GRCh38
rs745997084 Chr.7: 90384936-90384936 on Build GRCh38
rs148357215 Chr.7: 90385155-90385155 on Build GRCh38
rs374680644 Chr.7: 90385042-90385042 on Build GRCh38
rs201866556 Chr.7: 90385066-90385066 on Build GRCh38
rs762765294 Chr.7: 90384975-90384975 on Build GRCh38
rs770957088 Chr.7: 90384880-90384883 on Build GRCh38
rs886812416 Chr.7: 90385325-90385325 on Build GRCh38
rs762150802 Chr.7: 90385159-90385159 on Build GRCh38
rs749081567 Chr.7: 90385116-90385116 on Build GRCh38
rs754176936 Chr.7: 90385048-90385048 on Build GRCh38
rs924636055 Chr.7: 90385252-90385252 on Build GRCh38
rs978807346 Chr.7: 90385244-90385244 on Build GRCh38
rs770142683 Chr.7: 90385121-90385126 on Build GRCh38
rs747023041 Chr.7: 90385091-90385091 on Build GRCh38
rs984947902 Chr.7: 90385111-90385111 on Build GRCh38
rs968803078 Chr.7: 90385235-90385235 on Build GRCh38
rs370318776 Chr.7: 90385135-90385135 on Build GRCh38
rs781318104 Chr.7: 90384909-90384910 on Build GRCh38
rs772580798 Chr.7: 90385343-90385343 on Build GRCh38
rs377658515 Chr.7: 90384935-90384935 on Build GRCh38
rs1004274204 Chr.7: 90385332-90385332 on Build GRCh38
rs904970826 Chr.7: 90385099-90385099 on Build GRCh38
rs1040046125 Chr.7: 90385162-90385162 on Build GRCh38
rs772107421 Chr.7: 90384938-90384938 on Build GRCh38
rs17863999 Chr.7: 90385094-90385094 on Build GRCh38
rs769647269 Chr.7: 90385092-90385092 on Build GRCh38
rs370884460 Chr.7: 90384986-90384986 on Build GRCh38
rs917812787 Chr.7: 90385283-90385283 on Build GRCh38
rs17869357 Chr.7: 90385317-90385317 on Build GRCh38
rs372736306 Chr.7: 90385342-90385342 on Build GRCh38
rs769091506 Chr.7: 90385017-90385017 on Build GRCh38
rs764624426 Chr.7: 90385043-90385043 on Build GRCh38
rs747347928 Chr.7: 90385004-90385004 on Build GRCh38
rs1015064442 Chr.7: 90385378-90385378 on Build GRCh38
rs771785955 Chr.7: 90385130-90385130 on Build GRCh38
rs780119986 Chr.7: 90385086-90385086 on Build GRCh38
rs17869500 Chr.7: 90385289-90385289 on Build GRCh38
rs892876601 Chr.7: 90385227-90385227 on Build GRCh38
rs765619753 Chr.7: 90385168-90385168 on Build GRCh38
rs764557944 Chr.7: 90385141-90385141 on Build GRCh38
rs769659138 Chr.7: 90384954-90384954 on Build GRCh38
rs115223124 Chr.7: 90385299-90385299 on Build GRCh38
rs757453617 Chr.7: 90384925-90384925 on Build GRCh38
rs1023759643 Chr.7: 90385335-90385335 on Build GRCh38
rs773936273 Chr.7: 90385122-90385122 on Build GRCh38

View Full Product Details