Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143264862-143265442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTK1
Entrez Gene ID
373156
Gene Name
glutathione S-transferase kappa 1
Gene Aliases
GST, GST 13-13, GST13, GST13-13, GSTK1-1, hGSTK1
UniGene
Hs.390667
Gene Chromosome Location
Chr.7: 143263429-143269129 on Build GRCh38
Transcripts
NM_001143679.1 NM_001143680.1 NM_001143681.1 NM_015917.2 NR_133932.1

Gene Symbol
LOC105375545
Entrez Gene ID
105375545
Gene Name
uncharacterized LOC105375545
Gene Aliases
-
UniGene
Hs.699839
Gene Chromosome Location
Chr.7: 143255299-143286334 on Build GRCh38
Transcripts
NM_001143679.1 NM_001143680.1 NM_001143681.1 NM_015917.2 NR_133932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3662999 c.469C>G p.L157V Chr.7: 143265177-143265177 on Build GRCh38
7370474 c.323T>C p.L108S Chr.7: 143265031-143265031 on Build GRCh38
3635163 c.354C>T p.S118S Chr.7: 143265062-143265062 on Build GRCh38
4442227 c.301C>T p.R101C Chr.7: 143265009-143265009 on Build GRCh38
3698234 c.373G>A p.V125I Chr.7: 143265081-143265081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764917925 Chr.7: 143265226-143265226 on Build GRCh38
rs778290959 Chr.7: 143265176-143265176 on Build GRCh38
rs201201238 Chr.7: 143265103-143265103 on Build GRCh38
rs748132282 Chr.7: 143265329-143265329 on Build GRCh38
rs143879685 Chr.7: 143264895-143264895 on Build GRCh38
rs145599642 Chr.7: 143265013-143265014 on Build GRCh38
rs770163021 Chr.7: 143265186-143265186 on Build GRCh38
rs758934722 Chr.7: 143265079-143265079 on Build GRCh38
rs748802330 Chr.7: 143264949-143264949 on Build GRCh38
rs747358924 Chr.7: 143265024-143265024 on Build GRCh38
rs914042974 Chr.7: 143265088-143265088 on Build GRCh38
rs752226657 Chr.7: 143265172-143265172 on Build GRCh38
rs201961320 Chr.7: 143265110-143265110 on Build GRCh38
rs770920498 Chr.7: 143265340-143265340 on Build GRCh38
rs1043366349 Chr.7: 143265398-143265398 on Build GRCh38
rs147245180 Chr.7: 143265031-143265031 on Build GRCh38
rs745702032 Chr.7: 143265348-143265348 on Build GRCh38
rs759227977 Chr.7: 143265166-143265166 on Build GRCh38
rs958507949 Chr.7: 143265061-143265061 on Build GRCh38
rs1015821653 Chr.7: 143264962-143264962 on Build GRCh38
rs1029504347 Chr.7: 143265245-143265245 on Build GRCh38
rs776410147 Chr.7: 143265343-143265343 on Build GRCh38
rs549430902 Chr.7: 143265008-143265008 on Build GRCh38
rs779460183 Chr.7: 143264999-143264999 on Build GRCh38
rs763238862 Chr.7: 143265188-143265188 on Build GRCh38
rs575868481 Chr.7: 143265071-143265071 on Build GRCh38
rs370713285 Chr.7: 143265189-143265189 on Build GRCh38
rs888475206 Chr.7: 143265146-143265146 on Build GRCh38
rs750969769 Chr.7: 143265252-143265252 on Build GRCh38
rs752132541 Chr.7: 143265078-143265078 on Build GRCh38
rs766181029 Chr.7: 143265155-143265155 on Build GRCh38
rs753093362 Chr.7: 143265030-143265030 on Build GRCh38
rs184706110 Chr.7: 143265352-143265352 on Build GRCh38
rs371788051 Chr.7: 143265208-143265208 on Build GRCh38
rs1029453280 Chr.7: 143265010-143265010 on Build GRCh38
rs149801284 Chr.7: 143265232-143265232 on Build GRCh38
rs375554522 Chr.7: 143265095-143265095 on Build GRCh38
rs763609277 Chr.7: 143265242-143265242 on Build GRCh38
rs916543318 Chr.7: 143264890-143264890 on Build GRCh38
rs780493614 Chr.7: 143265296-143265296 on Build GRCh38
rs773083480 Chr.7: 143265201-143265201 on Build GRCh38
rs957088188 Chr.7: 143265379-143265379 on Build GRCh38
rs1055022153 Chr.7: 143265131-143265131 on Build GRCh38
rs1043865558 Chr.7: 143265072-143265072 on Build GRCh38
rs746117551 Chr.7: 143265105-143265105 on Build GRCh38
rs767532962 Chr.7: 143265185-143265185 on Build GRCh38
rs757880637 Chr.7: 143265175-143265175 on Build GRCh38
rs144260278 Chr.7: 143265058-143265058 on Build GRCh38
rs757110852 Chr.7: 143265081-143265081 on Build GRCh38
rs776516600 Chr.7: 143265217-143265217 on Build GRCh38
rs747935768 Chr.7: 143265020-143265020 on Build GRCh38
rs1021244748 Chr.7: 143265406-143265406 on Build GRCh38
rs762516112 Chr.7: 143265238-143265238 on Build GRCh38
rs754482839 Chr.7: 143264967-143264967 on Build GRCh38
rs748019638 Chr.7: 143264991-143264991 on Build GRCh38
rs772188539 Chr.7: 143265121-143265121 on Build GRCh38
rs749511359 Chr.7: 143265297-143265297 on Build GRCh38
rs748638302 Chr.7: 143265007-143265007 on Build GRCh38
rs780405817 Chr.7: 143265187-143265187 on Build GRCh38
rs773256350 Chr.7: 143265127-143265127 on Build GRCh38
rs779052336 Chr.7: 143265318-143265318 on Build GRCh38
rs781624647 Chr.7: 143265182-143265182 on Build GRCh38
rs946369808 Chr.7: 143265389-143265389 on Build GRCh38
rs1036943026 Chr.7: 143265006-143265006 on Build GRCh38
rs753526449 Chr.7: 143265065-143265065 on Build GRCh38
rs953742960 Chr.7: 143265285-143265285 on Build GRCh38
rs756520317 Chr.7: 143265276-143265276 on Build GRCh38
rs1005483424 Chr.7: 143264904-143264904 on Build GRCh38
rs199501326 Chr.7: 143265090-143265090 on Build GRCh38
rs753368454 Chr.7: 143264989-143264989 on Build GRCh38
rs1036981232 Chr.7: 143265192-143265192 on Build GRCh38
rs777393754 Chr.7: 143265223-143265223 on Build GRCh38
rs879097453 Chr.7: 143265168-143265168 on Build GRCh38
rs759277912 Chr.7: 143265224-143265224 on Build GRCh38
rs746976978 Chr.7: 143265216-143265216 on Build GRCh38
rs922412350 Chr.7: 143265301-143265301 on Build GRCh38
rs753515701 Chr.7: 143265164-143265164 on Build GRCh38
rs546731104 Chr.7: 143265057-143265057 on Build GRCh38
rs562656700 Chr.7: 143265397-143265397 on Build GRCh38
rs148563300 Chr.7: 143265044-143265044 on Build GRCh38
rs1005865188 Chr.7: 143265165-143265165 on Build GRCh38
rs562742677 Chr.7: 143264894-143264894 on Build GRCh38
rs145743461 Chr.7: 143265359-143265359 on Build GRCh38
rs984932513 Chr.7: 143265225-143265225 on Build GRCh38
rs757814489 Chr.7: 143265254-143265254 on Build GRCh38
rs368166019 Chr.7: 143265051-143265051 on Build GRCh38
rs763081198 Chr.7: 143265116-143265116 on Build GRCh38
rs1006663004 Chr.7: 143265336-143265336 on Build GRCh38
rs769906717 Chr.7: 143265042-143265042 on Build GRCh38
rs200469041 Chr.7: 143265021-143265021 on Build GRCh38
rs544446184 Chr.7: 143265360-143265360 on Build GRCh38
rs771944896 Chr.7: 143265185-143265186 on Build GRCh38
rs767523378 Chr.7: 143265054-143265054 on Build GRCh38
rs139754207 Chr.7: 143264889-143264889 on Build GRCh38
rs140666496 Chr.7: 143265128-143265128 on Build GRCh38
rs764825938 Chr.7: 143265171-143265171 on Build GRCh38
rs775743103 Chr.7: 143265111-143265111 on Build GRCh38
rs538829875 Chr.7: 143265063-143265063 on Build GRCh38
rs754501668 Chr.7: 143265068-143265068 on Build GRCh38
rs771294590 Chr.7: 143265091-143265091 on Build GRCh38
rs767864250 Chr.7: 143265275-143265275 on Build GRCh38
rs759573720 Chr.7: 143265038-143265038 on Build GRCh38

View Full Product Details