Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 30629878-30630356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO8
Entrez Gene ID
10526
Gene Name
importin 8
Gene Aliases
RANBP8
UniGene
Hs.505136
Gene Chromosome Location
Chr.12: 30628981-30695995 on Build GRCh38
Transcripts
NM_001190995.1 NM_006390.3 XM_017018691.1 XM_017018692.1

SNP Detail

SNP ID Chromosome Location
rs553612800 Chr.12: 30630146-30630146 on Build GRCh38
rs988435494 Chr.12: 30630120-30630120 on Build GRCh38
rs769350500 Chr.12: 30630271-30630271 on Build GRCh38
rs542296745 Chr.12: 30630232-30630232 on Build GRCh38
rs936840027 Chr.12: 30630151-30630151 on Build GRCh38
rs150097550 Chr.12: 30629950-30629950 on Build GRCh38
rs538141259 Chr.12: 30630038-30630038 on Build GRCh38
rs371742558 Chr.12: 30630136-30630136 on Build GRCh38
rs1029580332 Chr.12: 30630306-30630306 on Build GRCh38
rs905672117 Chr.12: 30630157-30630157 on Build GRCh38
rs935703384 Chr.12: 30630113-30630113 on Build GRCh38
rs567611336 Chr.12: 30630012-30630012 on Build GRCh38
rs556353229 Chr.12: 30630070-30630070 on Build GRCh38
rs987139273 Chr.12: 30630031-30630031 on Build GRCh38
rs368955101 Chr.12: 30630315-30630315 on Build GRCh38
rs953539762 Chr.12: 30630322-30630322 on Build GRCh38
rs1004925529 Chr.12: 30630256-30630256 on Build GRCh38
rs976678116 Chr.12: 30629958-30629958 on Build GRCh38
rs1015048190 Chr.12: 30629945-30629945 on Build GRCh38
rs141663832 Chr.12: 30629985-30629985 on Build GRCh38
rs4082413 Chr.12: 30630250-30630250 on Build GRCh38
rs1048972773 Chr.12: 30630236-30630236 on Build GRCh38
rs34994890 Chr.12: 30630191-30630192 on Build GRCh38
rs997589396 Chr.12: 30630284-30630284 on Build GRCh38
rs921526083 Chr.12: 30630145-30630145 on Build GRCh38
rs146112636 Chr.12: 30630132-30630132 on Build GRCh38
rs1002214582 Chr.12: 30630295-30630295 on Build GRCh38
rs369984165 Chr.12: 30630319-30630319 on Build GRCh38
rs193174208 Chr.12: 30630140-30630140 on Build GRCh38
rs190244536 Chr.12: 30629938-30629938 on Build GRCh38
rs951985953 Chr.12: 30629911-30629911 on Build GRCh38
rs574849501 Chr.12: 30630182-30630182 on Build GRCh38
rs1002099917 Chr.12: 30629954-30629954 on Build GRCh38
rs994673833 Chr.12: 30629961-30629961 on Build GRCh38
rs1026059317 Chr.12: 30629967-30629967 on Build GRCh38
rs906500035 Chr.12: 30630280-30630280 on Build GRCh38
rs1045987679 Chr.12: 30630155-30630155 on Build GRCh38
rs975606992 Chr.12: 30630137-30630137 on Build GRCh38
rs138432570 Chr.12: 30629959-30629959 on Build GRCh38
rs979564335 Chr.12: 30630325-30630325 on Build GRCh38
rs759313812 Chr.12: 30630269-30630269 on Build GRCh38
rs911571161 Chr.12: 30630126-30630126 on Build GRCh38

View Full Product Details