Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 99645771-99646357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALMD
Entrez Gene ID
54873
Gene Name
palmdelphin
Gene Aliases
C1orf11, PALML
UniGene
Hs.483993
Gene Chromosome Location
Chr.1: 99645875-99694541 on Build GRCh38
Transcripts
NM_017734.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1952836 c.36G>T p.Q12H Chr.1: 99646353-99646353 on Build GRCh38
674758 c.39C>T p.A13A Chr.1: 99646356-99646356 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953377002 Chr.1: 99645805-99645805 on Build GRCh38
rs754595931 Chr.1: 99646197-99646197 on Build GRCh38
rs986118034 Chr.1: 99646143-99646143 on Build GRCh38
rs949494453 Chr.1: 99646181-99646181 on Build GRCh38
rs959489162 Chr.1: 99646008-99646008 on Build GRCh38
rs926835596 Chr.1: 99646236-99646236 on Build GRCh38
rs986181149 Chr.1: 99645810-99645810 on Build GRCh38
rs759641389 Chr.1: 99646141-99646141 on Build GRCh38
rs887428036 Chr.1: 99646257-99646257 on Build GRCh38
rs763824424 Chr.1: 99646305-99646305 on Build GRCh38
rs758528750 Chr.1: 99646270-99646270 on Build GRCh38
rs566019235 Chr.1: 99646201-99646201 on Build GRCh38
rs914115207 Chr.1: 99646158-99646158 on Build GRCh38
rs140178482 Chr.1: 99645947-99645947 on Build GRCh38
rs575014063 Chr.1: 99646333-99646333 on Build GRCh38
rs988616714 Chr.1: 99646157-99646157 on Build GRCh38
rs971333011 Chr.1: 99646121-99646124 on Build GRCh38
rs116419334 Chr.1: 99645869-99645869 on Build GRCh38
rs371843474 Chr.1: 99645932-99645932 on Build GRCh38
rs774347792 Chr.1: 99646281-99646284 on Build GRCh38
rs1036519146 Chr.1: 99646271-99646271 on Build GRCh38
rs866884201 Chr.1: 99646177-99646177 on Build GRCh38
rs1020676305 Chr.1: 99645928-99645928 on Build GRCh38
rs981418038 Chr.1: 99646121-99646121 on Build GRCh38
rs936846137 Chr.1: 99645825-99645825 on Build GRCh38
rs997427850 Chr.1: 99646117-99646117 on Build GRCh38
rs745749594 Chr.1: 99646289-99646290 on Build GRCh38
rs909369555 Chr.1: 99646170-99646170 on Build GRCh38
rs1000600434 Chr.1: 99646328-99646328 on Build GRCh38
rs760081424 Chr.1: 99646281-99646281 on Build GRCh38
rs897665490 Chr.1: 99646282-99646282 on Build GRCh38
rs1046682513 Chr.1: 99645887-99645887 on Build GRCh38
rs534722346 Chr.1: 99646238-99646238 on Build GRCh38
rs937009980 Chr.1: 99646249-99646249 on Build GRCh38
rs966494286 Chr.1: 99645930-99645930 on Build GRCh38
rs1032192372 Chr.1: 99645981-99645981 on Build GRCh38
rs558678804 Chr.1: 99646159-99646159 on Build GRCh38
rs925356785 Chr.1: 99645812-99645812 on Build GRCh38
rs1007792116 Chr.1: 99645806-99645806 on Build GRCh38
rs906873608 Chr.1: 99645894-99645894 on Build GRCh38
rs1042088027 Chr.1: 99646262-99646262 on Build GRCh38
rs958169405 Chr.1: 99646152-99646152 on Build GRCh38
rs770126653 Chr.1: 99646293-99646293 on Build GRCh38
rs917865554 Chr.1: 99646118-99646119 on Build GRCh38
rs369664376 Chr.1: 99646294-99646294 on Build GRCh38
rs997221426 Chr.1: 99645923-99645923 on Build GRCh38
rs889007070 Chr.1: 99646321-99646321 on Build GRCh38
rs1001169323 Chr.1: 99645896-99645896 on Build GRCh38
rs575832110 Chr.1: 99646214-99646215 on Build GRCh38
rs961846088 Chr.1: 99646110-99646110 on Build GRCh38
rs1039302822 Chr.1: 99646220-99646220 on Build GRCh38
rs1006365910 Chr.1: 99645956-99645956 on Build GRCh38
rs982370503 Chr.1: 99646202-99646202 on Build GRCh38
rs374336429 Chr.1: 99646250-99646250 on Build GRCh38
rs1052590575 Chr.1: 99645831-99645831 on Build GRCh38
rs766595537 Chr.1: 99645877-99645877 on Build GRCh38
rs997150073 Chr.1: 99646254-99646254 on Build GRCh38
rs111874805 Chr.1: 99646078-99646078 on Build GRCh38
rs887512097 Chr.1: 99645950-99645950 on Build GRCh38
rs777088503 Chr.1: 99646276-99646276 on Build GRCh38
rs1014789152 Chr.1: 99645965-99645965 on Build GRCh38
rs1030165650 Chr.1: 99646132-99646132 on Build GRCh38
rs77121907 Chr.1: 99646146-99646146 on Build GRCh38

View Full Product Details