Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63240130-63240689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC7
Entrez Gene ID
79444
Gene Name
baculoviral IAP repeat containing 7
Gene Aliases
KIAP, LIVIN, ML-IAP, MLIAP, RNF50
UniGene
Hs.256126
Gene Chromosome Location
Chr.20: 63235883-63240507 on Build GRCh38
Transcripts
NM_022161.3 NM_139317.2

SNP Detail

SNP ID Chromosome Location
rs1045808487 Chr.20: 63240388-63240388 on Build GRCh38
rs372341579 Chr.20: 63240657-63240657 on Build GRCh38
rs1055918027 Chr.20: 63240158-63240158 on Build GRCh38
rs554219550 Chr.20: 63240267-63240267 on Build GRCh38
rs903244592 Chr.20: 63240415-63240415 on Build GRCh38
rs553544446 Chr.20: 63240351-63240351 on Build GRCh38
rs950549261 Chr.20: 63240544-63240544 on Build GRCh38
rs6062856 Chr.20: 63240645-63240645 on Build GRCh38
rs959750433 Chr.20: 63240359-63240359 on Build GRCh38
rs945917279 Chr.20: 63240180-63240180 on Build GRCh38
rs796735145 Chr.20: 63240527-63240528 on Build GRCh38
rs374854695 Chr.20: 63240255-63240255 on Build GRCh38
rs908509614 Chr.20: 63240589-63240589 on Build GRCh38
rs115140499 Chr.20: 63240385-63240385 on Build GRCh38
rs1032949861 Chr.20: 63240228-63240228 on Build GRCh38
rs1047196242 Chr.20: 63240518-63240518 on Build GRCh38
rs893131259 Chr.20: 63240441-63240441 on Build GRCh38
rs959031704 Chr.20: 63240578-63240578 on Build GRCh38
rs1017497812 Chr.20: 63240465-63240465 on Build GRCh38
rs1032379661 Chr.20: 63240536-63240536 on Build GRCh38
rs941640939 Chr.20: 63240353-63240353 on Build GRCh38
rs1019885656 Chr.20: 63240376-63240376 on Build GRCh38
rs574098893 Chr.20: 63240427-63240427 on Build GRCh38
rs554188193 Chr.20: 63240425-63240425 on Build GRCh38
rs920318590 Chr.20: 63240488-63240489 on Build GRCh38
rs941602688 Chr.20: 63240155-63240155 on Build GRCh38
rs758162537 Chr.20: 63240393-63240393 on Build GRCh38
rs1017486394 Chr.20: 63240259-63240259 on Build GRCh38
rs180774596 Chr.20: 63240170-63240170 on Build GRCh38
rs934024036 Chr.20: 63240501-63240501 on Build GRCh38
rs1039791624 Chr.20: 63240218-63240218 on Build GRCh38
rs978721786 Chr.20: 63240214-63240214 on Build GRCh38
rs6122407 Chr.20: 63240193-63240193 on Build GRCh38
rs758834783 Chr.20: 63240217-63240217 on Build GRCh38
rs571947623 Chr.20: 63240382-63240382 on Build GRCh38
rs372628348 Chr.20: 63240550-63240550 on Build GRCh38
rs916135281 Chr.20: 63240629-63240629 on Build GRCh38
rs576108774 Chr.20: 63240549-63240549 on Build GRCh38
rs1013963573 Chr.20: 63240373-63240373 on Build GRCh38
rs924773431 Chr.20: 63240652-63240652 on Build GRCh38
rs1039025583 Chr.20: 63240644-63240644 on Build GRCh38
rs918979628 Chr.20: 63240297-63240297 on Build GRCh38
rs1007482751 Chr.20: 63240442-63240442 on Build GRCh38
rs1014331631 Chr.20: 63240202-63240202 on Build GRCh38
rs544933029 Chr.20: 63240339-63240339 on Build GRCh38
rs114358311 Chr.20: 63240248-63240248 on Build GRCh38
rs1038628673 Chr.20: 63240290-63240290 on Build GRCh38
rs567753918 Chr.20: 63240215-63240215 on Build GRCh38
rs941394294 Chr.20: 63240635-63240635 on Build GRCh38
rs922719128 Chr.20: 63240223-63240223 on Build GRCh38
rs1003032969 Chr.20: 63240532-63240532 on Build GRCh38
rs984886502 Chr.20: 63240316-63240316 on Build GRCh38
rs979071686 Chr.20: 63240392-63240392 on Build GRCh38
rs964215213 Chr.20: 63240417-63240417 on Build GRCh38
rs905342235 Chr.20: 63240294-63240294 on Build GRCh38
rs991710118 Chr.20: 63240622-63240622 on Build GRCh38
rs903193110 Chr.20: 63240216-63240216 on Build GRCh38
rs892029027 Chr.20: 63240540-63240540 on Build GRCh38
rs1002419651 Chr.20: 63240655-63240655 on Build GRCh38
rs556539590 Chr.20: 63240266-63240266 on Build GRCh38
rs1026127902 Chr.20: 63240533-63240533 on Build GRCh38
rs1002416215 Chr.20: 63240304-63240304 on Build GRCh38
rs981601269 Chr.20: 63240646-63240646 on Build GRCh38
rs560773808 Chr.20: 63240473-63240473 on Build GRCh38
rs6122406 Chr.20: 63240173-63240173 on Build GRCh38

View Full Product Details