Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51177411-51177934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEX3C
Entrez Gene ID
51320
Gene Name
mex-3 RNA binding family member C
Gene Aliases
BM-013, MEX-3C, RKHD2, RNF194
UniGene
Hs.465144
Gene Chromosome Location
Chr.18: 51174550-51197681 on Build GRCh38
Transcripts
NM_016626.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
988914 c.325C>T p.R109C Chr.18: 51177421-51177421 on Build GRCh38
2879650 c.259A>C p.R87R Chr.18: 51177487-51177487 on Build GRCh38
2879640 c.327T>C p.R109R Chr.18: 51177419-51177419 on Build GRCh38
5677733 c.815G>C p.G272A Chr.18: 51177516-51177516 on Build GRCh38
1141462 c.903C>T p.S301S Chr.18: 51177428-51177428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054795085 Chr.18: 51177738-51177738 on Build GRCh38
rs117501522 Chr.18: 51177894-51177894 on Build GRCh38
rs939049330 Chr.18: 51177806-51177806 on Build GRCh38
rs906254104 Chr.18: 51177671-51177671 on Build GRCh38
rs879614815 Chr.18: 51177459-51177459 on Build GRCh38
rs373388741 Chr.18: 51177449-51177449 on Build GRCh38
rs571494470 Chr.18: 51177893-51177893 on Build GRCh38
rs754601042 Chr.18: 51177519-51177519 on Build GRCh38
rs141476539 Chr.18: 51177780-51177780 on Build GRCh38
rs961211229 Chr.18: 51177794-51177794 on Build GRCh38
rs746996546 Chr.18: 51177841-51177845 on Build GRCh38
rs757927594 Chr.18: 51177607-51177607 on Build GRCh38
rs74755782 Chr.18: 51177839-51177839 on Build GRCh38
rs559716751 Chr.18: 51177470-51177470 on Build GRCh38
rs754213465 Chr.18: 51177500-51177500 on Build GRCh38
rs911487827 Chr.18: 51177598-51177598 on Build GRCh38
rs957569903 Chr.18: 51177892-51177892 on Build GRCh38
rs769062501 Chr.18: 51177442-51177442 on Build GRCh38
rs892182279 Chr.18: 51177836-51177836 on Build GRCh38
rs913410105 Chr.18: 51177636-51177636 on Build GRCh38
rs540792408 Chr.18: 51177539-51177539 on Build GRCh38
rs771141916 Chr.18: 51177776-51177776 on Build GRCh38
rs200651484 Chr.18: 51177626-51177626 on Build GRCh38
rs955440459 Chr.18: 51177826-51177826 on Build GRCh38
rs927648975 Chr.18: 51177798-51177798 on Build GRCh38
rs149948055 Chr.18: 51177476-51177476 on Build GRCh38
rs936265360 Chr.18: 51177617-51177617 on Build GRCh38
rs184339910 Chr.18: 51177813-51177813 on Build GRCh38
rs766759832 Chr.18: 51177494-51177494 on Build GRCh38
rs762012261 Chr.18: 51177469-51177469 on Build GRCh38
rs975750691 Chr.18: 51177520-51177520 on Build GRCh38
rs985412635 Chr.18: 51177482-51177482 on Build GRCh38
rs987563927 Chr.18: 51177724-51177724 on Build GRCh38
rs750917572 Chr.18: 51177473-51177473 on Build GRCh38
rs191393069 Chr.18: 51177773-51177773 on Build GRCh38
rs948292889 Chr.18: 51177779-51177779 on Build GRCh38
rs1042179746 Chr.18: 51177660-51177660 on Build GRCh38
rs899698253 Chr.18: 51177861-51177861 on Build GRCh38
rs776655974 Chr.18: 51177777-51177777 on Build GRCh38
rs954910881 Chr.18: 51177650-51177650 on Build GRCh38
rs752163632 Chr.18: 51177559-51177559 on Build GRCh38
rs1043551740 Chr.18: 51177842-51177842 on Build GRCh38
rs1003240583 Chr.18: 51177708-51177708 on Build GRCh38
rs774098853 Chr.18: 51177582-51177585 on Build GRCh38
rs147841372 Chr.18: 51177739-51177739 on Build GRCh38

View Full Product Details