Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140855829-140856383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA10
Entrez Gene ID
56139
Gene Name
protocadherin alpha 10
Gene Aliases
CNR8, CNRN8, CNRS8, CRNR8, PCDH-ALPHA10
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140855883-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140807074-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA5
Entrez Gene ID
56143
Gene Name
protocadherin alpha 5
Gene Aliases
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140821776-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA6
Entrez Gene ID
56142
Gene Name
protocadherin alpha 6
Gene Aliases
CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140827922-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA7
Entrez Gene ID
56141
Gene Name
protocadherin alpha 7
Gene Aliases
CNR4, CNRN4, CNRS4, CRNR4, PCDH-ALPHA7
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140834384-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA8
Entrez Gene ID
56140
Gene Name
protocadherin alpha 8
Gene Aliases
PCDH-ALPHA8
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140841322-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA9
Entrez Gene ID
9752
Gene Name
protocadherin alpha 9
Gene Aliases
PCDH-ALPHA9
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140847772-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5668254 c.124A>C p.T42P Chr.5: 140856172-140856172 on Build GRCh38
4712042 c.137G>A p.R46H Chr.5: 140856185-140856185 on Build GRCh38
125179 c.310C>A p.H104N Chr.5: 140856358-140856358 on Build GRCh38
448806 c.168G>A p.A56A Chr.5: 140856216-140856216 on Build GRCh38
4425680 c.291G>C p.R97R Chr.5: 140856339-140856339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782167466 Chr.5: 140856120-140856120 on Build GRCh38
rs782449893 Chr.5: 140856143-140856143 on Build GRCh38
rs781959235 Chr.5: 140856168-140856168 on Build GRCh38
rs149846721 Chr.5: 140856069-140856069 on Build GRCh38
rs782115498 Chr.5: 140856112-140856112 on Build GRCh38
rs782323132 Chr.5: 140856305-140856305 on Build GRCh38
rs782325790 Chr.5: 140856295-140856295 on Build GRCh38
rs200204071 Chr.5: 140856318-140856318 on Build GRCh38
rs782584293 Chr.5: 140856341-140856341 on Build GRCh38
rs533914587 Chr.5: 140856070-140856070 on Build GRCh38
rs782602500 Chr.5: 140856145-140856145 on Build GRCh38
rs147800828 Chr.5: 140856154-140856154 on Build GRCh38
rs782007386 Chr.5: 140856148-140856148 on Build GRCh38
rs782782390 Chr.5: 140856337-140856337 on Build GRCh38
rs782200483 Chr.5: 140856013-140856013 on Build GRCh38
rs782417138 Chr.5: 140856223-140856223 on Build GRCh38
rs781872854 Chr.5: 140856136-140856136 on Build GRCh38
rs782472888 Chr.5: 140856271-140856271 on Build GRCh38
rs781846770 Chr.5: 140856323-140856323 on Build GRCh38
rs781874204 Chr.5: 140856187-140856187 on Build GRCh38
rs560286204 Chr.5: 140855913-140855913 on Build GRCh38
rs782147679 Chr.5: 140856216-140856216 on Build GRCh38
rs542615615 Chr.5: 140855959-140855959 on Build GRCh38
rs782775861 Chr.5: 140856173-140856173 on Build GRCh38
rs941733432 Chr.5: 140855994-140855994 on Build GRCh38
rs17844339 Chr.5: 140856230-140856230 on Build GRCh38
rs367902357 Chr.5: 140856269-140856269 on Build GRCh38
rs782772973 Chr.5: 140855915-140855915 on Build GRCh38
rs556174610 Chr.5: 140856210-140856210 on Build GRCh38
rs138367057 Chr.5: 140856289-140856289 on Build GRCh38
rs781975666 Chr.5: 140856109-140856109 on Build GRCh38
rs781849353 Chr.5: 140855940-140855940 on Build GRCh38
rs149039484 Chr.5: 140856129-140856129 on Build GRCh38
rs914508327 Chr.5: 140855929-140855929 on Build GRCh38
rs527332783 Chr.5: 140855952-140855952 on Build GRCh38
rs191873949 Chr.5: 140855986-140855986 on Build GRCh38
rs374070531 Chr.5: 140856211-140856211 on Build GRCh38
rs782689818 Chr.5: 140856178-140856178 on Build GRCh38
rs782114957 Chr.5: 140856304-140856304 on Build GRCh38
rs781811609 Chr.5: 140856015-140856015 on Build GRCh38
rs781911942 Chr.5: 140856046-140856046 on Build GRCh38
rs377602570 Chr.5: 140856297-140856297 on Build GRCh38
rs782220766 Chr.5: 140856204-140856204 on Build GRCh38
rs782523397 Chr.5: 140856336-140856336 on Build GRCh38
rs782182532 Chr.5: 140856196-140856196 on Build GRCh38
rs199529084 Chr.5: 140856259-140856259 on Build GRCh38
rs923077189 Chr.5: 140855951-140855951 on Build GRCh38
rs782812320 Chr.5: 140856011-140856011 on Build GRCh38
rs141221498 Chr.5: 140856167-140856167 on Build GRCh38
rs200004763 Chr.5: 140856177-140856177 on Build GRCh38
rs150172685 Chr.5: 140856202-140856202 on Build GRCh38
rs544932528 Chr.5: 140856321-140856321 on Build GRCh38
rs782207999 Chr.5: 140856061-140856061 on Build GRCh38
rs782635462 Chr.5: 140856281-140856281 on Build GRCh38
rs782672623 Chr.5: 140856140-140856140 on Build GRCh38
rs782149074 Chr.5: 140856006-140856006 on Build GRCh38
rs1031755500 Chr.5: 140856096-140856096 on Build GRCh38
rs782332920 Chr.5: 140856198-140856198 on Build GRCh38
rs782568790 Chr.5: 140856315-140856315 on Build GRCh38
rs369328632 Chr.5: 140856073-140856073 on Build GRCh38
rs533343357 Chr.5: 140856093-140856093 on Build GRCh38
rs782335067 Chr.5: 140856344-140856344 on Build GRCh38
rs145079458 Chr.5: 140855999-140855999 on Build GRCh38
rs149352769 Chr.5: 140856227-140856227 on Build GRCh38
rs782183125 Chr.5: 140856100-140856100 on Build GRCh38
rs182267691 Chr.5: 140855885-140855885 on Build GRCh38
rs781798552 Chr.5: 140856082-140856082 on Build GRCh38
rs553967078 Chr.5: 140856258-140856258 on Build GRCh38
rs577890350 Chr.5: 140855877-140855877 on Build GRCh38
rs533990024 Chr.5: 140856155-140856155 on Build GRCh38
rs199611545 Chr.5: 140856347-140856347 on Build GRCh38
rs782376274 Chr.5: 140856001-140856001 on Build GRCh38
rs782180138 Chr.5: 140856292-140856292 on Build GRCh38
rs782244369 Chr.5: 140856203-140856204 on Build GRCh38
rs782290263 Chr.5: 140856141-140856141 on Build GRCh38
rs782761072 Chr.5: 140856246-140856246 on Build GRCh38
rs782001586 Chr.5: 140856004-140856004 on Build GRCh38
rs782071685 Chr.5: 140856162-140856162 on Build GRCh38
rs17844338 Chr.5: 140856229-140856229 on Build GRCh38
rs781924604 Chr.5: 140856160-140856160 on Build GRCh38
rs201064172 Chr.5: 140856130-140856130 on Build GRCh38
rs374986437 Chr.5: 140856040-140856040 on Build GRCh38
rs781995878 Chr.5: 140856215-140856215 on Build GRCh38
rs773891459 Chr.5: 140856191-140856191 on Build GRCh38
rs782159422 Chr.5: 140856176-140856176 on Build GRCh38
rs990164410 Chr.5: 140855924-140855924 on Build GRCh38
rs782020835 Chr.5: 140856119-140856119 on Build GRCh38
rs782636519 Chr.5: 140856184-140856184 on Build GRCh38
rs149114226 Chr.5: 140856042-140856042 on Build GRCh38
rs782092511 Chr.5: 140856319-140856319 on Build GRCh38
rs782444703 Chr.5: 140856052-140856052 on Build GRCh38
rs782497272 Chr.5: 140856356-140856356 on Build GRCh38
rs782661319 Chr.5: 140856364-140856364 on Build GRCh38
rs138092357 Chr.5: 140856175-140856175 on Build GRCh38
rs907541138 Chr.5: 140855991-140855991 on Build GRCh38
rs371048948 Chr.5: 140855973-140855973 on Build GRCh38
rs782062442 Chr.5: 140856245-140856245 on Build GRCh38
rs781950915 Chr.5: 140855998-140855998 on Build GRCh38
rs782708607 Chr.5: 140856123-140856123 on Build GRCh38
rs781799826 Chr.5: 140856126-140856126 on Build GRCh38
rs782595746 Chr.5: 140856286-140856286 on Build GRCh38
rs941778432 Chr.5: 140856193-140856193 on Build GRCh38
rs142037616 Chr.5: 140856147-140856147 on Build GRCh38
rs782445424 Chr.5: 140856260-140856260 on Build GRCh38
rs933070909 Chr.5: 140855960-140855960 on Build GRCh38
rs782387382 Chr.5: 140856310-140856310 on Build GRCh38
rs782797869 Chr.5: 140856116-140856116 on Build GRCh38
rs3806841 Chr.5: 140855953-140855953 on Build GRCh38
rs200441286 Chr.5: 140856169-140856169 on Build GRCh38
rs782811970 Chr.5: 140856044-140856044 on Build GRCh38
rs782347472 Chr.5: 140856108-140856108 on Build GRCh38
rs782543440 Chr.5: 140855990-140855990 on Build GRCh38
rs145849392 Chr.5: 140856078-140856078 on Build GRCh38
rs782582964 Chr.5: 140856261-140856261 on Build GRCh38
rs781903292 Chr.5: 140856338-140856338 on Build GRCh38

View Full Product Details