Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 135318320-135318778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHI1
Entrez Gene ID
54806
Gene Name
Abelson helper integration site 1
Gene Aliases
AHI-1, JBTS3, ORF1, dJ71N10.1
UniGene
Hs.386684
Gene Chromosome Location
Chr.6: 135283972-135497775 on Build GRCh38
Transcripts
NM_001134830.1 NM_001134831.1 NM_017651.4 XM_011535910.2 XM_011535911.2 XM_017010977.1 XM_017010978.1 XM_017010979.1 XM_017010980.1 XM_017010981.1 XM_017010982.1 XM_017010983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3174641 c.3329C>T p.T1110I Chr.6: 135318616-135318616 on Build GRCh38
5061428 c.3364C>T p.R1122* Chr.6: 135318581-135318581 on Build GRCh38
3858476 c.3380G>T p.S1127I Chr.6: 135318565-135318565 on Build GRCh38
4988782 c.3332T>C p.L1111P Chr.6: 135318613-135318613 on Build GRCh38
3620893 c.3350C>T p.P1117L Chr.6: 135318595-135318595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143261524 Chr.6: 135318359-135318359 on Build GRCh38
rs372673953 Chr.6: 135318552-135318552 on Build GRCh38
rs749635663 Chr.6: 135318465-135318466 on Build GRCh38
rs114874016 Chr.6: 135318451-135318451 on Build GRCh38
rs182916933 Chr.6: 135318683-135318683 on Build GRCh38
rs773514922 Chr.6: 135318615-135318615 on Build GRCh38
rs117447608 Chr.6: 135318577-135318577 on Build GRCh38
rs920897302 Chr.6: 135318550-135318550 on Build GRCh38
rs749512217 Chr.6: 135318619-135318619 on Build GRCh38
rs893650468 Chr.6: 135318517-135318517 on Build GRCh38
rs200532937 Chr.6: 135318471-135318471 on Build GRCh38
rs539653309 Chr.6: 135318391-135318391 on Build GRCh38
rs375647379 Chr.6: 135318564-135318564 on Build GRCh38
rs891027780 Chr.6: 135318616-135318616 on Build GRCh38
rs971631961 Chr.6: 135318698-135318698 on Build GRCh38
rs766441368 Chr.6: 135318499-135318499 on Build GRCh38
rs372888581 Chr.6: 135318580-135318580 on Build GRCh38
rs6914831 Chr.6: 135318506-135318506 on Build GRCh38
rs190677416 Chr.6: 135318464-135318464 on Build GRCh38
rs1004117268 Chr.6: 135318636-135318636 on Build GRCh38
rs755957905 Chr.6: 135318531-135318531 on Build GRCh38
rs763598267 Chr.6: 135318536-135318536 on Build GRCh38
rs771191530 Chr.6: 135318621-135318621 on Build GRCh38
rs752391496 Chr.6: 135318529-135318529 on Build GRCh38
rs746002430 Chr.6: 135318658-135318658 on Build GRCh38
rs1027751873 Chr.6: 135318677-135318677 on Build GRCh38
rs773495107 Chr.6: 135318496-135318496 on Build GRCh38
rs201148693 Chr.6: 135318527-135318527 on Build GRCh38
rs931519427 Chr.6: 135318363-135318363 on Build GRCh38
rs562653082 Chr.6: 135318663-135318663 on Build GRCh38
rs751696699 Chr.6: 135318502-135318502 on Build GRCh38
rs28417237 Chr.6: 135318631-135318631 on Build GRCh38
rs771754539 Chr.6: 135318579-135318580 on Build GRCh38
rs908825818 Chr.6: 135318416-135318416 on Build GRCh38
rs775508831 Chr.6: 135318646-135318646 on Build GRCh38
rs1054847144 Chr.6: 135318515-135318515 on Build GRCh38
rs541370297 Chr.6: 135318508-135318508 on Build GRCh38
rs992121221 Chr.6: 135318415-135318415 on Build GRCh38
rs762102067 Chr.6: 135318610-135318610 on Build GRCh38
rs547992509 Chr.6: 135318709-135318709 on Build GRCh38
rs756931330 Chr.6: 135318548-135318548 on Build GRCh38
rs952854603 Chr.6: 135318715-135318715 on Build GRCh38
rs773745153 Chr.6: 135318682-135318682 on Build GRCh38
rs753536518 Chr.6: 135318543-135318543 on Build GRCh38
rs975553680 Chr.6: 135318354-135318354 on Build GRCh38
rs373503518 Chr.6: 135318498-135318498 on Build GRCh38
rs1036820704 Chr.6: 135318640-135318640 on Build GRCh38
rs908780616 Chr.6: 135318731-135318731 on Build GRCh38
rs770197175 Chr.6: 135318613-135318613 on Build GRCh38
rs760322271 Chr.6: 135318655-135318655 on Build GRCh38
rs984718007 Chr.6: 135318724-135318724 on Build GRCh38
rs780408017 Chr.6: 135318490-135318490 on Build GRCh38
rs34299308 Chr.6: 135318727-135318727 on Build GRCh38
rs1012902737 Chr.6: 135318689-135318690 on Build GRCh38
rs370808684 Chr.6: 135318623-135318623 on Build GRCh38
rs199708272 Chr.6: 135318603-135318603 on Build GRCh38
rs759639934 Chr.6: 135318625-135318625 on Build GRCh38
rs745419871 Chr.6: 135318557-135318557 on Build GRCh38
rs767390315 Chr.6: 135318642-135318642 on Build GRCh38
rs565249188 Chr.6: 135318441-135318441 on Build GRCh38
rs1020870112 Chr.6: 135318572-135318572 on Build GRCh38

View Full Product Details