Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95771429-95771905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM76B
Entrez Gene ID
143684
Gene Name
family with sequence similarity 76 member B
Gene Aliases
-
UniGene
Hs.288304
Gene Chromosome Location
Chr.11: 95768942-95790396 on Build GRCh38
Transcripts
NM_144664.4 XM_005273775.4 XM_005273776.4 XM_011542610.2 XM_011542611.2 XM_011542612.1 XM_011542613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6251816 c.1010C>A p.T337K Chr.11: 95771571-95771571 on Build GRCh38
6588135 c.941G>C p.R314T Chr.11: 95771640-95771640 on Build GRCh38
7159709 c.951C>G p.L317L Chr.11: 95771630-95771630 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1050352552 Chr.11: 95771822-95771822 on Build GRCh38
rs982534774 Chr.11: 95771508-95771508 on Build GRCh38
rs934762974 Chr.11: 95771699-95771699 on Build GRCh38
rs754187254 Chr.11: 95771514-95771514 on Build GRCh38
rs757486610 Chr.11: 95771621-95771621 on Build GRCh38
rs373243898 Chr.11: 95771573-95771573 on Build GRCh38
rs760838810 Chr.11: 95771602-95771602 on Build GRCh38
rs769398471 Chr.11: 95771542-95771542 on Build GRCh38
rs950065031 Chr.11: 95771750-95771750 on Build GRCh38
rs991711687 Chr.11: 95771714-95771714 on Build GRCh38
rs921187052 Chr.11: 95771605-95771605 on Build GRCh38
rs747614674 Chr.11: 95771655-95771655 on Build GRCh38
rs1018638565 Chr.11: 95771845-95771845 on Build GRCh38
rs757476270 Chr.11: 95771493-95771493 on Build GRCh38
rs771349510 Chr.11: 95771564-95771566 on Build GRCh38
rs752781261 Chr.11: 95771597-95771597 on Build GRCh38
rs537242062 Chr.11: 95771718-95771718 on Build GRCh38
rs541500180 Chr.11: 95771784-95771784 on Build GRCh38
rs888756350 Chr.11: 95771827-95771827 on Build GRCh38
rs757652925 Chr.11: 95771515-95771515 on Build GRCh38
rs375781739 Chr.11: 95771566-95771566 on Build GRCh38
rs986695926 Chr.11: 95771859-95771859 on Build GRCh38
rs780659284 Chr.11: 95771712-95771712 on Build GRCh38
rs994718013 Chr.11: 95771818-95771818 on Build GRCh38
rs779212467 Chr.11: 95771521-95771521 on Build GRCh38
rs200981048 Chr.11: 95771523-95771523 on Build GRCh38
rs1026411909 Chr.11: 95771487-95771487 on Build GRCh38
rs764352249 Chr.11: 95771613-95771613 on Build GRCh38
rs769314392 Chr.11: 95771667-95771667 on Build GRCh38
rs555495615 Chr.11: 95771671-95771671 on Build GRCh38
rs899074102 Chr.11: 95771529-95771529 on Build GRCh38
rs775064699 Chr.11: 95771637-95771638 on Build GRCh38
rs906335975 Chr.11: 95771503-95771503 on Build GRCh38
rs200339806 Chr.11: 95771664-95771664 on Build GRCh38
rs747681751 Chr.11: 95771538-95771538 on Build GRCh38
rs376848080 Chr.11: 95771643-95771643 on Build GRCh38
rs558150856 Chr.11: 95771632-95771632 on Build GRCh38
rs998652358 Chr.11: 95771867-95771867 on Build GRCh38
rs759491549 Chr.11: 95771575-95771575 on Build GRCh38
rs578060310 Chr.11: 95771806-95771806 on Build GRCh38
rs770552232 Chr.11: 95771674-95771674 on Build GRCh38
rs926183345 Chr.11: 95771706-95771706 on Build GRCh38
rs772858364 Chr.11: 95771557-95771557 on Build GRCh38
rs754097720 Chr.11: 95771617-95771617 on Build GRCh38
rs772098419 Chr.11: 95771693-95771693 on Build GRCh38
rs1012878554 Chr.11: 95771727-95771727 on Build GRCh38
rs1008555653 Chr.11: 95771835-95771835 on Build GRCh38
rs1033563162 Chr.11: 95771868-95771868 on Build GRCh38
rs780571127 Chr.11: 95771646-95771646 on Build GRCh38
rs754084051 Chr.11: 95771461-95771461 on Build GRCh38
rs775593628 Chr.11: 95771697-95771697 on Build GRCh38
rs1022881337 Chr.11: 95771746-95771746 on Build GRCh38
rs767580908 Chr.11: 95771595-95771595 on Build GRCh38
rs116380897 Chr.11: 95771757-95771757 on Build GRCh38
rs575443253 Chr.11: 95771686-95771686 on Build GRCh38
rs746381454 Chr.11: 95771518-95771518 on Build GRCh38
rs759257231 Chr.11: 95771687-95771687 on Build GRCh38
rs967530639 Chr.11: 95771865-95771865 on Build GRCh38
rs749750689 Chr.11: 95771515-95771516 on Build GRCh38
rs1036597316 Chr.11: 95771790-95771790 on Build GRCh38
rs117505137 Chr.11: 95771762-95771762 on Build GRCh38
rs955301306 Chr.11: 95771838-95771838 on Build GRCh38
rs779304798 Chr.11: 95771623-95771623 on Build GRCh38
rs1026465713 Chr.11: 95771775-95771775 on Build GRCh38
rs568874939 Chr.11: 95771507-95771508 on Build GRCh38
rs369049259 Chr.11: 95771568-95771568 on Build GRCh38
rs950911258 Chr.11: 95771777-95771777 on Build GRCh38
rs760164832 Chr.11: 95771669-95771669 on Build GRCh38
rs779201665 Chr.11: 95771516-95771516 on Build GRCh38
rs559815418 Chr.11: 95771800-95771800 on Build GRCh38
rs576616886 Chr.11: 95771716-95771716 on Build GRCh38
rs565308842 Chr.11: 95771483-95771483 on Build GRCh38

View Full Product Details