Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119132365-119132964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINFP
Entrez Gene ID
25988
Gene Name
histone H4 transcription factor
Gene Aliases
HiNF-P, MIZF, ZNF743
UniGene
Hs.504091
Gene Chromosome Location
Chr.11: 119121523-119135055 on Build GRCh38
Transcripts
NM_001243259.1 NM_015517.4 NM_198971.2 XM_011542744.2 XM_011542745.2 XM_011542746.1 XM_017017499.1 XM_017017500.1 XM_017017501.1 XM_017017502.1 XM_017017503.1 XM_017017504.1 XM_017017505.1 XM_017017506.1 XM_017017507.1 XM_017017508.1 XM_017017509.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6328660 c.755-5C>T p.? Chr.11: 119132656-119132656 on Build GRCh38
4018475 c.955T>C p.C319R Chr.11: 119132943-119132943 on Build GRCh38
4018474 c.725G>A p.R242Q Chr.11: 119132544-119132544 on Build GRCh38
6854960 c.870C>T p.D290D Chr.11: 119132776-119132776 on Build GRCh38
5520988 c.746G>A p.R249H Chr.11: 119132565-119132565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139354951 Chr.11: 119132760-119132760 on Build GRCh38
rs753418944 Chr.11: 119132628-119132628 on Build GRCh38
rs374266980 Chr.11: 119132546-119132546 on Build GRCh38
rs374160093 Chr.11: 119132831-119132831 on Build GRCh38
rs562905964 Chr.11: 119132717-119132717 on Build GRCh38
rs760606813 Chr.11: 119132735-119132735 on Build GRCh38
rs777150630 Chr.11: 119132700-119132700 on Build GRCh38
rs765009336 Chr.11: 119132878-119132878 on Build GRCh38
rs960302696 Chr.11: 119132395-119132395 on Build GRCh38
rs113330716 Chr.11: 119132543-119132543 on Build GRCh38
rs146974311 Chr.11: 119132730-119132730 on Build GRCh38
rs773853212 Chr.11: 119132565-119132565 on Build GRCh38
rs780798872 Chr.11: 119132552-119132552 on Build GRCh38
rs369983660 Chr.11: 119132462-119132462 on Build GRCh38
rs770473895 Chr.11: 119132496-119132496 on Build GRCh38
rs765351974 Chr.11: 119132610-119132610 on Build GRCh38
rs920086089 Chr.11: 119132801-119132801 on Build GRCh38
rs1022984774 Chr.11: 119132434-119132434 on Build GRCh38
rs1014251124 Chr.11: 119132407-119132407 on Build GRCh38
rs368077854 Chr.11: 119132915-119132915 on Build GRCh38
rs755900940 Chr.11: 119132553-119132553 on Build GRCh38
rs754959400 Chr.11: 119132550-119132550 on Build GRCh38
rs747537186 Chr.11: 119132670-119132670 on Build GRCh38
rs746300693 Chr.11: 119132572-119132572 on Build GRCh38
rs775948465 Chr.11: 119132516-119132516 on Build GRCh38
rs775904186 Chr.11: 119132587-119132587 on Build GRCh38
rs754573616 Chr.11: 119132909-119132909 on Build GRCh38
rs771079583 Chr.11: 119132933-119132933 on Build GRCh38
rs921607956 Chr.11: 119132562-119132562 on Build GRCh38
rs773470395 Chr.11: 119132822-119132822 on Build GRCh38
rs750083718 Chr.11: 119132750-119132750 on Build GRCh38
rs201442297 Chr.11: 119132614-119132614 on Build GRCh38
rs751477070 Chr.11: 119132549-119132549 on Build GRCh38
rs753653489 Chr.11: 119132767-119132767 on Build GRCh38
rs761081653 Chr.11: 119132595-119132595 on Build GRCh38
rs774305870 Chr.11: 119132833-119132833 on Build GRCh38
rs370577473 Chr.11: 119132503-119132503 on Build GRCh38
rs761583730 Chr.11: 119132877-119132877 on Build GRCh38
rs202020241 Chr.11: 119132701-119132701 on Build GRCh38
rs776989356 Chr.11: 119132605-119132605 on Build GRCh38
rs1028149356 Chr.11: 119132834-119132834 on Build GRCh38
rs375828893 Chr.11: 119132835-119132835 on Build GRCh38
rs756492224 Chr.11: 119132924-119132924 on Build GRCh38
rs200102995 Chr.11: 119132582-119132582 on Build GRCh38
rs577404339 Chr.11: 119132436-119132436 on Build GRCh38
rs749878705 Chr.11: 119132471-119132474 on Build GRCh38
rs141315890 Chr.11: 119132702-119132702 on Build GRCh38
rs755886037 Chr.11: 119132622-119132622 on Build GRCh38
rs148039711 Chr.11: 119132882-119132882 on Build GRCh38
rs375947209 Chr.11: 119132487-119132487 on Build GRCh38
rs376530533 Chr.11: 119132501-119132501 on Build GRCh38
rs776601278 Chr.11: 119132866-119132866 on Build GRCh38
rs878930321 Chr.11: 119132485-119132485 on Build GRCh38
rs575006452 Chr.11: 119132736-119132736 on Build GRCh38
rs200770412 Chr.11: 119132775-119132775 on Build GRCh38
rs370250147 Chr.11: 119132814-119132814 on Build GRCh38
rs767978551 Chr.11: 119132445-119132445 on Build GRCh38
rs771535254 Chr.11: 119132648-119132648 on Build GRCh38
rs774768553 Chr.11: 119132464-119132464 on Build GRCh38
rs760555535 Chr.11: 119132854-119132854 on Build GRCh38
rs745355710 Chr.11: 119132646-119132646 on Build GRCh38
rs1022932442 Chr.11: 119132796-119132796 on Build GRCh38
rs765889765 Chr.11: 119132798-119132798 on Build GRCh38
rs764384793 Chr.11: 119132537-119132537 on Build GRCh38
rs763989664 Chr.11: 119132856-119132856 on Build GRCh38
rs752855227 Chr.11: 119132446-119132446 on Build GRCh38
rs200152500 Chr.11: 119132790-119132790 on Build GRCh38
rs778004690 Chr.11: 119132927-119132927 on Build GRCh38
rs747493397 Chr.11: 119132799-119132799 on Build GRCh38
rs770419129 Chr.11: 119132564-119132564 on Build GRCh38
rs751148237 Chr.11: 119132908-119132908 on Build GRCh38
rs773345381 Chr.11: 119132706-119132706 on Build GRCh38
rs778356419 Chr.11: 119132644-119132644 on Build GRCh38
rs769076814 Chr.11: 119132679-119132679 on Build GRCh38
rs772668341 Chr.11: 119132491-119132491 on Build GRCh38
rs754726645 Chr.11: 119132467-119132467 on Build GRCh38
rs920405866 Chr.11: 119132481-119132481 on Build GRCh38
rs970053879 Chr.11: 119132802-119132802 on Build GRCh38
rs765481086 Chr.11: 119132544-119132544 on Build GRCh38
rs544787874 Chr.11: 119132508-119132508 on Build GRCh38
rs183673787 Chr.11: 119132616-119132616 on Build GRCh38
rs762131169 Chr.11: 119132608-119132608 on Build GRCh38
rs769507372 Chr.11: 119132488-119132488 on Build GRCh38
rs781653457 Chr.11: 119132800-119132800 on Build GRCh38
rs777382562 Chr.11: 119132557-119132557 on Build GRCh38
rs757926763 Chr.11: 119132795-119132795 on Build GRCh38
rs373000641 Chr.11: 119132493-119132493 on Build GRCh38
rs748719286 Chr.11: 119132921-119132921 on Build GRCh38
rs757455572 Chr.11: 119132463-119132463 on Build GRCh38
rs662069 Chr.11: 119132612-119132612 on Build GRCh38
rs996607907 Chr.11: 119132545-119132545 on Build GRCh38
rs544241823 Chr.11: 119132635-119132635 on Build GRCh38
rs757857020 Chr.11: 119132875-119132876 on Build GRCh38
rs756361195 Chr.11: 119132449-119132449 on Build GRCh38
rs35118112 Chr.11: 119132524-119132524 on Build GRCh38
rs575227348 Chr.11: 119132597-119132597 on Build GRCh38
rs200108358 Chr.11: 119132711-119132711 on Build GRCh38
rs754166978 Chr.11: 119132538-119132538 on Build GRCh38
rs373409976 Chr.11: 119132476-119132476 on Build GRCh38
rs780601233 Chr.11: 119132652-119132652 on Build GRCh38
rs752671821 Chr.11: 119132734-119132734 on Build GRCh38
rs377637914 Chr.11: 119132563-119132563 on Build GRCh38
rs911778686 Chr.11: 119132529-119132529 on Build GRCh38
rs749467609 Chr.11: 119132931-119132931 on Build GRCh38
rs761232973 Chr.11: 119132586-119132586 on Build GRCh38
rs777667179 Chr.11: 119132455-119132455 on Build GRCh38
rs950851384 Chr.11: 119132840-119132840 on Build GRCh38
rs756913692 Chr.11: 119132643-119132643 on Build GRCh38
rs770330530 Chr.11: 119132820-119132820 on Build GRCh38
rs558967965 Chr.11: 119132396-119132396 on Build GRCh38
rs751371937 Chr.11: 119132935-119132935 on Build GRCh38
rs747000509 Chr.11: 119132466-119132466 on Build GRCh38
rs1053319844 Chr.11: 119132784-119132784 on Build GRCh38
rs777607392 Chr.11: 119132623-119132623 on Build GRCh38
rs758004053 Chr.11: 119132895-119132895 on Build GRCh38
rs766427405 Chr.11: 119132718-119132718 on Build GRCh38
rs778608840 Chr.11: 119132786-119132786 on Build GRCh38

View Full Product Details