Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21001179-21001752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A2
Entrez Gene ID
29986
Gene Name
solute carrier family 39 member 2
Gene Aliases
6A1, ETI-1, ZIP-2, ZIP2
UniGene
Hs.175783
Gene Chromosome Location
Chr.14: 20999255-21001875 on Build GRCh38
Transcripts
NM_001256588.1 NM_014579.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
72695 c.604C>T p.H202Y Chr.14: 21001253-21001253 on Build GRCh38
4987488 c.893G>T p.G298V Chr.14: 21001542-21001542 on Build GRCh38
1677857 c.601G>T p.A201S Chr.14: 21001250-21001250 on Build GRCh38
7276008 c.671C>T p.A224V Chr.14: 21001320-21001320 on Build GRCh38
4652458 c.777T>C p.A259A Chr.14: 21001426-21001426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762289107 Chr.14: 21001520-21001520 on Build GRCh38
rs759621913 Chr.14: 21001314-21001314 on Build GRCh38
rs748980161 Chr.14: 21001394-21001394 on Build GRCh38
rs866511844 Chr.14: 21001363-21001363 on Build GRCh38
rs766452966 Chr.14: 21001250-21001250 on Build GRCh38
rs755574284 Chr.14: 21001555-21001555 on Build GRCh38
rs776178193 Chr.14: 21001414-21001414 on Build GRCh38
rs199812093 Chr.14: 21001490-21001490 on Build GRCh38
rs201004359 Chr.14: 21001487-21001487 on Build GRCh38
rs765447780 Chr.14: 21001213-21001213 on Build GRCh38
rs778879063 Chr.14: 21001567-21001567 on Build GRCh38
rs751193511 Chr.14: 21001463-21001463 on Build GRCh38
rs774710308 Chr.14: 21001401-21001401 on Build GRCh38
rs750064810 Chr.14: 21001277-21001277 on Build GRCh38
rs773526524 Chr.14: 21001531-21001531 on Build GRCh38
rs1010190035 Chr.14: 21001704-21001704 on Build GRCh38
rs957308824 Chr.14: 21001679-21001679 on Build GRCh38
rs769580091 Chr.14: 21001373-21001373 on Build GRCh38
rs185949718 Chr.14: 21001311-21001311 on Build GRCh38
rs138343150 Chr.14: 21001558-21001558 on Build GRCh38
rs147806155 Chr.14: 21001209-21001209 on Build GRCh38
rs779619376 Chr.14: 21001359-21001359 on Build GRCh38
rs539767630 Chr.14: 21001499-21001499 on Build GRCh38
rs537502967 Chr.14: 21001395-21001395 on Build GRCh38
rs200748795 Chr.14: 21001357-21001357 on Build GRCh38
rs990941999 Chr.14: 21001619-21001619 on Build GRCh38
rs757883727 Chr.14: 21001281-21001281 on Build GRCh38
rs1051624119 Chr.14: 21001721-21001721 on Build GRCh38
rs764455481 Chr.14: 21001427-21001427 on Build GRCh38
rs756091366 Chr.14: 21001476-21001476 on Build GRCh38
rs772377404 Chr.14: 21001503-21001503 on Build GRCh38
rs576512392 Chr.14: 21001607-21001607 on Build GRCh38
rs746999749 Chr.14: 21001504-21001504 on Build GRCh38
rs370867815 Chr.14: 21001298-21001298 on Build GRCh38
rs963873760 Chr.14: 21001225-21001225 on Build GRCh38
rs769611697 Chr.14: 21001297-21001297 on Build GRCh38
rs915272077 Chr.14: 21001623-21001623 on Build GRCh38
rs754525772 Chr.14: 21001467-21001467 on Build GRCh38
rs768588995 Chr.14: 21001292-21001292 on Build GRCh38
rs145679995 Chr.14: 21001522-21001522 on Build GRCh38
rs1007298312 Chr.14: 21001452-21001452 on Build GRCh38
rs375310383 Chr.14: 21001538-21001538 on Build GRCh38
rs201278435 Chr.14: 21001313-21001313 on Build GRCh38
rs758704040 Chr.14: 21001593-21001593 on Build GRCh38
rs761273431 Chr.14: 21001424-21001424 on Build GRCh38
rs374140607 Chr.14: 21001286-21001286 on Build GRCh38
rs776994944 Chr.14: 21001512-21001512 on Build GRCh38
rs867731844 Chr.14: 21001269-21001269 on Build GRCh38
rs752101178 Chr.14: 21001470-21001470 on Build GRCh38
rs924451673 Chr.14: 21001645-21001645 on Build GRCh38
rs756494342 Chr.14: 21001268-21001268 on Build GRCh38
rs545174532 Chr.14: 21001353-21001353 on Build GRCh38
rs562183286 Chr.14: 21001665-21001665 on Build GRCh38
rs78369198 Chr.14: 21001446-21001446 on Build GRCh38
rs145048254 Chr.14: 21001226-21001226 on Build GRCh38
rs766082856 Chr.14: 21001459-21001459 on Build GRCh38
rs982078195 Chr.14: 21001285-21001285 on Build GRCh38
rs1020276664 Chr.14: 21001710-21001710 on Build GRCh38
rs533770961 Chr.14: 21001468-21001468 on Build GRCh38
rs2234638 Chr.14: 21001210-21001210 on Build GRCh38
rs1017308491 Chr.14: 21001464-21001464 on Build GRCh38
rs778487362 Chr.14: 21001274-21001274 on Build GRCh38
rs762993649 Chr.14: 21001248-21001248 on Build GRCh38
rs760141922 Chr.14: 21001251-21001251 on Build GRCh38
rs777646887 Chr.14: 21001389-21001389 on Build GRCh38
rs745607282 Chr.14: 21001573-21001573 on Build GRCh38
rs1026579305 Chr.14: 21001489-21001489 on Build GRCh38
rs764485792 Chr.14: 21001334-21001334 on Build GRCh38
rs763268658 Chr.14: 21001303-21001303 on Build GRCh38
rs371660387 Chr.14: 21001203-21001203 on Build GRCh38
rs934608616 Chr.14: 21001689-21001689 on Build GRCh38
rs753240970 Chr.14: 21001321-21001321 on Build GRCh38
rs772255805 Chr.14: 21001410-21001410 on Build GRCh38
rs140359641 Chr.14: 21001488-21001488 on Build GRCh38
rs900353404 Chr.14: 21001615-21001615 on Build GRCh38
rs886179576 Chr.14: 21001578-21001578 on Build GRCh38
rs115791592 Chr.14: 21001437-21001437 on Build GRCh38
rs1009073609 Chr.14: 21001582-21001582 on Build GRCh38
rs760991848 Chr.14: 21001330-21001330 on Build GRCh38
rs200324289 Chr.14: 21001435-21001435 on Build GRCh38
rs554255537 Chr.14: 21001409-21001409 on Build GRCh38
rs369197232 Chr.14: 21001614-21001614 on Build GRCh38
rs998551208 Chr.14: 21001397-21001397 on Build GRCh38
rs767099737 Chr.14: 21001542-21001542 on Build GRCh38
rs947248010 Chr.14: 21001629-21001629 on Build GRCh38
rs769276266 Chr.14: 21001505-21001505 on Build GRCh38
rs780070956 Chr.14: 21001601-21001601 on Build GRCh38
rs751140614 Chr.14: 21001361-21001361 on Build GRCh38
rs149113931 Chr.14: 21001317-21001317 on Build GRCh38
rs777356846 Chr.14: 21001547-21001555 on Build GRCh38
rs572528359 Chr.14: 21001682-21001682 on Build GRCh38
rs754474127 Chr.14: 21001364-21001364 on Build GRCh38
rs759745730 Chr.14: 21001406-21001406 on Build GRCh38
rs781159174 Chr.14: 21001369-21001369 on Build GRCh38
rs2234639 Chr.14: 21001370-21001370 on Build GRCh38
rs768079811 Chr.14: 21001255-21001255 on Build GRCh38
rs775631460 Chr.14: 21001206-21001206 on Build GRCh38
rs753116315 Chr.14: 21001261-21001261 on Build GRCh38
rs183186857 Chr.14: 21001686-21001686 on Build GRCh38
rs2234640 Chr.14: 21001638-21001638 on Build GRCh38
rs527893268 Chr.14: 21001296-21001296 on Build GRCh38
rs145056031 Chr.14: 21001287-21001287 on Build GRCh38
rs777063466 Chr.14: 21001434-21001434 on Build GRCh38
rs757157290 Chr.14: 21001563-21001563 on Build GRCh38

View Full Product Details