Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50382575-50383132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC59
Entrez Gene ID
55379
Gene Name
leucine rich repeat containing 59
Gene Aliases
PRO1855, p34
UniGene
Hs.370927
Gene Chromosome Location
Chr.17: 50381233-50397553 on Build GRCh38
Transcripts
NM_018509.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6413889 c.890T>C p.L297P Chr.17: 50383022-50383022 on Build GRCh38
6703998 c.874C>T p.R292C Chr.17: 50383038-50383038 on Build GRCh38
6419842 c.898G>T p.V300F Chr.17: 50383014-50383014 on Build GRCh38
6703999 c.879C>T p.R293R Chr.17: 50383033-50383033 on Build GRCh38
4898755 c.873A>T p.L291L Chr.17: 50383039-50383039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201185497 Chr.17: 50382998-50382998 on Build GRCh38
rs1014767295 Chr.17: 50383078-50383078 on Build GRCh38
rs1053533217 Chr.17: 50382993-50382993 on Build GRCh38
rs149087944 Chr.17: 50383051-50383051 on Build GRCh38
rs544291551 Chr.17: 50383072-50383072 on Build GRCh38
rs772143736 Chr.17: 50383037-50383037 on Build GRCh38
rs1052138250 Chr.17: 50382878-50382878 on Build GRCh38
rs919958128 Chr.17: 50382798-50382798 on Build GRCh38
rs753495143 Chr.17: 50383109-50383109 on Build GRCh38
rs745674948 Chr.17: 50383113-50383113 on Build GRCh38
rs190245654 Chr.17: 50382759-50382759 on Build GRCh38
rs537463024 Chr.17: 50382844-50382844 on Build GRCh38
rs137985988 Chr.17: 50383038-50383038 on Build GRCh38
rs972801767 Chr.17: 50382795-50382795 on Build GRCh38
rs778788884 Chr.17: 50383112-50383112 on Build GRCh38
rs187926967 Chr.17: 50382849-50382849 on Build GRCh38
rs536412929 Chr.17: 50383035-50383035 on Build GRCh38
rs1019057046 Chr.17: 50383017-50383017 on Build GRCh38
rs761661372 Chr.17: 50383005-50383006 on Build GRCh38
rs199717142 Chr.17: 50383083-50383083 on Build GRCh38
rs182564736 Chr.17: 50382782-50382782 on Build GRCh38
rs568752344 Chr.17: 50382996-50382996 on Build GRCh38
rs757116582 Chr.17: 50383111-50383111 on Build GRCh38
rs780746800 Chr.17: 50382991-50382991 on Build GRCh38
rs377582963 Chr.17: 50382641-50382641 on Build GRCh38
rs146642281 Chr.17: 50383034-50383034 on Build GRCh38
rs373546654 Chr.17: 50382607-50382607 on Build GRCh38
rs764232630 Chr.17: 50382938-50382938 on Build GRCh38
rs770681229 Chr.17: 50383030-50383030 on Build GRCh38
rs777976263 Chr.17: 50382598-50382598 on Build GRCh38
rs930756638 Chr.17: 50382768-50382768 on Build GRCh38
rs755621027 Chr.17: 50383090-50383090 on Build GRCh38
rs770529197 Chr.17: 50382773-50382773 on Build GRCh38
rs143159492 Chr.17: 50383052-50383052 on Build GRCh38
rs576240945 Chr.17: 50383065-50383065 on Build GRCh38
rs571373096 Chr.17: 50382926-50382926 on Build GRCh38
rs750975889 Chr.17: 50382980-50382980 on Build GRCh38
rs533213855 Chr.17: 50382693-50382693 on Build GRCh38
rs544566628 Chr.17: 50382639-50382639 on Build GRCh38
rs939233341 Chr.17: 50382873-50382873 on Build GRCh38
rs947892218 Chr.17: 50382790-50382790 on Build GRCh38
rs961766319 Chr.17: 50382781-50382781 on Build GRCh38
rs764801621 Chr.17: 50383069-50383070 on Build GRCh38
rs1022003808 Chr.17: 50382927-50382927 on Build GRCh38
rs549398671 Chr.17: 50382776-50382776 on Build GRCh38
rs776755349 Chr.17: 50383055-50383055 on Build GRCh38
rs893471217 Chr.17: 50382680-50382680 on Build GRCh38
rs115294777 Chr.17: 50382627-50382627 on Build GRCh38
rs201332762 Chr.17: 50383002-50383002 on Build GRCh38
rs1011079030 Chr.17: 50382913-50382913 on Build GRCh38
rs762215142 Chr.17: 50383061-50383061 on Build GRCh38
rs145371594 Chr.17: 50383003-50383003 on Build GRCh38
rs1010650655 Chr.17: 50382638-50382638 on Build GRCh38
rs954550168 Chr.17: 50382709-50382709 on Build GRCh38
rs909709068 Chr.17: 50382780-50382780 on Build GRCh38
rs776276398 Chr.17: 50382969-50382974 on Build GRCh38
rs985299470 Chr.17: 50383095-50383095 on Build GRCh38
rs370133634 Chr.17: 50382668-50382668 on Build GRCh38
rs766895386 Chr.17: 50383081-50383081 on Build GRCh38
rs1045018102 Chr.17: 50382843-50382843 on Build GRCh38
rs999251033 Chr.17: 50382666-50382666 on Build GRCh38
rs202162924 Chr.17: 50382979-50382979 on Build GRCh38
rs564427157 Chr.17: 50382667-50382667 on Build GRCh38
rs996531645 Chr.17: 50382681-50382681 on Build GRCh38
rs919258469 Chr.17: 50382766-50382766 on Build GRCh38
rs777461127 Chr.17: 50383092-50383092 on Build GRCh38
rs916851282 Chr.17: 50382868-50382868 on Build GRCh38
rs754469903 Chr.17: 50382986-50382986 on Build GRCh38
rs754178248 Chr.17: 50382957-50382957 on Build GRCh38
rs912122257 Chr.17: 50382733-50382733 on Build GRCh38
rs11548984 Chr.17: 50382636-50382636 on Build GRCh38
rs763383397 Chr.17: 50383074-50383074 on Build GRCh38
rs116114267 Chr.17: 50382774-50382774 on Build GRCh38
rs1022917925 Chr.17: 50382656-50382656 on Build GRCh38
rs749080506 Chr.17: 50383009-50383009 on Build GRCh38
rs1046528769 Chr.17: 50382628-50382628 on Build GRCh38
rs562141195 Chr.17: 50382626-50382626 on Build GRCh38
rs966030097 Chr.17: 50382734-50382734 on Build GRCh38
rs892121372 Chr.17: 50382910-50382910 on Build GRCh38

View Full Product Details