Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49558754-49559316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOSIP
Entrez Gene ID
51070
Gene Name
nitric oxide synthase interacting protein
Gene Aliases
CGI-25
UniGene
Hs.7236
Gene Chromosome Location
Chr.19: 49555468-49580599 on Build GRCh38
Transcripts
NM_001270960.1 NM_015953.4 XM_005258964.4 XM_006723235.3 XM_011527015.2 XM_011527017.2 XM_017026851.1 XM_017026852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
999330 c.247C>T p.R83W Chr.19: 49558908-49558908 on Build GRCh38
6243058 c.209T>C p.I70T Chr.19: 49558946-49558946 on Build GRCh38
6727989 c.200delG p.R67fs*19 Chr.19: 49558955-49558955 on Build GRCh38
2752751 c.199C>T p.R67C Chr.19: 49558956-49558956 on Build GRCh38
1395265 c.235_237delAAG p.K79delK Chr.19: 49558918-49558920 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748280866 Chr.19: 49559012-49559012 on Build GRCh38
rs983901611 Chr.19: 49559178-49559178 on Build GRCh38
rs761989429 Chr.19: 49559000-49559003 on Build GRCh38
rs1017618660 Chr.19: 49559076-49559076 on Build GRCh38
rs773691874 Chr.19: 49558878-49558878 on Build GRCh38
rs976126100 Chr.19: 49558830-49558830 on Build GRCh38
rs763988310 Chr.19: 49558915-49558917 on Build GRCh38
rs770349642 Chr.19: 49558875-49558875 on Build GRCh38
rs777545441 Chr.19: 49559026-49559026 on Build GRCh38
rs901334848 Chr.19: 49559063-49559063 on Build GRCh38
rs79151664 Chr.19: 49559023-49559023 on Build GRCh38
rs140289937 Chr.19: 49559074-49559074 on Build GRCh38
rs550600095 Chr.19: 49559210-49559210 on Build GRCh38
rs771296306 Chr.19: 49559022-49559022 on Build GRCh38
rs778790721 Chr.19: 49558996-49558996 on Build GRCh38
rs947515519 Chr.19: 49559273-49559273 on Build GRCh38
rs147601481 Chr.19: 49558868-49558868 on Build GRCh38
rs766703222 Chr.19: 49558907-49558907 on Build GRCh38
rs759763761 Chr.19: 49558949-49558949 on Build GRCh38
rs938456550 Chr.19: 49558955-49558955 on Build GRCh38
rs763728144 Chr.19: 49559028-49559028 on Build GRCh38
rs59279440 Chr.19: 49559036-49559036 on Build GRCh38
rs571602809 Chr.19: 49559133-49559133 on Build GRCh38
rs769838519 Chr.19: 49559015-49559015 on Build GRCh38
rs983813358 Chr.19: 49558842-49558842 on Build GRCh38
rs4426453 Chr.19: 49559223-49559223 on Build GRCh38
rs749829794 Chr.19: 49559019-49559019 on Build GRCh38
rs1018530654 Chr.19: 49559061-49559061 on Build GRCh38
rs764184868 Chr.19: 49558956-49558956 on Build GRCh38
rs767747125 Chr.19: 49558999-49559000 on Build GRCh38
rs750571557 Chr.19: 49559018-49559030 on Build GRCh38
rs923289265 Chr.19: 49558836-49558836 on Build GRCh38
rs111299973 Chr.19: 49558867-49558867 on Build GRCh38
rs747287256 Chr.19: 49558858-49558858 on Build GRCh38
rs374814242 Chr.19: 49558889-49558889 on Build GRCh38
rs909570443 Chr.19: 49558846-49558846 on Build GRCh38
rs753806135 Chr.19: 49558967-49558967 on Build GRCh38
rs188502013 Chr.19: 49559214-49559214 on Build GRCh38
rs888243909 Chr.19: 49559038-49559038 on Build GRCh38
rs778917118 Chr.19: 49559176-49559176 on Build GRCh38
rs191741403 Chr.19: 49559293-49559293 on Build GRCh38
rs372577019 Chr.19: 49558874-49558874 on Build GRCh38
rs980249765 Chr.19: 49559289-49559289 on Build GRCh38
rs752427783 Chr.19: 49558963-49558963 on Build GRCh38
rs981096095 Chr.19: 49559218-49559218 on Build GRCh38
rs751353745 Chr.19: 49558918-49558920 on Build GRCh38
rs548585453 Chr.19: 49559260-49559260 on Build GRCh38
rs1007519988 Chr.19: 49559055-49559055 on Build GRCh38
rs951016088 Chr.19: 49558777-49558777 on Build GRCh38
rs370070206 Chr.19: 49558908-49558908 on Build GRCh38
rs757220647 Chr.19: 49558989-49558989 on Build GRCh38
rs532378653 Chr.19: 49559148-49559148 on Build GRCh38
rs759258487 Chr.19: 49558914-49558914 on Build GRCh38
rs764747040 Chr.19: 49559254-49559254 on Build GRCh38
rs755682821 Chr.19: 49558929-49558929 on Build GRCh38
rs771615227 Chr.19: 49558782-49558782 on Build GRCh38
rs745590059 Chr.19: 49559004-49559004 on Build GRCh38
rs773752957 Chr.19: 49559016-49559016 on Build GRCh38
rs1017649597 Chr.19: 49558773-49558773 on Build GRCh38
rs781263509 Chr.19: 49559009-49559009 on Build GRCh38
rs574288559 Chr.19: 49559209-49559209 on Build GRCh38
rs1025381569 Chr.19: 49559127-49559127 on Build GRCh38
rs553629696 Chr.19: 49559240-49559240 on Build GRCh38
rs752257192 Chr.19: 49558920-49558920 on Build GRCh38
rs753349099 Chr.19: 49559029-49559029 on Build GRCh38
rs767268699 Chr.19: 49559024-49559024 on Build GRCh38
rs547090881 Chr.19: 49559099-49559099 on Build GRCh38
rs767023635 Chr.19: 49558916-49558916 on Build GRCh38
rs755229511 Chr.19: 49559005-49559005 on Build GRCh38
rs946031606 Chr.19: 49559018-49559018 on Build GRCh38

View Full Product Details