Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 125238571-125239076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA5
Entrez Gene ID
3309
Gene Name
heat shock protein family A (Hsp70) member 5
Gene Aliases
BIP, GRP78, HEL-S-89n, MIF2
UniGene
Hs.743241
Gene Chromosome Location
Chr.9: 125234848-125241387 on Build GRCh38
Transcripts
NM_005347.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3375079 c.903T>C p.S301S Chr.9: 125239034-125239034 on Build GRCh38
6189448 c.1110G>C p.K370N Chr.9: 125238714-125238714 on Build GRCh38
7152472 c.865C>T p.R289W Chr.9: 125239072-125239072 on Build GRCh38
6182296 c.1154C>G p.S385C Chr.9: 125238670-125238670 on Build GRCh38
487041 c.862C>G p.L288V Chr.9: 125239075-125239075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761352039 Chr.9: 125238857-125238857 on Build GRCh38
rs771120515 Chr.9: 125238844-125238844 on Build GRCh38
rs201819093 Chr.9: 125238974-125238974 on Build GRCh38
rs866597853 Chr.9: 125238964-125238964 on Build GRCh38
rs200526775 Chr.9: 125238647-125238647 on Build GRCh38
rs780605687 Chr.9: 125238831-125238832 on Build GRCh38
rs756951363 Chr.9: 125238609-125238609 on Build GRCh38
rs768548100 Chr.9: 125238812-125238812 on Build GRCh38
rs764555947 Chr.9: 125239048-125239048 on Build GRCh38
rs772811684 Chr.9: 125238941-125238941 on Build GRCh38
rs775362964 Chr.9: 125238639-125238639 on Build GRCh38
rs746186875 Chr.9: 125238985-125238985 on Build GRCh38
rs779884716 Chr.9: 125238805-125238805 on Build GRCh38
rs745612349 Chr.9: 125238624-125238624 on Build GRCh38
rs768638015 Chr.9: 125238661-125238661 on Build GRCh38
rs147872079 Chr.9: 125238702-125238702 on Build GRCh38
rs766767689 Chr.9: 125238716-125238716 on Build GRCh38
rs776753213 Chr.9: 125238982-125238982 on Build GRCh38
rs947809704 Chr.9: 125238950-125238950 on Build GRCh38
rs574075188 Chr.9: 125238801-125238801 on Build GRCh38
rs11542739 Chr.9: 125238790-125238790 on Build GRCh38
rs576419119 Chr.9: 125238908-125238908 on Build GRCh38
rs748080410 Chr.9: 125238929-125238929 on Build GRCh38
rs772238271 Chr.9: 125238694-125238694 on Build GRCh38
rs369271426 Chr.9: 125238866-125238866 on Build GRCh38
rs773236710 Chr.9: 125238841-125238841 on Build GRCh38
rs991681206 Chr.9: 125238940-125238940 on Build GRCh38
rs777034647 Chr.9: 125238850-125238850 on Build GRCh38
rs916098425 Chr.9: 125238967-125238967 on Build GRCh38
rs545601947 Chr.9: 125238999-125238999 on Build GRCh38
rs141506328 Chr.9: 125238789-125238789 on Build GRCh38
rs748174426 Chr.9: 125238822-125238822 on Build GRCh38
rs757316929 Chr.9: 125238860-125238864 on Build GRCh38
rs1046058280 Chr.9: 125238704-125238704 on Build GRCh38
rs763551125 Chr.9: 125239023-125239023 on Build GRCh38
rs776075958 Chr.9: 125239011-125239011 on Build GRCh38
rs1039991057 Chr.9: 125238836-125238836 on Build GRCh38
rs150434402 Chr.9: 125238943-125238943 on Build GRCh38
rs765987586 Chr.9: 125238891-125238891 on Build GRCh38
rs747246754 Chr.9: 125238842-125238842 on Build GRCh38
rs373962277 Chr.9: 125238786-125238786 on Build GRCh38
rs868145616 Chr.9: 125238673-125238673 on Build GRCh38
rs774390329 Chr.9: 125238667-125238667 on Build GRCh38
rs914981092 Chr.9: 125238893-125238893 on Build GRCh38
rs1045278040 Chr.9: 125239007-125239007 on Build GRCh38
rs868706078 Chr.9: 125239047-125239048 on Build GRCh38
rs374053540 Chr.9: 125238870-125238870 on Build GRCh38
rs377636827 Chr.9: 125238900-125238900 on Build GRCh38
rs760021194 Chr.9: 125238851-125238851 on Build GRCh38
rs962813421 Chr.9: 125238937-125238937 on Build GRCh38
rs749243565 Chr.9: 125238654-125238654 on Build GRCh38
rs935054387 Chr.9: 125238767-125238767 on Build GRCh38
rs769654200 Chr.9: 125238636-125238636 on Build GRCh38
rs1016994579 Chr.9: 125238922-125238922 on Build GRCh38
rs986689660 Chr.9: 125238714-125238714 on Build GRCh38
rs373901475 Chr.9: 125238852-125238852 on Build GRCh38
rs866181541 Chr.9: 125239046-125239047 on Build GRCh38
rs61755726 Chr.9: 125238825-125238825 on Build GRCh38
rs866590944 Chr.9: 125238707-125238707 on Build GRCh38
rs750103794 Chr.9: 125238800-125238800 on Build GRCh38
rs376077344 Chr.9: 125238798-125238798 on Build GRCh38
rs746945148 Chr.9: 125238711-125238711 on Build GRCh38
rs754225932 Chr.9: 125238729-125238729 on Build GRCh38
rs747110106 Chr.9: 125238960-125238960 on Build GRCh38
rs761919160 Chr.9: 125238685-125238685 on Build GRCh38
rs751174810 Chr.9: 125238602-125238602 on Build GRCh38
rs762582529 Chr.9: 125238966-125238966 on Build GRCh38
rs982129586 Chr.9: 125239031-125239031 on Build GRCh38
rs430397 Chr.9: 125238840-125238840 on Build GRCh38
rs760079687 Chr.9: 125238755-125238755 on Build GRCh38
rs776164280 Chr.9: 125238855-125238855 on Build GRCh38
rs778760844 Chr.9: 125238924-125238924 on Build GRCh38
rs542802064 Chr.9: 125238819-125238819 on Build GRCh38
rs201202868 Chr.9: 125238612-125238612 on Build GRCh38
rs200135468 Chr.9: 125238679-125238679 on Build GRCh38
rs749957636 Chr.9: 125238868-125238868 on Build GRCh38
rs760887016 Chr.9: 125238708-125238708 on Build GRCh38

View Full Product Details