Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 139350962-139351558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAA15
Entrez Gene ID
80155
Gene Name
N(alpha)-acetyltransferase 15, NatA auxiliary subunit
Gene Aliases
Ga19, NARG1, NAT1P, NATH, TBDN, TBDN100
UniGene
Hs.745047
Gene Chromosome Location
Chr.4: 139301467-139390781 on Build GRCh38
Transcripts
NM_057175.3 XM_005263236.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1051377 c.949A>C p.N317H Chr.4: 139351546-139351546 on Build GRCh38
1427187 c.815A>T p.N272I Chr.4: 139351194-139351194 on Build GRCh38
1719610 c.940C>T p.L314L Chr.4: 139351537-139351537 on Build GRCh38
4455846 c.924A>G p.E308E Chr.4: 139351521-139351521 on Build GRCh38
6011643 c.954C>T p.F318F Chr.4: 139351551-139351551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054312232 Chr.4: 139351357-139351357 on Build GRCh38
rs780163068 Chr.4: 139351143-139351143 on Build GRCh38
rs766438983 Chr.4: 139351325-139351325 on Build GRCh38
rs776335714 Chr.4: 139351437-139351437 on Build GRCh38
rs935414275 Chr.4: 139351332-139351332 on Build GRCh38
rs758429913 Chr.4: 139351193-139351193 on Build GRCh38
rs565254704 Chr.4: 139351452-139351452 on Build GRCh38
rs922189480 Chr.4: 139351211-139351211 on Build GRCh38
rs770286890 Chr.4: 139351310-139351310 on Build GRCh38
rs777529256 Chr.4: 139351226-139351226 on Build GRCh38
rs894440401 Chr.4: 139351096-139351096 on Build GRCh38
rs750474534 Chr.4: 139351171-139351171 on Build GRCh38
rs947378978 Chr.4: 139351136-139351136 on Build GRCh38
rs999433239 Chr.4: 139351220-139351220 on Build GRCh38
rs983635941 Chr.4: 139351136-139351137 on Build GRCh38
rs1028728044 Chr.4: 139351413-139351413 on Build GRCh38
rs746648645 Chr.4: 139351276-139351276 on Build GRCh38
rs11736015 Chr.4: 139351364-139351364 on Build GRCh38
rs1055900605 Chr.4: 139351087-139351087 on Build GRCh38
rs754916386 Chr.4: 139351464-139351464 on Build GRCh38
rs890322200 Chr.4: 139351155-139351170 on Build GRCh38
rs762325556 Chr.4: 139351146-139351146 on Build GRCh38
rs750324671 Chr.4: 139351506-139351506 on Build GRCh38
rs1012663772 Chr.4: 139351447-139351447 on Build GRCh38
rs972217633 Chr.4: 139351036-139351036 on Build GRCh38
rs757182339 Chr.4: 139351236-139351236 on Build GRCh38
rs768437780 Chr.4: 139351285-139351285 on Build GRCh38
rs796798038 Chr.4: 139351394-139351394 on Build GRCh38
rs940961035 Chr.4: 139351034-139351034 on Build GRCh38
rs78607223 Chr.4: 139351257-139351257 on Build GRCh38
rs202230897 Chr.4: 139351229-139351229 on Build GRCh38
rs768760652 Chr.4: 139351073-139351076 on Build GRCh38
rs529369697 Chr.4: 139351419-139351419 on Build GRCh38
rs376665519 Chr.4: 139351313-139351313 on Build GRCh38
rs371981648 Chr.4: 139351270-139351270 on Build GRCh38
rs779777784 Chr.4: 139351273-139351273 on Build GRCh38
rs767760353 Chr.4: 139351476-139351476 on Build GRCh38
rs762114041 Chr.4: 139351309-139351309 on Build GRCh38
rs778741013 Chr.4: 139351242-139351242 on Build GRCh38
rs776985851 Chr.4: 139351308-139351308 on Build GRCh38
rs369021673 Chr.4: 139351470-139351470 on Build GRCh38
rs766494727 Chr.4: 139351205-139351205 on Build GRCh38
rs763487215 Chr.4: 139351323-139351323 on Build GRCh38
rs564502007 Chr.4: 139351006-139351006 on Build GRCh38
rs547946962 Chr.4: 139351454-139351454 on Build GRCh38
rs756984620 Chr.4: 139351481-139351481 on Build GRCh38
rs770801370 Chr.4: 139351312-139351312 on Build GRCh38
rs902705854 Chr.4: 139351300-139351302 on Build GRCh38
rs781003731 Chr.4: 139351487-139351487 on Build GRCh38
rs751693020 Chr.4: 139351330-139351330 on Build GRCh38
rs745870672 Chr.4: 139351264-139351264 on Build GRCh38
rs541181417 Chr.4: 139351149-139351149 on Build GRCh38
rs901680614 Chr.4: 139351239-139351239 on Build GRCh38
rs938724521 Chr.4: 139351061-139351061 on Build GRCh38
rs373488592 Chr.4: 139351515-139351515 on Build GRCh38
rs377052939 Chr.4: 139351206-139351206 on Build GRCh38
rs770678718 Chr.4: 139351387-139351387 on Build GRCh38
rs569551157 Chr.4: 139351482-139351482 on Build GRCh38
rs561993597 Chr.4: 139351395-139351395 on Build GRCh38
rs928825158 Chr.4: 139351060-139351060 on Build GRCh38
rs546981635 Chr.4: 139351215-139351215 on Build GRCh38
rs762787385 Chr.4: 139351403-139351403 on Build GRCh38
rs763831546 Chr.4: 139351477-139351477 on Build GRCh38
rs950478713 Chr.4: 139351109-139351109 on Build GRCh38

View Full Product Details