Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133554215-133554716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL2
Entrez Gene ID
9719
Gene Name
ADAMTS like 2
Gene Aliases
GPHYSD1
UniGene
Hs.522543
Gene Chromosome Location
Chr.9: 133532164-133575519 on Build GRCh38
Transcripts
NM_001145320.1 NM_014694.3 XM_005272237.3 XM_005272238.3 XM_005272239.2 XM_011519241.2 XM_011519242.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6388387 c.1380G>T p.E460D Chr.9: 133554470-133554470 on Build GRCh38
6402526 c.1493C>T p.P498L Chr.9: 133554583-133554583 on Build GRCh38
4955564 c.1588G>A p.G530S Chr.9: 133554678-133554678 on Build GRCh38
4955563 c.1261G>A p.G421S Chr.9: 133554678-133554678 on Build GRCh38
6388386 c.1053G>T p.E351D Chr.9: 133554470-133554470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778189023 Chr.9: 133554654-133554654 on Build GRCh38
rs760098538 Chr.9: 133554513-133554513 on Build GRCh38
rs770463380 Chr.9: 133554536-133554536 on Build GRCh38
rs951307887 Chr.9: 133554615-133554615 on Build GRCh38
rs754962186 Chr.9: 133554455-133554455 on Build GRCh38
rs753606710 Chr.9: 133554423-133554423 on Build GRCh38
rs1028974722 Chr.9: 133554403-133554404 on Build GRCh38
rs1026897116 Chr.9: 133554485-133554485 on Build GRCh38
rs367726537 Chr.9: 133554567-133554567 on Build GRCh38
rs776340163 Chr.9: 133554512-133554512 on Build GRCh38
rs577516860 Chr.9: 133554442-133554442 on Build GRCh38
rs757684437 Chr.9: 133554691-133554691 on Build GRCh38
rs1052225145 Chr.9: 133554260-133554260 on Build GRCh38
rs974026101 Chr.9: 133554465-133554465 on Build GRCh38
rs1048312943 Chr.9: 133554273-133554273 on Build GRCh38
rs996554992 Chr.9: 133554396-133554396 on Build GRCh38
rs907919956 Chr.9: 133554296-133554296 on Build GRCh38
rs751779809 Chr.9: 133554400-133554400 on Build GRCh38
rs932262126 Chr.9: 133554404-133554404 on Build GRCh38
rs538102556 Chr.9: 133554395-133554396 on Build GRCh38
rs944709273 Chr.9: 133554337-133554337 on Build GRCh38
rs1044402763 Chr.9: 133554530-133554530 on Build GRCh38
rs886063638 Chr.9: 133554371-133554371 on Build GRCh38
rs753767762 Chr.9: 133554434-133554434 on Build GRCh38
rs995138068 Chr.9: 133554584-133554584 on Build GRCh38
rs936356646 Chr.9: 133554523-133554523 on Build GRCh38
rs969024836 Chr.9: 133554464-133554464 on Build GRCh38
rs886063639 Chr.9: 133554615-133554615 on Build GRCh38
rs35767802 Chr.9: 133554507-133554507 on Build GRCh38
rs904894177 Chr.9: 133554583-133554583 on Build GRCh38
rs754694330 Chr.9: 133554430-133554430 on Build GRCh38
rs757873971 Chr.9: 133554639-133554639 on Build GRCh38
rs981120741 Chr.9: 133554499-133554499 on Build GRCh38
rs984831879 Chr.9: 133554417-133554417 on Build GRCh38
rs762086890 Chr.9: 133554578-133554578 on Build GRCh38
rs369962641 Chr.9: 133554647-133554647 on Build GRCh38
rs925544120 Chr.9: 133554662-133554662 on Build GRCh38
rs1027321376 Chr.9: 133554587-133554587 on Build GRCh38
rs1000602352 Chr.9: 133554250-133554250 on Build GRCh38
rs868264505 Chr.9: 133554432-133554432 on Build GRCh38
rs1013366056 Chr.9: 133554575-133554575 on Build GRCh38
rs929824437 Chr.9: 133554258-133554258 on Build GRCh38
rs543220749 Chr.9: 133554555-133554555 on Build GRCh38
rs777497620 Chr.9: 133554492-133554492 on Build GRCh38
rs758444844 Chr.9: 133554554-133554583 on Build GRCh38
rs746439642 Chr.9: 133554506-133554506 on Build GRCh38
rs931990483 Chr.9: 133554651-133554651 on Build GRCh38
rs766276641 Chr.9: 133554416-133554416 on Build GRCh38
rs576039842 Chr.9: 133554550-133554550 on Build GRCh38
rs200335966 Chr.9: 133554291-133554291 on Build GRCh38
rs751807903 Chr.9: 133554576-133554576 on Build GRCh38
rs370480318 Chr.9: 133554491-133554491 on Build GRCh38
rs919778163 Chr.9: 133554255-133554255 on Build GRCh38
rs900778844 Chr.9: 133554395-133554395 on Build GRCh38
rs904068964 Chr.9: 133554541-133554541 on Build GRCh38
rs764918755 Chr.9: 133554458-133554458 on Build GRCh38
rs965162793 Chr.9: 133554642-133554642 on Build GRCh38
rs931980391 Chr.9: 133554518-133554518 on Build GRCh38
rs1019170377 Chr.9: 133554313-133554313 on Build GRCh38
rs922011097 Chr.9: 133554650-133554650 on Build GRCh38
rs199582731 Chr.9: 133554659-133554659 on Build GRCh38
rs1052592021 Chr.9: 133554554-133554554 on Build GRCh38
rs896196462 Chr.9: 133554443-133554443 on Build GRCh38
rs1040331678 Chr.9: 133554392-133554392 on Build GRCh38
rs35767802 Chr.9: 133554507-133554507 on Build GRCh38
rs538276028 Chr.9: 133554459-133554459 on Build GRCh38
rs904056906 Chr.9: 133554422-133554422 on Build GRCh38
rs944765676 Chr.9: 133554510-133554510 on Build GRCh38
rs770561616 Chr.9: 133554511-133554511 on Build GRCh38
rs983037163 Chr.9: 133554630-133554630 on Build GRCh38
rs752071055 Chr.9: 133554678-133554678 on Build GRCh38

View Full Product Details