Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 113452074-113452618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
UniGene
Hs.144114
Gene Chromosome Location
Chr.4: 113451032-113761927 on Build GRCh38
Transcripts
NM_001221.3 NM_001321566.1 NM_001321567.1 NM_001321568.1 NM_001321569.1 NM_001321570.1 NM_001321571.1 NM_001321572.1 NM_001321573.1 NM_001321574.1 NM_001321575.1 NM_001321576.1 NM_001321577.1 NM_001321578.1 NM_001321579.1 NM_001321580.1 NM_001321581.1 NM_001321582.1 NM_001321583.1 NM_001321584.1 NM_001321585.1 NM_001321586.1 NM_001321587.1 NM_001321588.1 NM_001321589.1 NM_001321590.1 NM_001321591.1 NM_172127.2 NM_172128.2 XM_005263253.3 XM_005263254.3 XM_006714330.2 XM_011532289.1 XM_011532290.1 XM_011532291.1 XM_011532292.2 XM_011532295.1 XM_017008672.1 XM_017008673.1 XM_017008674.1 XM_017008675.1 XM_017008676.1 XM_017008677.1 XM_017008678.1

SNP Detail

SNP ID Chromosome Location
rs952102845 Chr.4: 113452311-113452311 on Build GRCh38
rs1018585579 Chr.4: 113452337-113452348 on Build GRCh38
rs147249536 Chr.4: 113452298-113452298 on Build GRCh38
rs955129769 Chr.4: 113452186-113452186 on Build GRCh38
rs1044181505 Chr.4: 113452200-113452200 on Build GRCh38
rs562911826 Chr.4: 113452582-113452582 on Build GRCh38
rs997631870 Chr.4: 113452149-113452149 on Build GRCh38
rs555451101 Chr.4: 113452424-113452424 on Build GRCh38
rs368608352 Chr.4: 113452347-113452347 on Build GRCh38
rs575548410 Chr.4: 113452567-113452567 on Build GRCh38
rs10553216 Chr.4: 113452134-113452135 on Build GRCh38
rs967674799 Chr.4: 113452453-113452453 on Build GRCh38
rs796349006 Chr.4: 113452149-113452150 on Build GRCh38
rs929384062 Chr.4: 113452502-113452502 on Build GRCh38
rs750546724 Chr.4: 113452481-113452481 on Build GRCh38
rs397878493 Chr.4: 113452135-113452135 on Build GRCh38
rs1009104386 Chr.4: 113452241-113452241 on Build GRCh38
rs987229624 Chr.4: 113452199-113452200 on Build GRCh38
rs1019589469 Chr.4: 113452269-113452269 on Build GRCh38
rs992002730 Chr.4: 113452288-113452288 on Build GRCh38
rs544152195 Chr.4: 113452576-113452576 on Build GRCh38
rs148706700 Chr.4: 113452301-113452301 on Build GRCh38
rs888056116 Chr.4: 113452096-113452096 on Build GRCh38
rs887739477 Chr.4: 113452316-113452317 on Build GRCh38
rs555682889 Chr.4: 113452394-113452394 on Build GRCh38
rs182697945 Chr.4: 113452386-113452386 on Build GRCh38
rs62314973 Chr.4: 113452135-113452135 on Build GRCh38
rs939310353 Chr.4: 113452366-113452366 on Build GRCh38
rs3733619 Chr.4: 113452099-113452099 on Build GRCh38
rs550796955 Chr.4: 113452271-113452271 on Build GRCh38
rs34030509 Chr.4: 113452133-113452134 on Build GRCh38
rs1013134906 Chr.4: 113452198-113452198 on Build GRCh38
rs191496065 Chr.4: 113452487-113452487 on Build GRCh38
rs1052761682 Chr.4: 113452462-113452462 on Build GRCh38
rs907878763 Chr.4: 113452365-113452365 on Build GRCh38
rs967977777 Chr.4: 113452217-113452217 on Build GRCh38
rs918018879 Chr.4: 113452294-113452294 on Build GRCh38
rs1033635842 Chr.4: 113452477-113452477 on Build GRCh38
rs992756853 Chr.4: 113452377-113452377 on Build GRCh38
rs900644718 Chr.4: 113452107-113452107 on Build GRCh38
rs111656928 Chr.4: 113452131-113452131 on Build GRCh38
rs900750576 Chr.4: 113452486-113452486 on Build GRCh38
rs917730229 Chr.4: 113452500-113452500 on Build GRCh38
rs533578881 Chr.4: 113452150-113452151 on Build GRCh38
rs757975113 Chr.4: 113452106-113452107 on Build GRCh38
rs562950191 Chr.4: 113452471-113452471 on Build GRCh38
rs535693611 Chr.4: 113452175-113452175 on Build GRCh38
rs955669559 Chr.4: 113452236-113452236 on Build GRCh38
rs186683405 Chr.4: 113452391-113452391 on Build GRCh38
rs890881098 Chr.4: 113452417-113452417 on Build GRCh38
rs796187485 Chr.4: 113452134-113452135 on Build GRCh38
rs904304437 Chr.4: 113452445-113452445 on Build GRCh38
rs556223150 Chr.4: 113452195-113452195 on Build GRCh38
rs767485627 Chr.4: 113452468-113452468 on Build GRCh38
rs573893837 Chr.4: 113452409-113452409 on Build GRCh38
rs904252727 Chr.4: 113452204-113452204 on Build GRCh38
rs555383293 Chr.4: 113452172-113452181 on Build GRCh38
rs191652913 Chr.4: 113452352-113452352 on Build GRCh38
rs983520612 Chr.4: 113452562-113452562 on Build GRCh38
rs942075421 Chr.4: 113452593-113452593 on Build GRCh38
rs999856530 Chr.4: 113452216-113452216 on Build GRCh38
rs979305292 Chr.4: 113452253-113452253 on Build GRCh38
rs1031288946 Chr.4: 113452226-113452226 on Build GRCh38
rs142458960 Chr.4: 113452451-113452451 on Build GRCh38
rs530683234 Chr.4: 113452591-113452591 on Build GRCh38
rs1039295238 Chr.4: 113452337-113452337 on Build GRCh38
rs964937923 Chr.4: 113452283-113452283 on Build GRCh38

View Full Product Details