Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69281385-69281892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC125
Entrez Gene ID
202243
Gene Name
coiled-coil domain containing 125
Gene Aliases
KENAE
UniGene
Hs.654850
Gene Chromosome Location
Chr.5: 69273087-69333069 on Build GRCh38
Transcripts
NM_001297696.1 NM_001297697.1 NM_176816.4 XM_005248458.4 XM_005248459.4 XM_005248461.3 XM_005248463.3 XM_006714569.3 XM_006714570.3 XM_011543255.2 XM_011543256.2 XM_011543257.2 XM_011543258.2 XM_011543259.2 XM_017009206.1 XM_017009207.1 XM_017009208.1 XM_017009209.1 XM_017009210.1 XM_017009211.1

SNP Detail

SNP ID Chromosome Location
rs539192233 Chr.5: 69281653-69281653 on Build GRCh38
rs763806153 Chr.5: 69281745-69281746 on Build GRCh38
rs1029303106 Chr.5: 69281828-69281828 on Build GRCh38
rs1043110845 Chr.5: 69281498-69281498 on Build GRCh38
rs924650163 Chr.5: 69281517-69281517 on Build GRCh38
rs922492267 Chr.5: 69281428-69281428 on Build GRCh38
rs756560167 Chr.5: 69281512-69281513 on Build GRCh38
rs745374773 Chr.5: 69281660-69281660 on Build GRCh38
rs189083350 Chr.5: 69281667-69281667 on Build GRCh38
rs1015338630 Chr.5: 69281704-69281704 on Build GRCh38
rs1057340276 Chr.5: 69281545-69281545 on Build GRCh38
rs942988034 Chr.5: 69281696-69281700 on Build GRCh38
rs973852301 Chr.5: 69281596-69281596 on Build GRCh38
rs904031455 Chr.5: 69281769-69281769 on Build GRCh38
rs577384368 Chr.5: 69281583-69281583 on Build GRCh38
rs184643439 Chr.5: 69281774-69281774 on Build GRCh38
rs573514599 Chr.5: 69281462-69281462 on Build GRCh38
rs915055748 Chr.5: 69281488-69281488 on Build GRCh38
rs12651858 Chr.5: 69281640-69281640 on Build GRCh38
rs997835821 Chr.5: 69281783-69281783 on Build GRCh38
rs560809267 Chr.5: 69281718-69281718 on Build GRCh38
rs934660720 Chr.5: 69281541-69281541 on Build GRCh38
rs191600758 Chr.5: 69281691-69281691 on Build GRCh38
rs377435414 Chr.5: 69281451-69281451 on Build GRCh38
rs192679776 Chr.5: 69281492-69281492 on Build GRCh38
rs961053504 Chr.5: 69281756-69281756 on Build GRCh38
rs1052542985 Chr.5: 69281826-69281826 on Build GRCh38
rs538383753 Chr.5: 69281842-69281842 on Build GRCh38
rs953961508 Chr.5: 69281869-69281869 on Build GRCh38
rs971329139 Chr.5: 69281495-69281495 on Build GRCh38
rs528275551 Chr.5: 69281723-69281723 on Build GRCh38
rs936815448 Chr.5: 69281791-69281795 on Build GRCh38
rs114133299 Chr.5: 69281567-69281567 on Build GRCh38
rs757641690 Chr.5: 69281826-69281851 on Build GRCh38
rs985710154 Chr.5: 69281463-69281463 on Build GRCh38
rs918308778 Chr.5: 69281625-69281629 on Build GRCh38
rs1013141869 Chr.5: 69281570-69281570 on Build GRCh38
rs184290407 Chr.5: 69281592-69281592 on Build GRCh38
rs571027099 Chr.5: 69281805-69281805 on Build GRCh38

View Full Product Details