Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113227964-113228476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL4A
Entrez Gene ID
8451
Gene Name
cullin 4A
Gene Aliases
-
UniGene
Hs.339735
Gene Chromosome Location
Chr.13: 113208193-113265078 on Build GRCh38
Transcripts
NM_001008895.2 NM_001278513.1 NM_001278514.1 NM_003589.2 XM_011537523.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6531823 c.413G>C p.C138S Chr.13: 113228020-113228020 on Build GRCh38
3688627 c.123C>T p.D41D Chr.13: 113228030-113228030 on Build GRCh38
3688626 c.423C>T p.D141D Chr.13: 113228030-113228030 on Build GRCh38
1579217 c.112T>G p.C38G Chr.13: 113228019-113228019 on Build GRCh38
6531824 c.113G>C p.C38S Chr.13: 113228020-113228020 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144456648 Chr.13: 113228280-113228280 on Build GRCh38
rs780464106 Chr.13: 113228163-113228163 on Build GRCh38
rs1009824639 Chr.13: 113228066-113228066 on Build GRCh38
rs771004997 Chr.13: 113228072-113228072 on Build GRCh38
rs1033558546 Chr.13: 113228419-113228419 on Build GRCh38
rs774417744 Chr.13: 113228075-113228075 on Build GRCh38
rs200015519 Chr.13: 113228025-113228025 on Build GRCh38
rs1009381421 Chr.13: 113228378-113228378 on Build GRCh38
rs777653049 Chr.13: 113228054-113228054 on Build GRCh38
rs151121982 Chr.13: 113228298-113228298 on Build GRCh38
rs917327922 Chr.13: 113228174-113228174 on Build GRCh38
rs967957926 Chr.13: 113228099-113228099 on Build GRCh38
rs940675670 Chr.13: 113228088-113228088 on Build GRCh38
rs920821341 Chr.13: 113228161-113228161 on Build GRCh38
rs189426451 Chr.13: 113228103-113228103 on Build GRCh38
rs747621457 Chr.13: 113228447-113228447 on Build GRCh38
rs757768264 Chr.13: 113228017-113228017 on Build GRCh38
rs370987328 Chr.13: 113228084-113228084 on Build GRCh38
rs780743841 Chr.13: 113228036-113228036 on Build GRCh38
rs958777798 Chr.13: 113228164-113228164 on Build GRCh38
rs745985279 Chr.13: 113228097-113228097 on Build GRCh38
rs950216667 Chr.13: 113228204-113228204 on Build GRCh38
rs751132176 Chr.13: 113228021-113228021 on Build GRCh38
rs1038034372 Chr.13: 113228274-113228274 on Build GRCh38
rs987282504 Chr.13: 113228363-113228363 on Build GRCh38
rs1042640033 Chr.13: 113228379-113228379 on Build GRCh38
rs895971661 Chr.13: 113228200-113228200 on Build GRCh38
rs769264884 Chr.13: 113228371-113228371 on Build GRCh38
rs1056099565 Chr.13: 113228199-113228199 on Build GRCh38
rs908775925 Chr.13: 113228242-113228242 on Build GRCh38
rs747811553 Chr.13: 113228042-113228042 on Build GRCh38
rs543907096 Chr.13: 113228396-113228396 on Build GRCh38
rs954514723 Chr.13: 113228290-113228290 on Build GRCh38
rs543860146 Chr.13: 113228344-113228344 on Build GRCh38
rs899539616 Chr.13: 113228275-113228275 on Build GRCh38
rs555231934 Chr.13: 113228223-113228223 on Build GRCh38
rs2025247 Chr.13: 113228250-113228250 on Build GRCh38
rs1032414306 Chr.13: 113228273-113228273 on Build GRCh38
rs765677995 Chr.13: 113228020-113228020 on Build GRCh38
rs756369838 Chr.13: 113228137-113228137 on Build GRCh38
rs181966705 Chr.13: 113228373-113228373 on Build GRCh38
rs903769934 Chr.13: 113228384-113228384 on Build GRCh38
rs778607338 Chr.13: 113228445-113228445 on Build GRCh38
rs902279809 Chr.13: 113228229-113228230 on Build GRCh38
rs749023680 Chr.13: 113228059-113228059 on Build GRCh38
rs755671736 Chr.13: 113228044-113228044 on Build GRCh38
rs867197733 Chr.13: 113228173-113228173 on Build GRCh38
rs754476192 Chr.13: 113228024-113228024 on Build GRCh38
rs937570992 Chr.13: 113228193-113228193 on Build GRCh38
rs113697945 Chr.13: 113227993-113227993 on Build GRCh38
rs1033005241 Chr.13: 113228121-113228121 on Build GRCh38

View Full Product Details