Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137390600-137391100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKI
Entrez Gene ID
9162
Gene Name
diacylglycerol kinase iota
Gene Aliases
DGK-IOTA
UniGene
Hs.242947
Gene Chromosome Location
Chr.7: 137384283-137846864 on Build GRCh38
Transcripts
NM_001321708.1 NM_001321709.1 NM_001321710.1 NM_004717.3 XM_011516684.2 XM_017012780.1 XM_017012781.1 XM_017012782.1 XM_017012783.1 XM_017012784.1 XM_017012785.1 XM_017012786.1 XM_017012787.1 XM_017012788.1 XM_017012789.1

SNP Detail

SNP ID Chromosome Location
rs540614689 Chr.7: 137390953-137390953 on Build GRCh38
rs544894886 Chr.7: 137390895-137390895 on Build GRCh38
rs779496972 Chr.7: 137390926-137390926 on Build GRCh38
rs577769505 Chr.7: 137390709-137390709 on Build GRCh38
rs75146337 Chr.7: 137391065-137391065 on Build GRCh38
rs184341737 Chr.7: 137390864-137390864 on Build GRCh38
rs573587647 Chr.7: 137391052-137391052 on Build GRCh38
rs747944363 Chr.7: 137390994-137390994 on Build GRCh38
rs1035330576 Chr.7: 137390816-137390816 on Build GRCh38
rs530810119 Chr.7: 137390876-137390876 on Build GRCh38
rs891110928 Chr.7: 137390677-137390677 on Build GRCh38
rs1023112508 Chr.7: 137390999-137390999 on Build GRCh38
rs180913379 Chr.7: 137391015-137391015 on Build GRCh38
rs891017454 Chr.7: 137390954-137390954 on Build GRCh38
rs913657766 Chr.7: 137390857-137390857 on Build GRCh38
rs963082235 Chr.7: 137390766-137390766 on Build GRCh38
rs939034308 Chr.7: 137390849-137390850 on Build GRCh38
rs181538870 Chr.7: 137390812-137390812 on Build GRCh38
rs775146751 Chr.7: 137390841-137390841 on Build GRCh38
rs987734478 Chr.7: 137390856-137390856 on Build GRCh38
rs1043602125 Chr.7: 137390902-137390902 on Build GRCh38
rs1015697718 Chr.7: 137390718-137390718 on Build GRCh38
rs758066548 Chr.7: 137390815-137390815 on Build GRCh38
rs1007989073 Chr.7: 137390697-137390697 on Build GRCh38
rs377156094 Chr.7: 137390809-137390809 on Build GRCh38
rs934401309 Chr.7: 137390627-137390627 on Build GRCh38
rs943935908 Chr.7: 137390928-137390928 on Build GRCh38
rs745652984 Chr.7: 137390631-137390631 on Build GRCh38
rs952184713 Chr.7: 137390796-137390796 on Build GRCh38
rs754564291 Chr.7: 137391037-137391037 on Build GRCh38
rs901932822 Chr.7: 137390734-137390734 on Build GRCh38
rs118161928 Chr.7: 137391068-137391068 on Build GRCh38
rs114364439 Chr.7: 137390958-137390958 on Build GRCh38
rs988519453 Chr.7: 137390842-137390842 on Build GRCh38
rs764443686 Chr.7: 137390713-137390713 on Build GRCh38
rs887969679 Chr.7: 137390979-137390979 on Build GRCh38
rs914433995 Chr.7: 137390850-137390850 on Build GRCh38
rs768170353 Chr.7: 137391001-137391001 on Build GRCh38
rs995988280 Chr.7: 137390988-137390988 on Build GRCh38
rs571921283 Chr.7: 137390632-137390632 on Build GRCh38
rs947339232 Chr.7: 137390633-137390633 on Build GRCh38
rs141022432 Chr.7: 137390788-137390788 on Build GRCh38
rs137996512 Chr.7: 137390665-137390666 on Build GRCh38
rs567048684 Chr.7: 137391026-137391026 on Build GRCh38
rs1035911663 Chr.7: 137391016-137391016 on Build GRCh38
rs561790130 Chr.7: 137390865-137390865 on Build GRCh38
rs1040346135 Chr.7: 137390700-137390700 on Build GRCh38
rs1004222490 Chr.7: 137390955-137390955 on Build GRCh38
rs748765965 Chr.7: 137390938-137390938 on Build GRCh38
rs997162311 Chr.7: 137390652-137390652 on Build GRCh38
rs889144756 Chr.7: 137390673-137390673 on Build GRCh38
rs1006507762 Chr.7: 137390798-137390798 on Build GRCh38
rs955987380 Chr.7: 137390840-137390840 on Build GRCh38
rs929410912 Chr.7: 137390946-137390946 on Build GRCh38
rs901839347 Chr.7: 137390944-137390944 on Build GRCh38
rs1049617554 Chr.7: 137390670-137390670 on Build GRCh38
rs188648614 Chr.7: 137390925-137390925 on Build GRCh38
rs993556268 Chr.7: 137390743-137390743 on Build GRCh38

View Full Product Details