Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17102028-17102535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17RA
Entrez Gene ID
23765
Gene Name
interleukin 17 receptor A
Gene Aliases
CANDF5, CD217, CDw217, IL-17RA, IL17R, hIL-17R
UniGene
Hs.48353
Gene Chromosome Location
Chr.22: 17084959-17115694 on Build GRCh38
Transcripts
NM_001289905.1 NM_014339.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1031832 c.762+2T>C p.? Chr.22: 17102304-17102304 on Build GRCh38
6350813 c.655G>T p.V219L Chr.22: 17102195-17102195 on Build GRCh38
5858855 c.713C>T p.P238L Chr.22: 17102253-17102253 on Build GRCh38
6559237 c.608G>T p.W203L Chr.22: 17102148-17102148 on Build GRCh38
725691 c.631A>T p.T211S Chr.22: 17102171-17102171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773889030 Chr.22: 17102314-17102314 on Build GRCh38
rs748859963 Chr.22: 17102301-17102301 on Build GRCh38
rs371674761 Chr.22: 17102215-17102215 on Build GRCh38
rs779662031 Chr.22: 17102298-17102298 on Build GRCh38
rs41332051 Chr.22: 17102096-17102096 on Build GRCh38
rs192318930 Chr.22: 17102416-17102416 on Build GRCh38
rs975235215 Chr.22: 17102063-17102063 on Build GRCh38
rs564498681 Chr.22: 17102380-17102380 on Build GRCh38
rs781768362 Chr.22: 17102102-17102104 on Build GRCh38
rs781306366 Chr.22: 17102079-17102079 on Build GRCh38
rs200302492 Chr.22: 17102184-17102184 on Build GRCh38
rs140868574 Chr.22: 17102261-17102261 on Build GRCh38
rs556317837 Chr.22: 17102288-17102288 on Build GRCh38
rs375647844 Chr.22: 17102058-17102058 on Build GRCh38
rs138105435 Chr.22: 17102406-17102406 on Build GRCh38
rs774999877 Chr.22: 17102197-17102197 on Build GRCh38
rs746066804 Chr.22: 17102080-17102080 on Build GRCh38
rs1046787585 Chr.22: 17102174-17102174 on Build GRCh38
rs950193347 Chr.22: 17102411-17102411 on Build GRCh38
rs756290540 Chr.22: 17102084-17102084 on Build GRCh38
rs369409494 Chr.22: 17102143-17102143 on Build GRCh38
rs200114306 Chr.22: 17102164-17102164 on Build GRCh38
rs746263912 Chr.22: 17102057-17102057 on Build GRCh38
rs144085995 Chr.22: 17102216-17102216 on Build GRCh38
rs768313727 Chr.22: 17102304-17102304 on Build GRCh38
rs560542290 Chr.22: 17102408-17102408 on Build GRCh38
rs183650149 Chr.22: 17102068-17102068 on Build GRCh38
rs377570438 Chr.22: 17102308-17102308 on Build GRCh38
rs908261847 Chr.22: 17102436-17102436 on Build GRCh38
rs747027956 Chr.22: 17102114-17102114 on Build GRCh38
rs899495549 Chr.22: 17102443-17102443 on Build GRCh38
rs780688927 Chr.22: 17102389-17102389 on Build GRCh38
rs780107646 Chr.22: 17102259-17102259 on Build GRCh38
rs1020357111 Chr.22: 17102324-17102324 on Build GRCh38
rs531498499 Chr.22: 17102087-17102087 on Build GRCh38
rs769414039 Chr.22: 17102291-17102291 on Build GRCh38
rs753988382 Chr.22: 17102249-17102249 on Build GRCh38
rs753999184 Chr.22: 17102133-17102133 on Build GRCh38
rs146464762 Chr.22: 17102254-17102254 on Build GRCh38
rs553774032 Chr.22: 17102124-17102124 on Build GRCh38
rs746204344 Chr.22: 17102447-17102447 on Build GRCh38
rs539545714 Chr.22: 17102082-17102087 on Build GRCh38
rs531611881 Chr.22: 17102450-17102450 on Build GRCh38
rs762025728 Chr.22: 17102192-17102192 on Build GRCh38
rs758299842 Chr.22: 17102242-17102242 on Build GRCh38
rs773697918 Chr.22: 17102341-17102341 on Build GRCh38
rs761221996 Chr.22: 17102346-17102346 on Build GRCh38
rs750935138 Chr.22: 17102232-17102232 on Build GRCh38
rs913348507 Chr.22: 17102103-17102103 on Build GRCh38
rs746893415 Chr.22: 17102182-17102182 on Build GRCh38
rs186541010 Chr.22: 17102195-17102195 on Build GRCh38
rs763158886 Chr.22: 17102128-17102128 on Build GRCh38
rs758810829 Chr.22: 17102062-17102062 on Build GRCh38
rs41444249 Chr.22: 17102122-17102122 on Build GRCh38
rs41537347 Chr.22: 17102438-17102438 on Build GRCh38
rs756200741 Chr.22: 17102159-17102159 on Build GRCh38
rs758116776 Chr.22: 17102173-17102173 on Build GRCh38
rs371494126 Chr.22: 17102219-17102219 on Build GRCh38
rs372584256 Chr.22: 17102285-17102285 on Build GRCh38
rs775651309 Chr.22: 17102367-17102368 on Build GRCh38
rs757277204 Chr.22: 17102076-17102076 on Build GRCh38
rs760741652 Chr.22: 17102245-17102245 on Build GRCh38
rs958771050 Chr.22: 17102390-17102390 on Build GRCh38
rs762431722 Chr.22: 17102140-17102140 on Build GRCh38
rs374891419 Chr.22: 17102246-17102246 on Build GRCh38
rs140367455 Chr.22: 17102193-17102193 on Build GRCh38
rs757772567 Chr.22: 17102100-17102100 on Build GRCh38
rs558799480 Chr.22: 17102181-17102181 on Build GRCh38
rs534512644 Chr.22: 17102165-17102165 on Build GRCh38
rs145541428 Chr.22: 17102322-17102322 on Build GRCh38
rs570076897 Chr.22: 17102134-17102134 on Build GRCh38
rs1038157688 Chr.22: 17102442-17102442 on Build GRCh38
rs550044753 Chr.22: 17102483-17102483 on Build GRCh38
rs773021948 Chr.22: 17102108-17102108 on Build GRCh38
rs978722497 Chr.22: 17102357-17102357 on Build GRCh38
rs750454090 Chr.22: 17102157-17102157 on Build GRCh38
rs747627070 Chr.22: 17102391-17102391 on Build GRCh38
rs752109820 Chr.22: 17102071-17102071 on Build GRCh38
rs760725110 Chr.22: 17102121-17102121 on Build GRCh38
rs755272262 Chr.22: 17102253-17102253 on Build GRCh38
rs762107871 Chr.22: 17102218-17102218 on Build GRCh38
rs546133354 Chr.22: 17102417-17102417 on Build GRCh38
rs770329430 Chr.22: 17102118-17102118 on Build GRCh38
rs780276810 Chr.22: 17102088-17102088 on Build GRCh38
rs369259606 Chr.22: 17102255-17102255 on Build GRCh38
rs140255319 Chr.22: 17102097-17102097 on Build GRCh38

View Full Product Details