Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95512759-95513261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METAP2
Entrez Gene ID
10988
Gene Name
methionyl aminopeptidase 2
Gene Aliases
MAP2, MNPEP, p67eIF2
UniGene
Hs.444986
Gene Chromosome Location
Chr.12: 95474046-95515839 on Build GRCh38
Transcripts
NM_001317182.1 NM_001317183.1 NM_006838.3 NR_133673.1 XM_005268583.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6365527 c.1085C>A p.A362E Chr.12: 95512817-95512817 on Build GRCh38
5470107 c.1080A>G p.V360V Chr.12: 95512812-95512812 on Build GRCh38
6711560 c.1142C>G p.S381* Chr.12: 95512874-95512874 on Build GRCh38
219981 c.1163A>T p.D388V Chr.12: 95512895-95512895 on Build GRCh38
3998989 c.1095C>A p.T365T Chr.12: 95512827-95512827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59022378 Chr.12: 95513092-95513092 on Build GRCh38
rs868375084 Chr.12: 95512872-95512872 on Build GRCh38
rs368326520 Chr.12: 95513089-95513090 on Build GRCh38
rs1031012401 Chr.12: 95513151-95513151 on Build GRCh38
rs199541160 Chr.12: 95512926-95512926 on Build GRCh38
rs938364661 Chr.12: 95513048-95513048 on Build GRCh38
rs753774762 Chr.12: 95512803-95512803 on Build GRCh38
rs373779339 Chr.12: 95513047-95513048 on Build GRCh38
rs149247945 Chr.12: 95512818-95512818 on Build GRCh38
rs982479218 Chr.12: 95512990-95512990 on Build GRCh38
rs1006444828 Chr.12: 95513075-95513075 on Build GRCh38
rs3794261 Chr.12: 95512827-95512827 on Build GRCh38
rs900738622 Chr.12: 95513090-95513090 on Build GRCh38
rs527743708 Chr.12: 95513118-95513118 on Build GRCh38
rs60788079 Chr.12: 95513130-95513133 on Build GRCh38
rs758870642 Chr.12: 95512786-95512789 on Build GRCh38
rs751082505 Chr.12: 95513091-95513092 on Build GRCh38
rs915834615 Chr.12: 95513057-95513057 on Build GRCh38
rs938425250 Chr.12: 95513052-95513052 on Build GRCh38
rs137949478 Chr.12: 95512830-95512830 on Build GRCh38
rs909512407 Chr.12: 95513136-95513136 on Build GRCh38
rs778867474 Chr.12: 95513017-95513017 on Build GRCh38
rs1008230389 Chr.12: 95513100-95513100 on Build GRCh38
rs754497638 Chr.12: 95512925-95512925 on Build GRCh38
rs780588841 Chr.12: 95512931-95512934 on Build GRCh38
rs771151741 Chr.12: 95512957-95512957 on Build GRCh38
rs1027133919 Chr.12: 95513141-95513143 on Build GRCh38
rs301029 Chr.12: 95513220-95513220 on Build GRCh38
rs954811764 Chr.12: 95513134-95513134 on Build GRCh38
rs68112561 Chr.12: 95513132-95513133 on Build GRCh38
rs200820149 Chr.12: 95512930-95512930 on Build GRCh38
rs371041952 Chr.12: 95513091-95513094 on Build GRCh38
rs778233340 Chr.12: 95512942-95512942 on Build GRCh38
rs116440422 Chr.12: 95513065-95513065 on Build GRCh38
rs773459881 Chr.12: 95513090-95513093 on Build GRCh38
rs369020696 Chr.12: 95513009-95513009 on Build GRCh38
rs991180552 Chr.12: 95513235-95513235 on Build GRCh38
rs902998689 Chr.12: 95513080-95513080 on Build GRCh38
rs1052533537 Chr.12: 95513090-95513091 on Build GRCh38
rs201546458 Chr.12: 95512905-95512905 on Build GRCh38
rs529826073 Chr.12: 95512901-95512901 on Build GRCh38
rs202010352 Chr.12: 95512897-95512897 on Build GRCh38
rs914503561 Chr.12: 95513227-95513227 on Build GRCh38
rs760430966 Chr.12: 95513090-95513091 on Build GRCh38
rs1030498938 Chr.12: 95513130-95513130 on Build GRCh38
rs763907822 Chr.12: 95513092-95513093 on Build GRCh38
rs764009609 Chr.12: 95512912-95512912 on Build GRCh38
rs377493820 Chr.12: 95512792-95512795 on Build GRCh38
rs1038328664 Chr.12: 95513084-95513084 on Build GRCh38
rs1047392634 Chr.12: 95513054-95513054 on Build GRCh38
rs764942287 Chr.12: 95512815-95512815 on Build GRCh38
rs964360202 Chr.12: 95513130-95513132 on Build GRCh38
rs770428790 Chr.12: 95512931-95512931 on Build GRCh38
rs757419916 Chr.12: 95513223-95513223 on Build GRCh38
rs200335324 Chr.12: 95512899-95512899 on Build GRCh38
rs542225048 Chr.12: 95513036-95513036 on Build GRCh38
rs138357368 Chr.12: 95512811-95512811 on Build GRCh38
rs774484167 Chr.12: 95512958-95512958 on Build GRCh38
rs867169872 Chr.12: 95512874-95512874 on Build GRCh38
rs779606861 Chr.12: 95512849-95512849 on Build GRCh38
rs1042931333 Chr.12: 95513066-95513066 on Build GRCh38
rs761589608 Chr.12: 95512806-95512806 on Build GRCh38
rs56936153 Chr.12: 95513089-95513089 on Build GRCh38
rs891191416 Chr.12: 95513098-95513098 on Build GRCh38
rs201404188 Chr.12: 95512949-95512949 on Build GRCh38
rs546809638 Chr.12: 95513150-95513150 on Build GRCh38
rs934573277 Chr.12: 95512824-95512824 on Build GRCh38
rs377202702 Chr.12: 95512952-95512952 on Build GRCh38

View Full Product Details