Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34113504-34114017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAT10
Entrez Gene ID
55226
Gene Name
N-acetyltransferase 10
Gene Aliases
ALP, NET43
UniGene
Hs.577281
Gene Chromosome Location
Chr.11: 34105564-34146911 on Build GRCh38
Transcripts
NM_001144030.1 NM_024662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4705521 c.495delG p.D166fs*70 Chr.11: 34113838-34113838 on Build GRCh38
4705520 c.403G>A p.A135T Chr.11: 34113746-34113746 on Build GRCh38
4495055 c.444C>T p.V148V Chr.11: 34113787-34113787 on Build GRCh38
926601 c.454C>T p.R152W Chr.11: 34113797-34113797 on Build GRCh38
3446934 c.495+1G>A p.? Chr.11: 34113839-34113839 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542003872 Chr.11: 34113596-34113596 on Build GRCh38
rs1044613274 Chr.11: 34113790-34113790 on Build GRCh38
rs776572548 Chr.11: 34113701-34113701 on Build GRCh38
rs918593367 Chr.11: 34113746-34113746 on Build GRCh38
rs767655683 Chr.11: 34113718-34113718 on Build GRCh38
rs551246719 Chr.11: 34113829-34113829 on Build GRCh38
rs752854377 Chr.11: 34113724-34113724 on Build GRCh38
rs765691041 Chr.11: 34113878-34113878 on Build GRCh38
rs779256457 Chr.11: 34113773-34113773 on Build GRCh38
rs776521063 Chr.11: 34113811-34113811 on Build GRCh38
rs759506616 Chr.11: 34113714-34113714 on Build GRCh38
rs987254552 Chr.11: 34113916-34113916 on Build GRCh38
rs775276900 Chr.11: 34113699-34113699 on Build GRCh38
rs987307095 Chr.11: 34113640-34113640 on Build GRCh38
rs948794647 Chr.11: 34113590-34113590 on Build GRCh38
rs1026188086 Chr.11: 34113875-34113875 on Build GRCh38
rs757832661 Chr.11: 34113780-34113780 on Build GRCh38
rs966463635 Chr.11: 34113655-34113656 on Build GRCh38
rs769674662 Chr.11: 34113828-34113828 on Build GRCh38
rs766122915 Chr.11: 34113637-34113639 on Build GRCh38
rs1013280425 Chr.11: 34113611-34113611 on Build GRCh38
rs74471251 Chr.11: 34113802-34113802 on Build GRCh38
rs771369897 Chr.11: 34113669-34113669 on Build GRCh38
rs760835437 Chr.11: 34113858-34113858 on Build GRCh38
rs1014412725 Chr.11: 34113737-34113737 on Build GRCh38
rs775574075 Chr.11: 34113847-34113847 on Build GRCh38
rs757687750 Chr.11: 34113764-34113764 on Build GRCh38
rs763009060 Chr.11: 34113832-34113832 on Build GRCh38
rs539842436 Chr.11: 34113530-34113530 on Build GRCh38
rs768456050 Chr.11: 34113808-34113808 on Build GRCh38
rs778068232 Chr.11: 34113736-34113736 on Build GRCh38
rs548166021 Chr.11: 34113637-34113637 on Build GRCh38
rs764276204 Chr.11: 34113871-34113871 on Build GRCh38
rs554726356 Chr.11: 34113615-34113615 on Build GRCh38
rs574581978 Chr.11: 34113630-34113630 on Build GRCh38
rs963393046 Chr.11: 34113571-34113571 on Build GRCh38
rs201730594 Chr.11: 34113726-34113726 on Build GRCh38
rs112626590 Chr.11: 34113918-34113918 on Build GRCh38
rs576569982 Chr.11: 34113637-34113638 on Build GRCh38
rs767514829 Chr.11: 34113836-34113836 on Build GRCh38
rs531934795 Chr.11: 34113686-34113686 on Build GRCh38
rs563265100 Chr.11: 34113679-34113679 on Build GRCh38
rs1015902792 Chr.11: 34113529-34113529 on Build GRCh38
rs543280700 Chr.11: 34113653-34113653 on Build GRCh38
rs149812318 Chr.11: 34113797-34113797 on Build GRCh38
rs745346952 Chr.11: 34113685-34113685 on Build GRCh38
rs758979168 Chr.11: 34113885-34113885 on Build GRCh38
rs778435054 Chr.11: 34113676-34113676 on Build GRCh38
rs919184986 Chr.11: 34113593-34113593 on Build GRCh38
rs943497358 Chr.11: 34113657-34113657 on Build GRCh38
rs765111741 Chr.11: 34113704-34113704 on Build GRCh38
rs930040317 Chr.11: 34113754-34113754 on Build GRCh38
rs573278917 Chr.11: 34113550-34113550 on Build GRCh38
rs1048464232 Chr.11: 34113782-34113782 on Build GRCh38
rs369494970 Chr.11: 34113705-34113705 on Build GRCh38
rs778826697 Chr.11: 34113742-34113742 on Build GRCh38
rs746453112 Chr.11: 34113774-34113774 on Build GRCh38
rs34445882 Chr.11: 34113660-34113660 on Build GRCh38
rs763429728 Chr.11: 34113772-34113772 on Build GRCh38
rs749954562 Chr.11: 34113638-34113639 on Build GRCh38
rs754172923 Chr.11: 34113761-34113761 on Build GRCh38
rs1043550683 Chr.11: 34113662-34113662 on Build GRCh38
rs1056009959 Chr.11: 34113555-34113555 on Build GRCh38
rs954652050 Chr.11: 34113889-34113889 on Build GRCh38
rs74497402 Chr.11: 34113525-34113525 on Build GRCh38
rs558967047 Chr.11: 34113703-34113703 on Build GRCh38
rs896095651 Chr.11: 34113983-34113983 on Build GRCh38
rs553463004 Chr.11: 34113541-34113541 on Build GRCh38
rs894806046 Chr.11: 34113603-34113603 on Build GRCh38
rs757556635 Chr.11: 34113873-34113873 on Build GRCh38
rs879188404 Chr.11: 34113664-34113664 on Build GRCh38
rs185281502 Chr.11: 34113700-34113700 on Build GRCh38
rs750838209 Chr.11: 34113881-34113881 on Build GRCh38
rs971917018 Chr.11: 34113580-34113580 on Build GRCh38
rs747876146 Chr.11: 34113818-34113818 on Build GRCh38
rs2957483 Chr.11: 34113610-34113610 on Build GRCh38
rs776160620 Chr.11: 34113701-34113702 on Build GRCh38
rs906039045 Chr.11: 34113793-34113793 on Build GRCh38
rs75440493 Chr.11: 34113709-34113709 on Build GRCh38

View Full Product Details