Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10570413-10570919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL10
Entrez Gene ID
79954
Gene Name
nucleolar protein 10
Gene Aliases
PQBP5
UniGene
Hs.222494
Gene Chromosome Location
Chr.2: 10570766-10689991 on Build GRCh38
Transcripts
NM_001261392.1 NM_001261394.1 NM_024894.3 NR_048552.1

SNP Detail

SNP ID Chromosome Location
rs909451283 Chr.2: 10570506-10570506 on Build GRCh38
rs943641590 Chr.2: 10570551-10570551 on Build GRCh38
rs34632282 Chr.2: 10570683-10570684 on Build GRCh38
rs919344668 Chr.2: 10570461-10570461 on Build GRCh38
rs923525927 Chr.2: 10570791-10570791 on Build GRCh38
rs991400535 Chr.2: 10570638-10570638 on Build GRCh38
rs1039318416 Chr.2: 10570752-10570752 on Build GRCh38
rs777312578 Chr.2: 10570674-10570674 on Build GRCh38
rs778304339 Chr.2: 10570715-10570715 on Build GRCh38
rs62128621 Chr.2: 10570852-10570852 on Build GRCh38
rs371857151 Chr.2: 10570726-10570726 on Build GRCh38
rs78773114 Chr.2: 10570854-10570854 on Build GRCh38
rs190116186 Chr.2: 10570563-10570563 on Build GRCh38
rs375436962 Chr.2: 10570884-10570885 on Build GRCh38
rs755318752 Chr.2: 10570541-10570541 on Build GRCh38
rs1032614178 Chr.2: 10570607-10570607 on Build GRCh38
rs959535350 Chr.2: 10570637-10570637 on Build GRCh38
rs868731958 Chr.2: 10570850-10570850 on Build GRCh38
rs71392236 Chr.2: 10570682-10570683 on Build GRCh38
rs80006092 Chr.2: 10570841-10570841 on Build GRCh38
rs563724565 Chr.2: 10570667-10570667 on Build GRCh38
rs901920604 Chr.2: 10570777-10570777 on Build GRCh38
rs547248903 Chr.2: 10570763-10570763 on Build GRCh38
rs575996412 Chr.2: 10570889-10570889 on Build GRCh38
rs1007817358 Chr.2: 10570520-10570520 on Build GRCh38
rs144132018 Chr.2: 10570441-10570441 on Build GRCh38
rs1042119330 Chr.2: 10570584-10570584 on Build GRCh38
rs34895208 Chr.2: 10570841-10570841 on Build GRCh38
rs902306773 Chr.2: 10570596-10570596 on Build GRCh38
rs185884808 Chr.2: 10570779-10570779 on Build GRCh38
rs573506279 Chr.2: 10570578-10570578 on Build GRCh38
rs142209561 Chr.2: 10570684-10570685 on Build GRCh38
rs56138710 Chr.2: 10570621-10570621 on Build GRCh38
rs539338754 Chr.2: 10570811-10570811 on Build GRCh38
rs397869862 Chr.2: 10570852-10570852 on Build GRCh38
rs74560553 Chr.2: 10570673-10570673 on Build GRCh38
rs551365657 Chr.2: 10570653-10570653 on Build GRCh38
rs997515332 Chr.2: 10570569-10570569 on Build GRCh38
rs117574411 Chr.2: 10570582-10570582 on Build GRCh38
rs9287719 Chr.2: 10570604-10570604 on Build GRCh38
rs1033238532 Chr.2: 10570501-10570501 on Build GRCh38
rs959256387 Chr.2: 10570610-10570610 on Build GRCh38
rs539444930 Chr.2: 10570593-10570593 on Build GRCh38
rs950804163 Chr.2: 10570704-10570704 on Build GRCh38
rs397800700 Chr.2: 10570686-10570687 on Build GRCh38
rs919331409 Chr.2: 10570681-10570681 on Build GRCh38
rs997145040 Chr.2: 10570571-10570571 on Build GRCh38
rs893050263 Chr.2: 10570728-10570728 on Build GRCh38
rs985037067 Chr.2: 10570614-10570614 on Build GRCh38
rs543518321 Chr.2: 10570483-10570483 on Build GRCh38
rs2160257 Chr.2: 10570468-10570468 on Build GRCh38
rs984956974 Chr.2: 10570712-10570712 on Build GRCh38
rs1045357513 Chr.2: 10570682-10570682 on Build GRCh38
rs950667554 Chr.2: 10570876-10570876 on Build GRCh38
rs915660804 Chr.2: 10570652-10570652 on Build GRCh38
rs374501344 Chr.2: 10570455-10570455 on Build GRCh38
rs779547850 Chr.2: 10570735-10570735 on Build GRCh38
rs567806598 Chr.2: 10570737-10570737 on Build GRCh38
rs751763963 Chr.2: 10570528-10570528 on Build GRCh38
rs77324976 Chr.2: 10570851-10570852 on Build GRCh38
rs749784627 Chr.2: 10570716-10570716 on Build GRCh38
rs892945371 Chr.2: 10570509-10570509 on Build GRCh38
rs971350410 Chr.2: 10570661-10570661 on Build GRCh38
rs535112626 Chr.2: 10570759-10570759 on Build GRCh38

View Full Product Details