Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 96985741-96986264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXGR1
Entrez Gene ID
27199
Gene Name
oxoglutarate receptor 1
Gene Aliases
GPR80, GPR99, P2RY15, P2Y15, aKGR
UniGene
Hs.352218
Gene Chromosome Location
Chr.13: 96985719-96995842 on Build GRCh38
Transcripts
NM_080818.3 XM_005254043.3 XM_005254044.3 XM_005254045.2 XM_005254046.2 XM_011521077.2

SNP Detail

SNP ID Chromosome Location
rs144105178 Chr.13: 96985852-96985852 on Build GRCh38
rs1021782553 Chr.13: 96986159-96986159 on Build GRCh38
rs544645753 Chr.13: 96985951-96985951 on Build GRCh38
rs958281369 Chr.13: 96986048-96986048 on Build GRCh38
rs931116039 Chr.13: 96986096-96986096 on Build GRCh38
rs1009791500 Chr.13: 96986052-96986052 on Build GRCh38
rs985135152 Chr.13: 96986023-96986023 on Build GRCh38
rs1051839890 Chr.13: 96985945-96985945 on Build GRCh38
rs912252987 Chr.13: 96986030-96986030 on Build GRCh38
rs986118246 Chr.13: 96986200-96986200 on Build GRCh38
rs139275313 Chr.13: 96985802-96985802 on Build GRCh38
rs146502547 Chr.13: 96986154-96986154 on Build GRCh38
rs774198882 Chr.13: 96986232-96986232 on Build GRCh38
rs1007839063 Chr.13: 96985766-96985766 on Build GRCh38
rs965437404 Chr.13: 96986207-96986207 on Build GRCh38
rs775962322 Chr.13: 96985841-96985841 on Build GRCh38
rs970895202 Chr.13: 96986092-96986092 on Build GRCh38
rs1023423671 Chr.13: 96985878-96985878 on Build GRCh38
rs1032583446 Chr.13: 96985781-96985782 on Build GRCh38
rs564543279 Chr.13: 96986223-96986223 on Build GRCh38
rs895056975 Chr.13: 96986110-96986110 on Build GRCh38
rs956218910 Chr.13: 96985972-96985972 on Build GRCh38
rs61975469 Chr.13: 96986149-96986149 on Build GRCh38
rs995395300 Chr.13: 96985819-96985819 on Build GRCh38
rs777063389 Chr.13: 96986004-96986004 on Build GRCh38
rs188156899 Chr.13: 96986212-96986212 on Build GRCh38
rs61368531 Chr.13: 96986152-96986152 on Build GRCh38
rs565013867 Chr.13: 96985952-96985952 on Build GRCh38
rs939948347 Chr.13: 96986173-96986173 on Build GRCh38
rs889145692 Chr.13: 96985955-96985955 on Build GRCh38
rs1012909217 Chr.13: 96985856-96985856 on Build GRCh38
rs998026876 Chr.13: 96985938-96985941 on Build GRCh38
rs1048326308 Chr.13: 96985885-96985885 on Build GRCh38
rs1048288384 Chr.13: 96986103-96986103 on Build GRCh38
rs954316439 Chr.13: 96986172-96986172 on Build GRCh38
rs1032635995 Chr.13: 96985975-96985975 on Build GRCh38
rs141110769 Chr.13: 96986231-96986231 on Build GRCh38
rs1019250000 Chr.13: 96986202-96986202 on Build GRCh38
rs759106033 Chr.13: 96986239-96986239 on Build GRCh38
rs972491906 Chr.13: 96986077-96986077 on Build GRCh38
rs747617943 Chr.13: 96985853-96985853 on Build GRCh38
rs918328095 Chr.13: 96986215-96986215 on Build GRCh38
rs558969136 Chr.13: 96985892-96985892 on Build GRCh38
rs1002930030 Chr.13: 96986151-96986151 on Build GRCh38
rs988307251 Chr.13: 96985790-96985790 on Build GRCh38
rs559156362 Chr.13: 96985800-96985800 on Build GRCh38
rs900969555 Chr.13: 96985937-96985937 on Build GRCh38
rs368565222 Chr.13: 96986000-96986000 on Build GRCh38
rs1049884922 Chr.13: 96985773-96985773 on Build GRCh38
rs573632264 Chr.13: 96985814-96985814 on Build GRCh38
rs1011686691 Chr.13: 96986157-96986157 on Build GRCh38
rs943817614 Chr.13: 96985782-96985782 on Build GRCh38
rs918400157 Chr.13: 96986089-96986089 on Build GRCh38
rs914805205 Chr.13: 96986002-96986002 on Build GRCh38
rs990335993 Chr.13: 96985980-96985980 on Build GRCh38
rs931086515 Chr.13: 96986218-96986218 on Build GRCh38
rs529664015 Chr.13: 96986183-96986183 on Build GRCh38
rs138296283 Chr.13: 96986181-96986181 on Build GRCh38
rs374560961 Chr.13: 96985963-96985963 on Build GRCh38
rs948006068 Chr.13: 96986131-96986131 on Build GRCh38
rs909794357 Chr.13: 96985889-96985889 on Build GRCh38
rs971505066 Chr.13: 96985886-96985886 on Build GRCh38
rs370563470 Chr.13: 96985909-96985909 on Build GRCh38
rs58399642 Chr.13: 96985781-96985781 on Build GRCh38
rs1044366053 Chr.13: 96986148-96986148 on Build GRCh38
rs556713042 Chr.13: 96985809-96985809 on Build GRCh38
rs937671578 Chr.13: 96985863-96985864 on Build GRCh38
rs943769245 Chr.13: 96986043-96986043 on Build GRCh38

View Full Product Details