Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15717787-15718293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHM2
Entrez Gene ID
23207
Gene Name
pleckstrin homology and RUN domain containing M2
Gene Aliases
SKIP
UniGene
Hs.646775
Gene Chromosome Location
Chr.1: 15681506-15734769 on Build GRCh38
Transcripts
NM_015164.2 XM_005245790.3 XM_017000757.1 XM_017000758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
898307 c.361C>T p.H121Y Chr.1: 15717976-15717976 on Build GRCh38
3399894 c.614A>G p.N205S Chr.1: 15717920-15717920 on Build GRCh38
7230203 c.291G>A p.L97L Chr.1: 15717906-15717906 on Build GRCh38
458507 c.608C>T p.A203V Chr.1: 15717914-15717914 on Build GRCh38
4517319 c.623_624CC>TT p.S208F Chr.1: 15717929-15717930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572193627 Chr.1: 15718233-15718233 on Build GRCh38
rs757274431 Chr.1: 15717889-15717889 on Build GRCh38
rs750279086 Chr.1: 15717899-15717899 on Build GRCh38
rs959070620 Chr.1: 15717966-15717966 on Build GRCh38
rs755932538 Chr.1: 15718014-15718014 on Build GRCh38
rs1051563960 Chr.1: 15718160-15718160 on Build GRCh38
rs752943991 Chr.1: 15717866-15717866 on Build GRCh38
rs201895179 Chr.1: 15717885-15717885 on Build GRCh38
rs761621510 Chr.1: 15718004-15718004 on Build GRCh38
rs757587145 Chr.1: 15718224-15718224 on Build GRCh38
rs764064755 Chr.1: 15717988-15717988 on Build GRCh38
rs962543394 Chr.1: 15718269-15718269 on Build GRCh38
rs184127702 Chr.1: 15718030-15718030 on Build GRCh38
rs778821268 Chr.1: 15717921-15717921 on Build GRCh38
rs776837720 Chr.1: 15717948-15717948 on Build GRCh38
rs1042120145 Chr.1: 15718025-15718025 on Build GRCh38
rs771509554 Chr.1: 15717947-15717947 on Build GRCh38
rs891112167 Chr.1: 15718213-15718213 on Build GRCh38
rs750407849 Chr.1: 15718008-15718008 on Build GRCh38
rs751725867 Chr.1: 15717999-15717999 on Build GRCh38
rs772031528 Chr.1: 15718101-15718101 on Build GRCh38
rs997103031 Chr.1: 15718152-15718152 on Build GRCh38
rs201347347 Chr.1: 15717936-15717936 on Build GRCh38
rs137990328 Chr.1: 15718106-15718106 on Build GRCh38
rs759723833 Chr.1: 15717857-15717857 on Build GRCh38
rs371787019 Chr.1: 15717880-15717880 on Build GRCh38
rs926490908 Chr.1: 15717957-15717957 on Build GRCh38
rs1015462025 Chr.1: 15717931-15717931 on Build GRCh38
rs779998624 Chr.1: 15718019-15718019 on Build GRCh38
rs759850212 Chr.1: 15717959-15717959 on Build GRCh38
rs548387299 Chr.1: 15717946-15717946 on Build GRCh38
rs770179616 Chr.1: 15717960-15717960 on Build GRCh38
rs191383513 Chr.1: 15717876-15717876 on Build GRCh38
rs1029663243 Chr.1: 15717897-15717897 on Build GRCh38
rs765492018 Chr.1: 15717865-15717865 on Build GRCh38
rs747744183 Chr.1: 15718041-15718041 on Build GRCh38
rs940200079 Chr.1: 15717816-15717816 on Build GRCh38
rs570664661 Chr.1: 15718108-15718108 on Build GRCh38
rs753580481 Chr.1: 15717896-15717896 on Build GRCh38
rs1041238941 Chr.1: 15717848-15717848 on Build GRCh38
rs577719675 Chr.1: 15718151-15718151 on Build GRCh38
rs16851970 Chr.1: 15717906-15717906 on Build GRCh38
rs977316809 Chr.1: 15718210-15718210 on Build GRCh38
rs201114976 Chr.1: 15717972-15717972 on Build GRCh38
rs973996783 Chr.1: 15717954-15717954 on Build GRCh38
rs1000423741 Chr.1: 15717822-15717822 on Build GRCh38
rs755758203 Chr.1: 15717849-15717849 on Build GRCh38
rs936138289 Chr.1: 15718089-15718089 on Build GRCh38
rs747106992 Chr.1: 15718124-15718124 on Build GRCh38
rs767536149 Chr.1: 15718007-15718007 on Build GRCh38
rs779804256 Chr.1: 15717907-15717907 on Build GRCh38
rs753481809 Chr.1: 15718021-15718021 on Build GRCh38
rs538620256 Chr.1: 15718131-15718131 on Build GRCh38
rs187603649 Chr.1: 15718180-15718180 on Build GRCh38
rs748978895 Chr.1: 15717908-15717908 on Build GRCh38
rs762983914 Chr.1: 15717868-15717868 on Build GRCh38
rs763178454 Chr.1: 15717987-15717987 on Build GRCh38
rs778256654 Chr.1: 15718037-15718037 on Build GRCh38
rs539802473 Chr.1: 15718202-15718202 on Build GRCh38

View Full Product Details