Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27311310-27311809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POM121L2
Entrez Gene ID
94026
Gene Name
POM121 transmembrane nucleoporin like 2
Gene Aliases
POM121-L, POM121L
UniGene
Hs.729955
Gene Chromosome Location
Chr.6: 27309063-27312232 on Build GRCh38
Transcripts
NM_033482.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4003842 c.855A>G p.P285P Chr.6: 27311316-27311316 on Build GRCh38
6054909 c.840G>T p.E280D Chr.6: 27311331-27311331 on Build GRCh38
5405002 c.404C>T p.P135L Chr.6: 27311767-27311767 on Build GRCh38
1221541 c.815G>T p.R272I Chr.6: 27311356-27311356 on Build GRCh38
4153174 c.684G>T p.L228F Chr.6: 27311487-27311487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767174201 Chr.6: 27311787-27311787 on Build GRCh38
rs760743347 Chr.6: 27311430-27311431 on Build GRCh38
rs773777808 Chr.6: 27311389-27311389 on Build GRCh38
rs992063981 Chr.6: 27311575-27311575 on Build GRCh38
rs763870440 Chr.6: 27311541-27311541 on Build GRCh38
rs975482457 Chr.6: 27311476-27311476 on Build GRCh38
rs984307030 Chr.6: 27311643-27311643 on Build GRCh38
rs766783664 Chr.6: 27311470-27311470 on Build GRCh38
rs766526203 Chr.6: 27311766-27311768 on Build GRCh38
rs777811535 Chr.6: 27311684-27311684 on Build GRCh38
rs369413879 Chr.6: 27311601-27311601 on Build GRCh38
rs772076557 Chr.6: 27311379-27311379 on Build GRCh38
rs533929055 Chr.6: 27311728-27311728 on Build GRCh38
rs937556866 Chr.6: 27311574-27311574 on Build GRCh38
rs779116411 Chr.6: 27311623-27311623 on Build GRCh38
rs149082261 Chr.6: 27311556-27311556 on Build GRCh38
rs16897553 Chr.6: 27311406-27311406 on Build GRCh38
rs202012145 Chr.6: 27311724-27311724 on Build GRCh38
rs754667540 Chr.6: 27311595-27311595 on Build GRCh38
rs761481095 Chr.6: 27311473-27311473 on Build GRCh38
rs767120845 Chr.6: 27311479-27311479 on Build GRCh38
rs753685622 Chr.6: 27311587-27311587 on Build GRCh38
rs952553995 Chr.6: 27311482-27311482 on Build GRCh38
rs1028606861 Chr.6: 27311521-27311521 on Build GRCh38
rs776784833 Chr.6: 27311542-27311542 on Build GRCh38
rs375550978 Chr.6: 27311640-27311640 on Build GRCh38
rs756813620 Chr.6: 27311335-27311337 on Build GRCh38
rs774080147 Chr.6: 27311472-27311472 on Build GRCh38
rs1003942952 Chr.6: 27311746-27311746 on Build GRCh38
rs997077310 Chr.6: 27311334-27311334 on Build GRCh38
rs142149892 Chr.6: 27311676-27311676 on Build GRCh38
rs747504039 Chr.6: 27311397-27311397 on Build GRCh38
rs568499246 Chr.6: 27311688-27311688 on Build GRCh38
rs778949486 Chr.6: 27311351-27311351 on Build GRCh38
rs961452007 Chr.6: 27311681-27311681 on Build GRCh38
rs928051923 Chr.6: 27311721-27311721 on Build GRCh38
rs761533908 Chr.6: 27311743-27311743 on Build GRCh38
rs943966790 Chr.6: 27311665-27311665 on Build GRCh38
rs770048144 Chr.6: 27311733-27311733 on Build GRCh38
rs574797263 Chr.6: 27311734-27311734 on Build GRCh38
rs902954688 Chr.6: 27311337-27311337 on Build GRCh38
rs375438522 Chr.6: 27311430-27311430 on Build GRCh38
rs367780723 Chr.6: 27311530-27311530 on Build GRCh38
rs748244122 Chr.6: 27311367-27311367 on Build GRCh38
rs756044102 Chr.6: 27311555-27311555 on Build GRCh38
rs981649668 Chr.6: 27311566-27311566 on Build GRCh38
rs201661976 Chr.6: 27311702-27311702 on Build GRCh38
rs777024577 Chr.6: 27311440-27311440 on Build GRCh38
rs915834575 Chr.6: 27311762-27311762 on Build GRCh38
rs914686008 Chr.6: 27311621-27311621 on Build GRCh38
rs928801406 Chr.6: 27311567-27311567 on Build GRCh38
rs759779931 Chr.6: 27311453-27311453 on Build GRCh38

View Full Product Details