Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 179102240-179102739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SESTD1
Entrez Gene ID
91404
Gene Name
SEC14 and spectrin domain containing 1
Gene Aliases
SOLO
UniGene
Hs.30977
Gene Chromosome Location
Chr.2: 179101692-179264723 on Build GRCh38
Transcripts
NM_178123.4 XM_011512139.2 XM_011512141.2 XM_017005250.1 XM_017005251.1

SNP Detail

SNP ID Chromosome Location
rs910375150 Chr.2: 179102408-179102408 on Build GRCh38
rs3180635 Chr.2: 179102519-179102519 on Build GRCh38
rs552834500 Chr.2: 179102607-179102607 on Build GRCh38
rs1003029808 Chr.2: 179102303-179102303 on Build GRCh38
rs186156933 Chr.2: 179102653-179102653 on Build GRCh38
rs867281531 Chr.2: 179102277-179102277 on Build GRCh38
rs993203300 Chr.2: 179102634-179102634 on Build GRCh38
rs530922640 Chr.2: 179102667-179102667 on Build GRCh38
rs986394990 Chr.2: 179102402-179102402 on Build GRCh38
rs542543070 Chr.2: 179102500-179102500 on Build GRCh38
rs958723140 Chr.2: 179102322-179102322 on Build GRCh38
rs571257772 Chr.2: 179102346-179102346 on Build GRCh38
rs1029985865 Chr.2: 179102637-179102637 on Build GRCh38
rs73973203 Chr.2: 179102484-179102484 on Build GRCh38
rs989682415 Chr.2: 179102709-179102709 on Build GRCh38
rs564999659 Chr.2: 179102480-179102492 on Build GRCh38
rs990266020 Chr.2: 179102470-179102470 on Build GRCh38
rs894656095 Chr.2: 179102575-179102575 on Build GRCh38
rs1016022877 Chr.2: 179102614-179102614 on Build GRCh38
rs946056499 Chr.2: 179102473-179102473 on Build GRCh38
rs914616999 Chr.2: 179102472-179102472 on Build GRCh38
rs890552537 Chr.2: 179102444-179102444 on Build GRCh38
rs957897271 Chr.2: 179102671-179102671 on Build GRCh38
rs572744739 Chr.2: 179102624-179102624 on Build GRCh38
rs539225609 Chr.2: 179102283-179102283 on Build GRCh38
rs971024188 Chr.2: 179102290-179102290 on Build GRCh38
rs1012771940 Chr.2: 179102686-179102686 on Build GRCh38
rs954641242 Chr.2: 179102650-179102650 on Build GRCh38
rs758131497 Chr.2: 179102388-179102388 on Build GRCh38
rs936475517 Chr.2: 179102413-179102413 on Build GRCh38
rs111276240 Chr.2: 179102381-179102381 on Build GRCh38
rs1006762534 Chr.2: 179102587-179102587 on Build GRCh38
rs914691972 Chr.2: 179102713-179102713 on Build GRCh38
rs1036374123 Chr.2: 179102559-179102559 on Build GRCh38
rs907503775 Chr.2: 179102494-179102494 on Build GRCh38
rs1041953546 Chr.2: 179102281-179102281 on Build GRCh38
rs141837306 Chr.2: 179102388-179102388 on Build GRCh38
rs544662739 Chr.2: 179102696-179102696 on Build GRCh38
rs1050456605 Chr.2: 179102422-179102422 on Build GRCh38
rs76285846 Chr.2: 179102282-179102282 on Build GRCh38
rs573553638 Chr.2: 179102376-179102376 on Build GRCh38
rs565048231 Chr.2: 179102656-179102656 on Build GRCh38
rs1056394890 Chr.2: 179102501-179102501 on Build GRCh38
rs556201214 Chr.2: 179102436-179102438 on Build GRCh38
rs1005956572 Chr.2: 179102338-179102338 on Build GRCh38
rs1041690868 Chr.2: 179102488-179102488 on Build GRCh38
rs557880595 Chr.2: 179102314-179102314 on Build GRCh38
rs556700077 Chr.2: 179102361-179102361 on Build GRCh38
rs537109065 Chr.2: 179102347-179102347 on Build GRCh38

View Full Product Details