Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 179102579-179103115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SESTD1
Entrez Gene ID
91404
Gene Name
SEC14 and spectrin domain containing 1
Gene Aliases
SOLO
UniGene
Hs.30977
Gene Chromosome Location
Chr.2: 179101692-179264723 on Build GRCh38
Transcripts
NM_178123.4 XM_011512139.2 XM_011512141.2 XM_017005250.1 XM_017005251.1

SNP Detail

SNP ID Chromosome Location
rs139968033 Chr.2: 179103079-179103079 on Build GRCh38
rs1007595448 Chr.2: 179102956-179102956 on Build GRCh38
rs547356316 Chr.2: 179102828-179102828 on Build GRCh38
rs377530867 Chr.2: 179102904-179102904 on Build GRCh38
rs944899681 Chr.2: 179103057-179103057 on Build GRCh38
rs552834500 Chr.2: 179102607-179102607 on Build GRCh38
rs553905273 Chr.2: 179102766-179102766 on Build GRCh38
rs536770521 Chr.2: 179102962-179102962 on Build GRCh38
rs894685795 Chr.2: 179102788-179102788 on Build GRCh38
rs977488496 Chr.2: 179103009-179103009 on Build GRCh38
rs186156933 Chr.2: 179102653-179102653 on Build GRCh38
rs143518419 Chr.2: 179103081-179103081 on Build GRCh38
rs942248009 Chr.2: 179102823-179102823 on Build GRCh38
rs993203300 Chr.2: 179102634-179102634 on Build GRCh38
rs938652915 Chr.2: 179102782-179102782 on Build GRCh38
rs530922640 Chr.2: 179102667-179102667 on Build GRCh38
rs928625634 Chr.2: 179102754-179102754 on Build GRCh38
rs567167563 Chr.2: 179102837-179102837 on Build GRCh38
rs1036893188 Chr.2: 179102867-179102868 on Build GRCh38
rs577722727 Chr.2: 179102805-179102805 on Build GRCh38
rs190570572 Chr.2: 179102916-179102916 on Build GRCh38
rs993235891 Chr.2: 179102910-179102910 on Build GRCh38
rs897807150 Chr.2: 179102876-179102876 on Build GRCh38
rs912059876 Chr.2: 179103056-179103057 on Build GRCh38
rs953631862 Chr.2: 179103042-179103042 on Build GRCh38
rs930669667 Chr.2: 179102888-179102888 on Build GRCh38
rs1029985865 Chr.2: 179102637-179102637 on Build GRCh38
rs998600439 Chr.2: 179102980-179102980 on Build GRCh38
rs894416462 Chr.2: 179102941-179102944 on Build GRCh38
rs1049535793 Chr.2: 179103086-179103089 on Build GRCh38
rs144089242 Chr.2: 179103010-179103010 on Build GRCh38
rs916204786 Chr.2: 179103068-179103068 on Build GRCh38
rs989682415 Chr.2: 179102709-179102709 on Build GRCh38
rs566959948 Chr.2: 179102990-179102990 on Build GRCh38
rs1024150516 Chr.2: 179102957-179102957 on Build GRCh38
rs77116194 Chr.2: 179102995-179102995 on Build GRCh38
rs1016022877 Chr.2: 179102614-179102614 on Build GRCh38
rs989729426 Chr.2: 179102778-179102778 on Build GRCh38
rs532807018 Chr.2: 179102909-179102909 on Build GRCh38
rs911943393 Chr.2: 179102845-179102845 on Build GRCh38
rs555845581 Chr.2: 179102737-179102737 on Build GRCh38
rs957897271 Chr.2: 179102671-179102671 on Build GRCh38
rs915507708 Chr.2: 179102784-179102784 on Build GRCh38
rs572744739 Chr.2: 179102624-179102624 on Build GRCh38
rs991892091 Chr.2: 179103055-179103055 on Build GRCh38
rs897603616 Chr.2: 179102848-179102848 on Build GRCh38
rs1022520567 Chr.2: 179103012-179103012 on Build GRCh38
rs558041632 Chr.2: 179102737-179102738 on Build GRCh38
rs530099658 Chr.2: 179102804-179102804 on Build GRCh38
rs1017527782 Chr.2: 179102960-179102960 on Build GRCh38
rs1012771940 Chr.2: 179102686-179102686 on Build GRCh38
rs561152854 Chr.2: 179102729-179102729 on Build GRCh38
rs954641242 Chr.2: 179102650-179102650 on Build GRCh38
rs181503234 Chr.2: 179103004-179103004 on Build GRCh38
rs914691972 Chr.2: 179102713-179102713 on Build GRCh38
rs989366679 Chr.2: 179102989-179102989 on Build GRCh38
rs1056053615 Chr.2: 179102785-179102785 on Build GRCh38
rs572807758 Chr.2: 179103065-179103065 on Build GRCh38
rs544662739 Chr.2: 179102696-179102696 on Build GRCh38
rs149510999 Chr.2: 179102968-179102968 on Build GRCh38
rs565048231 Chr.2: 179102656-179102656 on Build GRCh38
rs368470526 Chr.2: 179102771-179102771 on Build GRCh38
rs960076742 Chr.2: 179103049-179103049 on Build GRCh38
rs944785304 Chr.2: 179102847-179102847 on Build GRCh38
rs569400015 Chr.2: 179102946-179102946 on Build GRCh38

View Full Product Details