Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 64063165-64063704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCEA2
Entrez Gene ID
6919
Gene Name
transcription elongation factor A2
Gene Aliases
TFIIS
UniGene
Hs.505004
Gene Chromosome Location
Chr.20: 64042350-64072347 on Build GRCh38
Transcripts
NM_003195.4 NM_198723.1 XM_005260229.3 XM_011529022.1 XM_011529023.1 XM_011529027.2 XM_017028036.1 XM_017028037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1228699 c.34G>A p.A12T Chr.20: 64063346-64063346 on Build GRCh38
1029230 c.72G>A p.A24A Chr.20: 64063384-64063384 on Build GRCh38
3693669 c.38G>A p.R13Q Chr.20: 64063350-64063350 on Build GRCh38
3693668 c.38G>A p.R13Q Chr.20: 64063350-64063350 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1016869470 Chr.20: 64063210-64063210 on Build GRCh38
rs546241273 Chr.20: 64063375-64063375 on Build GRCh38
rs1045693419 Chr.20: 64063636-64063636 on Build GRCh38
rs923245808 Chr.20: 64063521-64063521 on Build GRCh38
rs943355141 Chr.20: 64063295-64063295 on Build GRCh38
rs577857417 Chr.20: 64063260-64063260 on Build GRCh38
rs934397352 Chr.20: 64063496-64063496 on Build GRCh38
rs916185304 Chr.20: 64063435-64063435 on Build GRCh38
rs1045811412 Chr.20: 64063466-64063466 on Build GRCh38
rs965012568 Chr.20: 64063516-64063516 on Build GRCh38
rs894366735 Chr.20: 64063593-64063593 on Build GRCh38
rs551389504 Chr.20: 64063538-64063538 on Build GRCh38
rs375173974 Chr.20: 64063376-64063376 on Build GRCh38
rs974005777 Chr.20: 64063536-64063536 on Build GRCh38
rs919472251 Chr.20: 64063183-64063183 on Build GRCh38
rs753832094 Chr.20: 64063414-64063414 on Build GRCh38
rs916126304 Chr.20: 64063674-64063674 on Build GRCh38
rs1011382009 Chr.20: 64063590-64063590 on Build GRCh38
rs878997428 Chr.20: 64063226-64063226 on Build GRCh38
rs906846053 Chr.20: 64063646-64063646 on Build GRCh38
rs73149297 Chr.20: 64063659-64063659 on Build GRCh38
rs540313866 Chr.20: 64063322-64063322 on Build GRCh38
rs370712578 Chr.20: 64063311-64063311 on Build GRCh38
rs998577758 Chr.20: 64063448-64063448 on Build GRCh38
rs763486482 Chr.20: 64063407-64063407 on Build GRCh38
rs564047778 Chr.20: 64063249-64063249 on Build GRCh38
rs985024564 Chr.20: 64063423-64063423 on Build GRCh38
rs757144862 Chr.20: 64063416-64063416 on Build GRCh38
rs764156502 Chr.20: 64063265-64063265 on Build GRCh38
rs972545349 Chr.20: 64063652-64063652 on Build GRCh38
rs757632854 Chr.20: 64063272-64063273 on Build GRCh38
rs547831911 Chr.20: 64063625-64063625 on Build GRCh38
rs111374609 Chr.20: 64063633-64063633 on Build GRCh38
rs759280400 Chr.20: 64063381-64063381 on Build GRCh38
rs909308667 Chr.20: 64063566-64063566 on Build GRCh38
rs753684729 Chr.20: 64063266-64063266 on Build GRCh38
rs1005386710 Chr.20: 64063397-64063397 on Build GRCh38
rs747262347 Chr.20: 64063290-64063290 on Build GRCh38
rs930881251 Chr.20: 64063184-64063184 on Build GRCh38
rs910789578 Chr.20: 64063498-64063498 on Build GRCh38
rs375782497 Chr.20: 64063666-64063666 on Build GRCh38
rs1038661193 Chr.20: 64063398-64063398 on Build GRCh38
rs952168444 Chr.20: 64063664-64063664 on Build GRCh38
rs749763855 Chr.20: 64063354-64063354 on Build GRCh38
rs199825146 Chr.20: 64063392-64063392 on Build GRCh38
rs534076179 Chr.20: 64063581-64063581 on Build GRCh38
rs774793941 Chr.20: 64063380-64063380 on Build GRCh38
rs769731700 Chr.20: 64063349-64063349 on Build GRCh38
rs1012612908 Chr.20: 64063242-64063242 on Build GRCh38
rs954549661 Chr.20: 64063235-64063235 on Build GRCh38
rs1028218756 Chr.20: 64063289-64063289 on Build GRCh38
rs991178867 Chr.20: 64063245-64063245 on Build GRCh38
rs923337740 Chr.20: 64063302-64063302 on Build GRCh38
rs961450102 Chr.20: 64063432-64063432 on Build GRCh38
rs544257745 Chr.20: 64063205-64063205 on Build GRCh38
rs773820158 Chr.20: 64063351-64063351 on Build GRCh38
rs1028166221 Chr.20: 64063598-64063598 on Build GRCh38
rs906954494 Chr.20: 64063250-64063250 on Build GRCh38
rs1052899849 Chr.20: 64063551-64063551 on Build GRCh38
rs1018996274 Chr.20: 64063362-64063362 on Build GRCh38
rs532806116 Chr.20: 64063194-64063194 on Build GRCh38
rs976475159 Chr.20: 64063520-64063520 on Build GRCh38
rs775004137 Chr.20: 64063393-64063393 on Build GRCh38
rs757089273 Chr.20: 64063275-64063275 on Build GRCh38
rs1007520950 Chr.20: 64063623-64063623 on Build GRCh38
rs567768026 Chr.20: 64063647-64063647 on Build GRCh38
rs929326116 Chr.20: 64063527-64063527 on Build GRCh38
rs1011432878 Chr.20: 64063286-64063286 on Build GRCh38
rs971125787 Chr.20: 64063431-64063431 on Build GRCh38
rs966108182 Chr.20: 64063642-64063642 on Build GRCh38
rs998658934 Chr.20: 64063252-64063252 on Build GRCh38
rs957541806 Chr.20: 64063264-64063264 on Build GRCh38
rs1024001908 Chr.20: 64063426-64063426 on Build GRCh38
rs748316892 Chr.20: 64063348-64063348 on Build GRCh38
rs562409180 Chr.20: 64063443-64063443 on Build GRCh38
rs201308894 Chr.20: 64063400-64063400 on Build GRCh38
rs906907466 Chr.20: 64063493-64063493 on Build GRCh38
rs531488114 Chr.20: 64063500-64063500 on Build GRCh38
rs1047814683 Chr.20: 64063346-64063346 on Build GRCh38
rs919936453 Chr.20: 64063549-64063549 on Build GRCh38
rs887012919 Chr.20: 64063193-64063193 on Build GRCh38
rs1047765409 Chr.20: 64063530-64063530 on Build GRCh38
rs986552338 Chr.20: 64063292-64063292 on Build GRCh38
rs983266221 Chr.20: 64063296-64063296 on Build GRCh38
rs571304688 Chr.20: 64063573-64063573 on Build GRCh38
rs893004092 Chr.20: 64063263-64063263 on Build GRCh38
rs957734299 Chr.20: 64063470-64063470 on Build GRCh38
rs778626284 Chr.20: 64063288-64063288 on Build GRCh38
rs948378724 Chr.20: 64063455-64063455 on Build GRCh38
rs919417819 Chr.20: 64063383-64063383 on Build GRCh38
rs894133399 Chr.20: 64063220-64063220 on Build GRCh38
rs889710023 Chr.20: 64063607-64063607 on Build GRCh38
rs1012996414 Chr.20: 64063402-64063402 on Build GRCh38
rs977569537 Chr.20: 64063257-64063257 on Build GRCh38

View Full Product Details