Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9676341-9676884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP43
Entrez Gene ID
124739
Gene Name
ubiquitin specific peptidase 43
Gene Aliases
-
UniGene
Hs.709621
Gene Chromosome Location
Chr.17: 9643493-9729687 on Build GRCh38
Transcripts
NM_001267576.1 NM_153210.4 XM_011523639.2 XM_011523640.1 XM_011523641.1 XM_011523642.2 XM_017024159.1 XM_017024160.1 XM_017024161.1 XM_017024162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5722546 c.887T>C p.V296A Chr.17: 9676799-9676799 on Build GRCh38
6897237 c.907C>T p.L303F Chr.17: 9676819-9676819 on Build GRCh38
5387787 c.958C>T p.P320S Chr.17: 9676870-9676870 on Build GRCh38
6523244 c.937G>A p.V313I Chr.17: 9676849-9676849 on Build GRCh38
5779848 c.875G>A p.R292Q Chr.17: 9676787-9676787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939728185 Chr.17: 9676833-9676833 on Build GRCh38
rs779050623 Chr.17: 9676697-9676697 on Build GRCh38
rs751312963 Chr.17: 9676802-9676802 on Build GRCh38
rs897979431 Chr.17: 9676551-9676551 on Build GRCh38
rs957757870 Chr.17: 9676640-9676640 on Build GRCh38
rs372807819 Chr.17: 9676758-9676758 on Build GRCh38
rs552504507 Chr.17: 9676518-9676518 on Build GRCh38
rs547944149 Chr.17: 9676592-9676592 on Build GRCh38
rs988611852 Chr.17: 9676663-9676663 on Build GRCh38
rs546979401 Chr.17: 9676771-9676771 on Build GRCh38
rs534758213 Chr.17: 9676678-9676678 on Build GRCh38
rs1013791672 Chr.17: 9676575-9676575 on Build GRCh38
rs770988998 Chr.17: 9676755-9676755 on Build GRCh38
rs1050832696 Chr.17: 9676469-9676469 on Build GRCh38
rs552940108 Chr.17: 9676604-9676604 on Build GRCh38
rs913726073 Chr.17: 9676419-9676419 on Build GRCh38
rs1037783910 Chr.17: 9676504-9676504 on Build GRCh38
rs571152378 Chr.17: 9676669-9676669 on Build GRCh38
rs932596847 Chr.17: 9676448-9676448 on Build GRCh38
rs373029742 Chr.17: 9676473-9676473 on Build GRCh38
rs201400428 Chr.17: 9676724-9676724 on Build GRCh38
rs376268405 Chr.17: 9676591-9676591 on Build GRCh38
rs777902138 Chr.17: 9676737-9676737 on Build GRCh38
rs531105872 Chr.17: 9676603-9676603 on Build GRCh38
rs921099843 Chr.17: 9676742-9676742 on Build GRCh38
rs943896365 Chr.17: 9676472-9676472 on Build GRCh38
rs748562301 Chr.17: 9676455-9676455 on Build GRCh38
rs932589381 Chr.17: 9676489-9676489 on Build GRCh38
rs766338318 Chr.17: 9676392-9676392 on Build GRCh38
rs761456862 Chr.17: 9676786-9676786 on Build GRCh38
rs576803088 Chr.17: 9676382-9676382 on Build GRCh38
rs757370893 Chr.17: 9676827-9676827 on Build GRCh38
rs375002344 Chr.17: 9676796-9676796 on Build GRCh38
rs910552121 Chr.17: 9676809-9676809 on Build GRCh38
rs945228188 Chr.17: 9676428-9676428 on Build GRCh38
rs772596055 Chr.17: 9676773-9676773 on Build GRCh38
rs368390814 Chr.17: 9676770-9676770 on Build GRCh38
rs371524163 Chr.17: 9676815-9676815 on Build GRCh38
rs563894498 Chr.17: 9676594-9676594 on Build GRCh38
rs749493365 Chr.17: 9676822-9676822 on Build GRCh38
rs770013453 Chr.17: 9676734-9676734 on Build GRCh38
rs753537186 Chr.17: 9676784-9676784 on Build GRCh38
rs529804834 Chr.17: 9676526-9676526 on Build GRCh38
rs746765850 Chr.17: 9676729-9676729 on Build GRCh38
rs371533171 Chr.17: 9676787-9676787 on Build GRCh38
rs368889332 Chr.17: 9676810-9676810 on Build GRCh38
rs749909327 Chr.17: 9676788-9676788 on Build GRCh38
rs199643890 Chr.17: 9676795-9676795 on Build GRCh38
rs989269812 Chr.17: 9676409-9676409 on Build GRCh38
rs778894555 Chr.17: 9676847-9676847 on Build GRCh38
rs921150732 Chr.17: 9676486-9676486 on Build GRCh38
rs564235218 Chr.17: 9676563-9676563 on Build GRCh38
rs958102122 Chr.17: 9676620-9676620 on Build GRCh38
rs1021066476 Chr.17: 9676687-9676687 on Build GRCh38
rs541017066 Chr.17: 9676493-9676493 on Build GRCh38
rs758602407 Chr.17: 9676705-9676705 on Build GRCh38
rs758290247 Chr.17: 9676789-9676789 on Build GRCh38
rs1020758163 Chr.17: 9676677-9676677 on Build GRCh38
rs1040899083 Chr.17: 9676429-9676429 on Build GRCh38
rs749434141 Chr.17: 9676746-9676746 on Build GRCh38
rs763845413 Chr.17: 9676782-9676782 on Build GRCh38
rs113165431 Chr.17: 9676497-9676497 on Build GRCh38
rs1048495359 Chr.17: 9676496-9676496 on Build GRCh38
rs775740636 Chr.17: 9676781-9676781 on Build GRCh38
rs1025163342 Chr.17: 9676589-9676589 on Build GRCh38
rs988275403 Chr.17: 9676436-9676436 on Build GRCh38
rs965446209 Chr.17: 9676696-9676696 on Build GRCh38

View Full Product Details