Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101884318-101884836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMAT1
Entrez Gene ID
84460
Gene Name
zinc finger matrin-type 1
Gene Aliases
-
UniGene
Hs.496512
Gene Chromosome Location
Chr.X: 101882288-101932135 on Build GRCh38
Transcripts
NM_001011657.3 NM_001282400.1 NM_001282401.1 XM_005262212.4 XM_005262213.4 XM_005262214.3 XM_005262216.4 XM_006724711.3 XM_017029903.1 XM_017029904.1

SNP Detail

SNP ID Chromosome Location
rs140202441 Chr.X: 101884649-101884649 on Build GRCh38
rs866887461 Chr.X: 101884372-101884372 on Build GRCh38
rs745816021 Chr.X: 101884559-101884559 on Build GRCh38
rs367837983 Chr.X: 101884628-101884628 on Build GRCh38
rs765131277 Chr.X: 101884378-101884378 on Build GRCh38
rs1026126400 Chr.X: 101884396-101884396 on Build GRCh38
rs939639420 Chr.X: 101884600-101884600 on Build GRCh38
rs368723231 Chr.X: 101884456-101884456 on Build GRCh38
rs964383934 Chr.X: 101884558-101884558 on Build GRCh38
rs371120505 Chr.X: 101884491-101884491 on Build GRCh38
rs376911023 Chr.X: 101884788-101884788 on Build GRCh38
rs768967766 Chr.X: 101884579-101884579 on Build GRCh38
rs767498025 Chr.X: 101884586-101884586 on Build GRCh38
rs768367195 Chr.X: 101884731-101884731 on Build GRCh38
rs777856893 Chr.X: 101884587-101884587 on Build GRCh38
rs17854380 Chr.X: 101884474-101884474 on Build GRCh38
rs751180162 Chr.X: 101884634-101884634 on Build GRCh38
rs754374119 Chr.X: 101884790-101884790 on Build GRCh38
rs761952893 Chr.X: 101884780-101884780 on Build GRCh38
rs142044704 Chr.X: 101884678-101884678 on Build GRCh38
rs113362805 Chr.X: 101884682-101884682 on Build GRCh38
rs151148157 Chr.X: 101884783-101884783 on Build GRCh38
rs774455122 Chr.X: 101884583-101884583 on Build GRCh38
rs757258463 Chr.X: 101884415-101884415 on Build GRCh38
rs889987288 Chr.X: 101884782-101884782 on Build GRCh38
rs755320591 Chr.X: 101884599-101884599 on Build GRCh38
rs371282679 Chr.X: 101884386-101884386 on Build GRCh38
rs201119309 Chr.X: 101884595-101884595 on Build GRCh38
rs890291236 Chr.X: 101884807-101884807 on Build GRCh38
rs1008806303 Chr.X: 101884469-101884469 on Build GRCh38
rs375125973 Chr.X: 101884686-101884686 on Build GRCh38
rs868279178 Chr.X: 101884624-101884624 on Build GRCh38
rs777269094 Chr.X: 101884515-101884515 on Build GRCh38
rs17282855 Chr.X: 101884786-101884786 on Build GRCh38
rs749196440 Chr.X: 101884706-101884706 on Build GRCh38
rs769889978 Chr.X: 101884670-101884670 on Build GRCh38
rs774127795 Chr.X: 101884754-101884754 on Build GRCh38
rs770382734 Chr.X: 101884543-101884543 on Build GRCh38
rs772185967 Chr.X: 101884628-101884629 on Build GRCh38
rs750958196 Chr.X: 101884419-101884419 on Build GRCh38
rs994196586 Chr.X: 101884741-101884741 on Build GRCh38
rs201830609 Chr.X: 101884750-101884750 on Build GRCh38
rs897268760 Chr.X: 101884687-101884687 on Build GRCh38
rs754251317 Chr.X: 101884596-101884596 on Build GRCh38
rs759509729 Chr.X: 101884366-101884366 on Build GRCh38
rs868738165 Chr.X: 101884531-101884531 on Build GRCh38
rs954814900 Chr.X: 101884398-101884398 on Build GRCh38
rs865847204 Chr.X: 101884574-101884574 on Build GRCh38
rs762054523 Chr.X: 101884585-101884585 on Build GRCh38
rs374675037 Chr.X: 101884555-101884555 on Build GRCh38
rs866003436 Chr.X: 101884352-101884352 on Build GRCh38
rs756879503 Chr.X: 101884635-101884635 on Build GRCh38
rs866507414 Chr.X: 101884355-101884355 on Build GRCh38
rs775489730 Chr.X: 101884571-101884571 on Build GRCh38
rs772266798 Chr.X: 101884781-101884781 on Build GRCh38
rs372023196 Chr.X: 101884650-101884650 on Build GRCh38
rs769843157 Chr.X: 101884563-101884563 on Build GRCh38
rs147524708 Chr.X: 101884594-101884594 on Build GRCh38
rs139010664 Chr.X: 101884466-101884466 on Build GRCh38
rs374330713 Chr.X: 101884407-101884407 on Build GRCh38
rs756546061 Chr.X: 101884420-101884420 on Build GRCh38
rs201873364 Chr.X: 101884418-101884418 on Build GRCh38
rs1049816808 Chr.X: 101884734-101884734 on Build GRCh38
rs746624461 Chr.X: 101884537-101884537 on Build GRCh38
rs376194900 Chr.X: 101884353-101884353 on Build GRCh38
rs200366802 Chr.X: 101884464-101884464 on Build GRCh38
rs77527361 Chr.X: 101884669-101884669 on Build GRCh38
rs751697393 Chr.X: 101884413-101884413 on Build GRCh38
rs763321835 Chr.X: 101884391-101884391 on Build GRCh38

View Full Product Details