Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212857684-212858191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLVCR1-AS1
Entrez Gene ID
642946
Gene Name
FLVCR1 antisense RNA 1 (head to head)
Gene Aliases
LQK1, NCRNA00292
UniGene
Hs.552649
Gene Chromosome Location
Chr.1: 212856604-212858138 on Build GRCh38
Transcripts
NR_027285.1 NR_027286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6289316 c.76C>T p.R26* Chr.1: 212857991-212857991 on Build GRCh38
6284323 c.136A>T p.T46S Chr.1: 212857931-212857931 on Build GRCh38
3746780 c.59G>A p.C20Y Chr.1: 212858008-212858008 on Build GRCh38
3746779 c.251G>A p.S84N Chr.1: 212857816-212857816 on Build GRCh38
1601629 c.173A>G p.Y58C Chr.1: 212857894-212857894 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs959843724 Chr.1: 212858057-212858057 on Build GRCh38
rs1057226211 Chr.1: 212858169-212858169 on Build GRCh38
rs2201602 Chr.1: 212858131-212858131 on Build GRCh38
rs997691020 Chr.1: 212857714-212857714 on Build GRCh38
rs563627182 Chr.1: 212858065-212858066 on Build GRCh38
rs555061212 Chr.1: 212858021-212858021 on Build GRCh38
rs910435445 Chr.1: 212857950-212857950 on Build GRCh38
rs932964305 Chr.1: 212857927-212857927 on Build GRCh38
rs1053284742 Chr.1: 212857994-212857994 on Build GRCh38
rs535311869 Chr.1: 212857839-212857839 on Build GRCh38
rs1035487170 Chr.1: 212857838-212857838 on Build GRCh38
rs933512473 Chr.1: 212857970-212857970 on Build GRCh38
rs964372054 Chr.1: 212858139-212858139 on Build GRCh38
rs1022321912 Chr.1: 212858074-212858074 on Build GRCh38
rs1012017976 Chr.1: 212857790-212857790 on Build GRCh38
rs574950324 Chr.1: 212858025-212858025 on Build GRCh38
rs41296686 Chr.1: 212858141-212858141 on Build GRCh38
rs3768554 Chr.1: 212858066-212858066 on Build GRCh38
rs1002838078 Chr.1: 212858050-212858050 on Build GRCh38
rs959894637 Chr.1: 212857843-212857843 on Build GRCh38
rs549502236 Chr.1: 212857924-212857924 on Build GRCh38
rs1043348269 Chr.1: 212858145-212858145 on Build GRCh38
rs994031844 Chr.1: 212858065-212858065 on Build GRCh38
rs1031920405 Chr.1: 212857728-212857728 on Build GRCh38
rs867687748 Chr.1: 212857954-212857954 on Build GRCh38
rs752380873 Chr.1: 212857929-212857929 on Build GRCh38
rs572645343 Chr.1: 212858067-212858067 on Build GRCh38
rs905870933 Chr.1: 212858020-212858020 on Build GRCh38
rs910846815 Chr.1: 212858100-212858100 on Build GRCh38
rs887191035 Chr.1: 212858040-212858040 on Build GRCh38
rs922382924 Chr.1: 212858109-212858109 on Build GRCh38
rs924368653 Chr.1: 212857903-212857903 on Build GRCh38
rs1047881 Chr.1: 212857816-212857816 on Build GRCh38
rs1047880 Chr.1: 212857822-212857822 on Build GRCh38
rs569340106 Chr.1: 212857928-212857928 on Build GRCh38
rs754833101 Chr.1: 212857786-212857786 on Build GRCh38
rs140978949 Chr.1: 212857793-212857793 on Build GRCh38
rs1011847317 Chr.1: 212858004-212858004 on Build GRCh38
rs980213408 Chr.1: 212857830-212857830 on Build GRCh38
rs531752151 Chr.1: 212858125-212858125 on Build GRCh38
rs958949986 Chr.1: 212857818-212857818 on Build GRCh38
rs529044833 Chr.1: 212858154-212858154 on Build GRCh38
rs963783024 Chr.1: 212858105-212858105 on Build GRCh38
rs899028449 Chr.1: 212857711-212857711 on Build GRCh38
rs12404942 Chr.1: 212858122-212858122 on Build GRCh38
rs892011621 Chr.1: 212857999-212857999 on Build GRCh38
rs923435587 Chr.1: 212857962-212857962 on Build GRCh38
rs539761646 Chr.1: 212857879-212857879 on Build GRCh38
rs947592568 Chr.1: 212858138-212858138 on Build GRCh38
rs551357554 Chr.1: 212857881-212857882 on Build GRCh38
rs561119655 Chr.1: 212857772-212857772 on Build GRCh38
rs565846406 Chr.1: 212857856-212857856 on Build GRCh38
rs866011374 Chr.1: 212858054-212858054 on Build GRCh38
rs542063223 Chr.1: 212858037-212858037 on Build GRCh38
rs895875259 Chr.1: 212858170-212858170 on Build GRCh38
rs748753532 Chr.1: 212857795-212857795 on Build GRCh38
rs183323430 Chr.1: 212857850-212857850 on Build GRCh38
rs918531481 Chr.1: 212857877-212857877 on Build GRCh38
rs984060570 Chr.1: 212858072-212858072 on Build GRCh38
rs1001560179 Chr.1: 212858163-212858163 on Build GRCh38
rs2201601 Chr.1: 212858106-212858106 on Build GRCh38
rs41296684 Chr.1: 212858066-212858067 on Build GRCh38
rs930982108 Chr.1: 212857908-212857908 on Build GRCh38
rs550331981 Chr.1: 212857796-212857796 on Build GRCh38
rs1001610927 Chr.1: 212858032-212858032 on Build GRCh38
rs751513759 Chr.1: 212857723-212857723 on Build GRCh38
rs969315149 Chr.1: 212857862-212857862 on Build GRCh38
rs2220949 Chr.1: 212858008-212858008 on Build GRCh38
rs1040782221 Chr.1: 212857955-212857955 on Build GRCh38

View Full Product Details